Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46860564-46860730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYL3
Entrez Gene ID
4634
Gene Name
myosin light chain 3
Gene Aliases
CMH8, MLC-lV/sb, MLC1SB, MLC1V, VLC1, VLCl
UniGene
Hs.517939
Gene Chromosome Location
Chr.3: 46857867-46863483 on Build GRCh38
Transcripts
NM_000258.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6410579 c.271G>A p.E91K Chr.3: 46860712-46860712 on Build GRCh38
4977900 c.253C>T p.Q85* Chr.3: 46860730-46860730 on Build GRCh38
3594591 c.286C>T p.L96L Chr.3: 46860697-46860697 on Build GRCh38
4453275 c.273A>G p.E91E Chr.3: 46860710-46860710 on Build GRCh38
347264 c.284T>A p.V95D Chr.3: 46860699-46860699 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs533243785 Chr.3: 46860591-46860591 on Build GRCh38
rs769503169 Chr.3: 46860689-46860689 on Build GRCh38
rs937808134 Chr.3: 46860613-46860613 on Build GRCh38
rs759754750 Chr.3: 46860660-46860660 on Build GRCh38
rs113993487 Chr.3: 46860675-46860675 on Build GRCh38
rs770961856 Chr.3: 46860709-46860709 on Build GRCh38
rs730880961 Chr.3: 46860703-46860703 on Build GRCh38
rs561570934 Chr.3: 46860590-46860590 on Build GRCh38
rs187668225 Chr.3: 46860593-46860593 on Build GRCh38
rs193217136 Chr.3: 46860612-46860612 on Build GRCh38
rs200593962 Chr.3: 46860620-46860620 on Build GRCh38
rs768956093 Chr.3: 46860671-46860671 on Build GRCh38
rs540550450 Chr.3: 46860636-46860636 on Build GRCh38
rs887145234 Chr.3: 46860708-46860708 on Build GRCh38
rs200526230 Chr.3: 46860633-46860633 on Build GRCh38
rs933664929 Chr.3: 46860588-46860588 on Build GRCh38
rs747691982 Chr.3: 46860682-46860682 on Build GRCh38
rs375395589 Chr.3: 46860672-46860672 on Build GRCh38
rs764356863 Chr.3: 46860637-46860637 on Build GRCh38
rs528471341 Chr.3: 46860635-46860635 on Build GRCh38
rs745968655 Chr.3: 46860648-46860648 on Build GRCh38
rs908313192 Chr.3: 46860592-46860592 on Build GRCh38
rs992388957 Chr.3: 46860601-46860601 on Build GRCh38
rs778410173 Chr.3: 46860670-46860670 on Build GRCh38
rs1056327125 Chr.3: 46860614-46860614 on Build GRCh38
rs2227294 Chr.3: 46860639-46860639 on Build GRCh38
rs112679764 Chr.3: 46860674-46860674 on Build GRCh38
rs730880957 Chr.3: 46860691-46860691 on Build GRCh38
rs762930148 Chr.3: 46860626-46860626 on Build GRCh38
rs184025552 Chr.3: 46860661-46860661 on Build GRCh38
rs947785437 Chr.3: 46860625-46860625 on Build GRCh38
rs772804040 Chr.3: 46860697-46860697 on Build GRCh38
rs960596932 Chr.3: 46860600-46860600 on Build GRCh38
rs905044253 Chr.3: 46860607-46860607 on Build GRCh38
rs754068239 Chr.3: 46860638-46860638 on Build GRCh38
rs199474703 Chr.3: 46860702-46860702 on Build GRCh38
rs778800102 Chr.3: 46860640-46860640 on Build GRCh38
rs376902603 Chr.3: 46860653-46860653 on Build GRCh38

View Full Product Details