Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53869782-53870183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR8
Entrez Gene ID
93973
Gene Name
ARP8 actin-related protein 8 homolog
Gene Aliases
ARP8, INO80N, hArp8
UniGene
Hs.412186
Gene Chromosome Location
Chr.3: 53863096-53882202 on Build GRCh38
Transcripts
NM_022899.4 XM_005265587.4 XM_011534249.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1047337 c.1695A>G p.R565R Chr.3: 53870018-53870018 on Build GRCh38
5018783 c.701delA p.K234fs*8 Chr.3: 53870127-53870127 on Build GRCh38
4119369 c.1643A>G p.Q548R Chr.3: 53870070-53870070 on Build GRCh38
4119370 c.758A>G p.Q253R Chr.3: 53870070-53870070 on Build GRCh38
5941197 c.1666C>T p.L556F Chr.3: 53870047-53870047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902429602 Chr.3: 53870140-53870140 on Build GRCh38
rs202219088 Chr.3: 53869966-53869966 on Build GRCh38
rs775870210 Chr.3: 53870045-53870045 on Build GRCh38
rs749997941 Chr.3: 53869964-53869964 on Build GRCh38
rs984446356 Chr.3: 53870011-53870011 on Build GRCh38
rs765836786 Chr.3: 53870107-53870107 on Build GRCh38
rs370209492 Chr.3: 53870158-53870158 on Build GRCh38
rs144617747 Chr.3: 53870027-53870027 on Build GRCh38
rs778077976 Chr.3: 53870139-53870139 on Build GRCh38
rs923894366 Chr.3: 53870141-53870141 on Build GRCh38
rs765554366 Chr.3: 53869938-53869938 on Build GRCh38
rs962316506 Chr.3: 53869842-53869842 on Build GRCh38
rs754914579 Chr.3: 53870126-53870126 on Build GRCh38
rs967311752 Chr.3: 53869815-53869815 on Build GRCh38
rs1042240474 Chr.3: 53870087-53870087 on Build GRCh38
rs754760494 Chr.3: 53869978-53869978 on Build GRCh38
rs74431192 Chr.3: 53869961-53869961 on Build GRCh38
rs756802645 Chr.3: 53870122-53870124 on Build GRCh38
rs957465704 Chr.3: 53869836-53869836 on Build GRCh38
rs1013479331 Chr.3: 53869805-53869805 on Build GRCh38
rs878973516 Chr.3: 53869901-53869901 on Build GRCh38
rs954987069 Chr.3: 53869925-53869925 on Build GRCh38
rs3833971 Chr.3: 53869854-53869854 on Build GRCh38
rs567747585 Chr.3: 53869942-53869942 on Build GRCh38
rs1015615202 Chr.3: 53869828-53869828 on Build GRCh38
rs761448050 Chr.3: 53870053-53870053 on Build GRCh38
rs556876582 Chr.3: 53869970-53869970 on Build GRCh38
rs754206571 Chr.3: 53869995-53869996 on Build GRCh38
rs747269929 Chr.3: 53870145-53870145 on Build GRCh38
rs373952185 Chr.3: 53870035-53870035 on Build GRCh38
rs778593714 Chr.3: 53869981-53869981 on Build GRCh38
rs949244723 Chr.3: 53870071-53870071 on Build GRCh38
rs397876190 Chr.3: 53869857-53869857 on Build GRCh38
rs748351305 Chr.3: 53869982-53869982 on Build GRCh38
rs369760477 Chr.3: 53869977-53869977 on Build GRCh38
rs761499842 Chr.3: 53869952-53869952 on Build GRCh38
rs767210588 Chr.3: 53870056-53870056 on Build GRCh38
rs761603656 Chr.3: 53869866-53869866 on Build GRCh38
rs985225206 Chr.3: 53869959-53869959 on Build GRCh38
rs1033008608 Chr.3: 53869835-53869835 on Build GRCh38
rs753759066 Chr.3: 53870120-53870120 on Build GRCh38
rs776381292 Chr.3: 53869816-53869816 on Build GRCh38
rs547789778 Chr.3: 53870020-53870020 on Build GRCh38
rs929582502 Chr.3: 53870002-53870002 on Build GRCh38
rs770170566 Chr.3: 53870044-53870044 on Build GRCh38
rs765120831 Chr.3: 53870127-53870127 on Build GRCh38
rs770924197 Chr.3: 53870024-53870024 on Build GRCh38
rs758761400 Chr.3: 53869991-53869991 on Build GRCh38
rs747032805 Chr.3: 53870016-53870016 on Build GRCh38
rs376058740 Chr.3: 53870079-53870079 on Build GRCh38
rs763591788 Chr.3: 53869810-53869810 on Build GRCh38
rs777986914 Chr.3: 53869997-53869997 on Build GRCh38
rs986645483 Chr.3: 53869881-53869881 on Build GRCh38
rs576722191 Chr.3: 53870138-53870138 on Build GRCh38
rs760163851 Chr.3: 53870089-53870089 on Build GRCh38
rs761373340 Chr.3: 53870148-53870148 on Build GRCh38
rs112256523 Chr.3: 53869817-53869817 on Build GRCh38
rs757313466 Chr.3: 53870149-53870149 on Build GRCh38

View Full Product Details