Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157477218-157477741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM19
Entrez Gene ID
8728
Gene Name
ADAM metallopeptidase domain 19
Gene Aliases
FKSG34, MADDAM, MLTNB
UniGene
Hs.483944
Gene Chromosome Location
Chr.5: 157477304-157575823 on Build GRCh38
Transcripts
NM_033274.4 XM_011534682.2 XM_017010009.1

SNP Detail

SNP ID Chromosome Location
rs972345938 Chr.5: 157477544-157477544 on Build GRCh38
rs192546051 Chr.5: 157477534-157477534 on Build GRCh38
rs187591027 Chr.5: 157477641-157477641 on Build GRCh38
rs1033068987 Chr.5: 157477502-157477502 on Build GRCh38
rs1047377869 Chr.5: 157477442-157477442 on Build GRCh38
rs148264060 Chr.5: 157477536-157477536 on Build GRCh38
rs963940382 Chr.5: 157477542-157477542 on Build GRCh38
rs528473264 Chr.5: 157477256-157477256 on Build GRCh38
rs920364473 Chr.5: 157477328-157477328 on Build GRCh38
rs1010315475 Chr.5: 157477702-157477702 on Build GRCh38
rs942880910 Chr.5: 157477309-157477309 on Build GRCh38
rs985820415 Chr.5: 157477537-157477537 on Build GRCh38
rs534745939 Chr.5: 157477669-157477669 on Build GRCh38
rs894472420 Chr.5: 157477412-157477412 on Build GRCh38
rs911384851 Chr.5: 157477294-157477294 on Build GRCh38
rs1038721408 Chr.5: 157477312-157477312 on Build GRCh38
rs938895671 Chr.5: 157477587-157477587 on Build GRCh38
rs11466825 Chr.5: 157477668-157477668 on Build GRCh38
rs141359105 Chr.5: 157477682-157477682 on Build GRCh38
rs892304721 Chr.5: 157477521-157477521 on Build GRCh38
rs565369281 Chr.5: 157477522-157477522 on Build GRCh38
rs1045712378 Chr.5: 157477449-157477449 on Build GRCh38
rs1000803133 Chr.5: 157477476-157477476 on Build GRCh38
rs913443794 Chr.5: 157477270-157477270 on Build GRCh38
rs949800 Chr.5: 157477626-157477626 on Build GRCh38
rs1001370569 Chr.5: 157477461-157477461 on Build GRCh38
rs1054275054 Chr.5: 157477681-157477681 on Build GRCh38
rs955595350 Chr.5: 157477535-157477535 on Build GRCh38
rs747191765 Chr.5: 157477302-157477302 on Build GRCh38
rs531427988 Chr.5: 157477265-157477265 on Build GRCh38
rs920122508 Chr.5: 157477589-157477589 on Build GRCh38
rs771733139 Chr.5: 157477621-157477621 on Build GRCh38
rs760291663 Chr.5: 157477652-157477652 on Build GRCh38
rs547164724 Chr.5: 157477511-157477511 on Build GRCh38
rs1045636638 Chr.5: 157477632-157477632 on Build GRCh38
rs11466826 Chr.5: 157477282-157477282 on Build GRCh38
rs866819025 Chr.5: 157477711-157477711 on Build GRCh38
rs10404 Chr.5: 157477625-157477625 on Build GRCh38
rs948762631 Chr.5: 157477435-157477435 on Build GRCh38
rs184355470 Chr.5: 157477614-157477614 on Build GRCh38
rs974226131 Chr.5: 157477588-157477588 on Build GRCh38
rs964157927 Chr.5: 157477555-157477555 on Build GRCh38
rs886127625 Chr.5: 157477447-157477447 on Build GRCh38
rs771673931 Chr.5: 157477654-157477654 on Build GRCh38
rs1018485282 Chr.5: 157477546-157477546 on Build GRCh38
rs993320607 Chr.5: 157477611-157477611 on Build GRCh38
rs1030071164 Chr.5: 157477689-157477689 on Build GRCh38
rs1052372829 Chr.5: 157477635-157477635 on Build GRCh38
rs919450636 Chr.5: 157477261-157477261 on Build GRCh38
rs987500673 Chr.5: 157477289-157477289 on Build GRCh38
rs997381622 Chr.5: 157477672-157477672 on Build GRCh38
rs1031230577 Chr.5: 157477486-157477486 on Build GRCh38
rs189011531 Chr.5: 157477448-157477448 on Build GRCh38
rs569717943 Chr.5: 157477434-157477434 on Build GRCh38
rs11134764 Chr.5: 157477242-157477242 on Build GRCh38
rs905658760 Chr.5: 157477450-157477450 on Build GRCh38

View Full Product Details