Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 64550985-64551528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS9
Entrez Gene ID
56999
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 9
Gene Aliases
-
UniGene
Hs.656071
Gene Chromosome Location
Chr.3: 64515654-64688000 on Build GRCh38
Transcripts
NM_001318781.1 NM_182920.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4549369 c.4717G>A p.E1573K Chr.3: 64551044-64551044 on Build GRCh38
460741 c.4753G>C p.D1585H Chr.3: 64551008-64551008 on Build GRCh38
3597013 c.4725C>T p.S1575S Chr.3: 64551036-64551036 on Build GRCh38
4005613 c.4711T>A p.C1571S Chr.3: 64551050-64551050 on Build GRCh38
6469075 c.4743G>T p.V1581V Chr.3: 64551018-64551018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951703873 Chr.3: 64551242-64551242 on Build GRCh38
rs377567025 Chr.3: 64551505-64551505 on Build GRCh38
rs572157368 Chr.3: 64551227-64551227 on Build GRCh38
rs761235278 Chr.3: 64551246-64551246 on Build GRCh38
rs146650362 Chr.3: 64551096-64551096 on Build GRCh38
rs73832311 Chr.3: 64551142-64551142 on Build GRCh38
rs756083396 Chr.3: 64551498-64551498 on Build GRCh38
rs767711432 Chr.3: 64551300-64551300 on Build GRCh38
rs965823060 Chr.3: 64551456-64551456 on Build GRCh38
rs575698374 Chr.3: 64551259-64551259 on Build GRCh38
rs530344787 Chr.3: 64551413-64551413 on Build GRCh38
rs975947162 Chr.3: 64551291-64551291 on Build GRCh38
rs966520084 Chr.3: 64551491-64551491 on Build GRCh38
rs531161189 Chr.3: 64551335-64551335 on Build GRCh38
rs951752412 Chr.3: 64551444-64551444 on Build GRCh38
rs766280038 Chr.3: 64551502-64551502 on Build GRCh38
rs566687448 Chr.3: 64551430-64551430 on Build GRCh38
rs376962279 Chr.3: 64551045-64551045 on Build GRCh38
rs757829104 Chr.3: 64551110-64551110 on Build GRCh38
rs372066084 Chr.3: 64551028-64551028 on Build GRCh38
rs1808944 Chr.3: 64551431-64551431 on Build GRCh38
rs139133767 Chr.3: 64551251-64551251 on Build GRCh38
rs945239455 Chr.3: 64551138-64551138 on Build GRCh38
rs748394099 Chr.3: 64551118-64551119 on Build GRCh38
rs995745400 Chr.3: 64551427-64551427 on Build GRCh38
rs369848708 Chr.3: 64551035-64551035 on Build GRCh38
rs1055652527 Chr.3: 64551359-64551359 on Build GRCh38
rs112774460 Chr.3: 64551489-64551489 on Build GRCh38
rs564092982 Chr.3: 64551323-64551323 on Build GRCh38
rs764801357 Chr.3: 64551079-64551079 on Build GRCh38
rs371010899 Chr.3: 64551113-64551113 on Build GRCh38
rs373804459 Chr.3: 64551095-64551095 on Build GRCh38
rs373075580 Chr.3: 64551073-64551073 on Build GRCh38
rs1044464065 Chr.3: 64551219-64551220 on Build GRCh38
rs752355567 Chr.3: 64551083-64551083 on Build GRCh38
rs1000456870 Chr.3: 64551015-64551015 on Build GRCh38
rs763391336 Chr.3: 64551129-64551129 on Build GRCh38
rs892032361 Chr.3: 64551267-64551267 on Build GRCh38
rs17071010 Chr.3: 64551026-64551026 on Build GRCh38
rs1027359040 Chr.3: 64551347-64551347 on Build GRCh38
rs948796742 Chr.3: 64551205-64551206 on Build GRCh38
rs138988394 Chr.3: 64551023-64551023 on Build GRCh38
rs964819799 Chr.3: 64551286-64551286 on Build GRCh38
rs374674971 Chr.3: 64551212-64551213 on Build GRCh38
rs751220780 Chr.3: 64551240-64551240 on Build GRCh38
rs1013747117 Chr.3: 64551196-64551196 on Build GRCh38
rs751978590 Chr.3: 64551102-64551102 on Build GRCh38
rs766996805 Chr.3: 64551358-64551358 on Build GRCh38
rs868002868 Chr.3: 64551310-64551310 on Build GRCh38
rs746064998 Chr.3: 64551037-64551037 on Build GRCh38
rs574618439 Chr.3: 64551116-64551116 on Build GRCh38
rs773279281 Chr.3: 64551302-64551302 on Build GRCh38
rs775861579 Chr.3: 64551044-64551044 on Build GRCh38
rs190803077 Chr.3: 64551062-64551062 on Build GRCh38
rs183574302 Chr.3: 64551132-64551132 on Build GRCh38
rs374660210 Chr.3: 64551042-64551042 on Build GRCh38
rs146412036 Chr.3: 64551018-64551018 on Build GRCh38
rs796717865 Chr.3: 64551500-64551500 on Build GRCh38
rs760711807 Chr.3: 64551063-64551063 on Build GRCh38
rs896687517 Chr.3: 64551174-64551174 on Build GRCh38
rs1017652462 Chr.3: 64551452-64551452 on Build GRCh38
rs771961128 Chr.3: 64551047-64551047 on Build GRCh38
rs548477784 Chr.3: 64551426-64551426 on Build GRCh38
rs1027292348 Chr.3: 64551219-64551219 on Build GRCh38
rs995694874 Chr.3: 64551218-64551218 on Build GRCh38
rs926016320 Chr.3: 64551483-64551483 on Build GRCh38
rs763898356 Chr.3: 64551158-64551158 on Build GRCh38
rs570482398 Chr.3: 64551064-64551064 on Build GRCh38
rs1044538461 Chr.3: 64551480-64551480 on Build GRCh38
rs1002301289 Chr.3: 64551152-64551152 on Build GRCh38
rs368887757 Chr.3: 64551048-64551048 on Build GRCh38
rs1040989975 Chr.3: 64551501-64551501 on Build GRCh38
rs1056282526 Chr.3: 64551178-64551178 on Build GRCh38
rs896577207 Chr.3: 64551367-64551367 on Build GRCh38
rs745957099 Chr.3: 64551071-64551072 on Build GRCh38
rs903832201 Chr.3: 64551146-64551146 on Build GRCh38
rs1007216947 Chr.3: 64551445-64551445 on Build GRCh38
rs563036613 Chr.3: 64551354-64551354 on Build GRCh38
rs966304496 Chr.3: 64551262-64551262 on Build GRCh38
rs113761233 Chr.3: 64551396-64551396 on Build GRCh38
rs1014047778 Chr.3: 64551400-64551400 on Build GRCh38
rs1016144077 Chr.3: 64551200-64551200 on Build GRCh38
rs370501725 Chr.3: 64551066-64551066 on Build GRCh38
rs1019128935 Chr.3: 64551278-64551278 on Build GRCh38
rs1009501836 Chr.3: 64551273-64551273 on Build GRCh38
rs961609136 Chr.3: 64551212-64551212 on Build GRCh38
rs544930184 Chr.3: 64551336-64551336 on Build GRCh38
rs976213303 Chr.3: 64551461-64551461 on Build GRCh38

View Full Product Details