Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114335835-114336369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKNA
Entrez Gene ID
80709
Gene Name
AT-hook transcription factor
Gene Aliases
-
UniGene
Hs.494895
Gene Chromosome Location
Chr.9: 114334149-114398498 on Build GRCh38
Transcripts
NM_001317950.1 NM_001317952.1 NM_030767.5 XM_005252244.2 XM_005252245.1 XM_005252247.4 XM_005252248.2 XM_006717294.1 XM_011519063.2 XM_011519065.2 XM_017015172.1

SNP Detail

SNP ID Chromosome Location
rs1038938646 Chr.9: 114336194-114336194 on Build GRCh38
rs759035245 Chr.9: 114336124-114336124 on Build GRCh38
rs780286273 Chr.9: 114336314-114336314 on Build GRCh38
rs776034771 Chr.9: 114336123-114336123 on Build GRCh38
rs929064797 Chr.9: 114336132-114336132 on Build GRCh38
rs531955393 Chr.9: 114336099-114336099 on Build GRCh38
rs1028473602 Chr.9: 114335964-114335964 on Build GRCh38
rs957675807 Chr.9: 114335944-114335944 on Build GRCh38
rs1013432050 Chr.9: 114335921-114335921 on Build GRCh38
rs943157821 Chr.9: 114336335-114336335 on Build GRCh38
rs539245194 Chr.9: 114336340-114336340 on Build GRCh38
rs752722831 Chr.9: 114336283-114336283 on Build GRCh38
rs41278653 Chr.9: 114336297-114336297 on Build GRCh38
rs541816085 Chr.9: 114336209-114336209 on Build GRCh38
rs893157342 Chr.9: 114336210-114336210 on Build GRCh38
rs957039645 Chr.9: 114336246-114336246 on Build GRCh38
rs560631918 Chr.9: 114335940-114335940 on Build GRCh38
rs980977524 Chr.9: 114336033-114336033 on Build GRCh38
rs1049408008 Chr.9: 114336138-114336138 on Build GRCh38
rs764519125 Chr.9: 114336139-114336139 on Build GRCh38
rs961916628 Chr.9: 114336295-114336295 on Build GRCh38
rs2636869 Chr.9: 114336321-114336321 on Build GRCh38
rs954155825 Chr.9: 114336271-114336271 on Build GRCh38
rs766933781 Chr.9: 114336205-114336205 on Build GRCh38
rs926305632 Chr.9: 114336066-114336066 on Build GRCh38
rs915404068 Chr.9: 114336003-114336003 on Build GRCh38
rs760369446 Chr.9: 114335987-114335987 on Build GRCh38
rs964024250 Chr.9: 114335874-114335874 on Build GRCh38
rs912821896 Chr.9: 114336041-114336041 on Build GRCh38
rs563174865 Chr.9: 114336071-114336071 on Build GRCh38
rs77907581 Chr.9: 114336167-114336167 on Build GRCh38
rs916385368 Chr.9: 114336134-114336134 on Build GRCh38
rs949175662 Chr.9: 114336342-114336342 on Build GRCh38
rs887590836 Chr.9: 114336154-114336154 on Build GRCh38
rs926840280 Chr.9: 114336296-114336296 on Build GRCh38
rs146496585 Chr.9: 114336193-114336193 on Build GRCh38
rs1019814908 Chr.9: 114336292-114336292 on Build GRCh38
rs537287626 Chr.9: 114336328-114336328 on Build GRCh38
rs368509608 Chr.9: 114336316-114336316 on Build GRCh38
rs750254577 Chr.9: 114336148-114336150 on Build GRCh38
rs750799869 Chr.9: 114336284-114336284 on Build GRCh38
rs779918963 Chr.9: 114336150-114336154 on Build GRCh38
rs968296112 Chr.9: 114336026-114336026 on Build GRCh38
rs372706280 Chr.9: 114336245-114336245 on Build GRCh38
rs907166086 Chr.9: 114335932-114335932 on Build GRCh38
rs1048790044 Chr.9: 114336322-114336322 on Build GRCh38
rs552860450 Chr.9: 114335984-114335984 on Build GRCh38
rs1024860040 Chr.9: 114335924-114335924 on Build GRCh38
rs954208759 Chr.9: 114335967-114335967 on Build GRCh38
rs574852485 Chr.9: 114335859-114335859 on Build GRCh38
rs1046205524 Chr.9: 114336343-114336343 on Build GRCh38
rs987018471 Chr.9: 114336019-114336019 on Build GRCh38
rs548154377 Chr.9: 114336115-114336115 on Build GRCh38
rs981008684 Chr.9: 114336294-114336294 on Build GRCh38
rs555814653 Chr.9: 114336339-114336339 on Build GRCh38
rs992271064 Chr.9: 114336303-114336303 on Build GRCh38
rs868786888 Chr.9: 114336195-114336195 on Build GRCh38
rs540390909 Chr.9: 114335939-114335939 on Build GRCh38
rs1011640456 Chr.9: 114336235-114336235 on Build GRCh38
rs1029754775 Chr.9: 114336219-114336219 on Build GRCh38
rs532765128 Chr.9: 114335978-114335978 on Build GRCh38
rs1055149450 Chr.9: 114335867-114335867 on Build GRCh38
rs988680656 Chr.9: 114335955-114335955 on Build GRCh38
rs999481323 Chr.9: 114336200-114336200 on Build GRCh38
rs976844451 Chr.9: 114335915-114335915 on Build GRCh38
rs768055622 Chr.9: 114336280-114336280 on Build GRCh38

View Full Product Details