Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109507456-109507960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMIGO1
Entrez Gene ID
57463
Gene Name
adhesion molecule with Ig like domain 1
Gene Aliases
ALI2, AMIGO, AMIGO-1
UniGene
Hs.726479
Gene Chromosome Location
Chr.1: 109504178-109509742 on Build GRCh38
Transcripts
NM_020703.2 XM_011541812.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1294678 c.1452G>A p.S484S Chr.1: 109507461-109507461 on Build GRCh38
3965691 c.1182C>T p.T394T Chr.1: 109507731-109507731 on Build GRCh38
4020368 c.1200C>A p.C400* Chr.1: 109507713-109507713 on Build GRCh38
5923488 c.1228C>T p.Q410* Chr.1: 109507685-109507685 on Build GRCh38
1688229 c.1028C>T p.A343V Chr.1: 109507885-109507885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780340228 Chr.1: 109507616-109507616 on Build GRCh38
rs770895028 Chr.1: 109507769-109507769 on Build GRCh38
rs759291273 Chr.1: 109507594-109507594 on Build GRCh38
rs775205886 Chr.1: 109507813-109507813 on Build GRCh38
rs530990414 Chr.1: 109507821-109507821 on Build GRCh38
rs149872562 Chr.1: 109507628-109507628 on Build GRCh38
rs769194902 Chr.1: 109507805-109507805 on Build GRCh38
rs369716753 Chr.1: 109507677-109507677 on Build GRCh38
rs760117977 Chr.1: 109507590-109507590 on Build GRCh38
rs779037602 Chr.1: 109507618-109507618 on Build GRCh38
rs757429542 Chr.1: 109507913-109507913 on Build GRCh38
rs766915531 Chr.1: 109507498-109507498 on Build GRCh38
rs772008087 Chr.1: 109507584-109507584 on Build GRCh38
rs750952983 Chr.1: 109507697-109507697 on Build GRCh38
rs756258730 Chr.1: 109507614-109507614 on Build GRCh38
rs765964759 Chr.1: 109507507-109507507 on Build GRCh38
rs1001422869 Chr.1: 109507542-109507542 on Build GRCh38
rs764343078 Chr.1: 109507595-109507595 on Build GRCh38
rs757591436 Chr.1: 109507598-109507598 on Build GRCh38
rs781021614 Chr.1: 109507790-109507790 on Build GRCh38
rs545255768 Chr.1: 109507882-109507882 on Build GRCh38
rs149188606 Chr.1: 109507708-109507708 on Build GRCh38
rs768061906 Chr.1: 109507691-109507691 on Build GRCh38
rs770410594 Chr.1: 109507642-109507642 on Build GRCh38
rs201103189 Chr.1: 109507712-109507712 on Build GRCh38
rs758181107 Chr.1: 109507742-109507742 on Build GRCh38
rs771865852 Chr.1: 109507625-109507625 on Build GRCh38
rs191722490 Chr.1: 109507506-109507506 on Build GRCh38
rs765297638 Chr.1: 109507908-109507908 on Build GRCh38
rs745901445 Chr.1: 109507551-109507551 on Build GRCh38
rs750971402 Chr.1: 109507613-109507613 on Build GRCh38
rs772542670 Chr.1: 109507836-109507836 on Build GRCh38
rs758623374 Chr.1: 109507527-109507527 on Build GRCh38
rs550801997 Chr.1: 109507847-109507847 on Build GRCh38
rs368837535 Chr.1: 109507826-109507826 on Build GRCh38
rs754317749 Chr.1: 109507893-109507893 on Build GRCh38
rs761156076 Chr.1: 109507869-109507869 on Build GRCh38
rs150743133 Chr.1: 109507558-109507558 on Build GRCh38
rs914949225 Chr.1: 109507703-109507703 on Build GRCh38
rs141877739 Chr.1: 109507683-109507683 on Build GRCh38
rs776166736 Chr.1: 109507649-109507649 on Build GRCh38
rs774251914 Chr.1: 109507577-109507577 on Build GRCh38
rs745333176 Chr.1: 109507799-109507799 on Build GRCh38
rs772750261 Chr.1: 109507501-109507501 on Build GRCh38
rs746927511 Chr.1: 109507755-109507755 on Build GRCh38
rs1005277760 Chr.1: 109507489-109507489 on Build GRCh38
rs773153721 Chr.1: 109507588-109507588 on Build GRCh38
rs753500867 Chr.1: 109507509-109507509 on Build GRCh38
rs865773833 Chr.1: 109507827-109507827 on Build GRCh38
rs776257575 Chr.1: 109507592-109507592 on Build GRCh38
rs751993737 Chr.1: 109507597-109507597 on Build GRCh38
rs752947718 Chr.1: 109507914-109507914 on Build GRCh38
rs55654518 Chr.1: 109507685-109507685 on Build GRCh38
rs923670714 Chr.1: 109507901-109507901 on Build GRCh38
rs572344278 Chr.1: 109507566-109507566 on Build GRCh38
rs761868421 Chr.1: 109507482-109507482 on Build GRCh38
rs780360711 Chr.1: 109507559-109507559 on Build GRCh38
rs781387041 Chr.1: 109507550-109507550 on Build GRCh38
rs777598886 Chr.1: 109507749-109507749 on Build GRCh38
rs766539441 Chr.1: 109507711-109507711 on Build GRCh38
rs201494815 Chr.1: 109507497-109507497 on Build GRCh38
rs974948294 Chr.1: 109507728-109507728 on Build GRCh38
rs17854055 Chr.1: 109507552-109507552 on Build GRCh38
rs546135330 Chr.1: 109507885-109507885 on Build GRCh38
rs768081948 Chr.1: 109507599-109507599 on Build GRCh38
rs908036887 Chr.1: 109507905-109507905 on Build GRCh38
rs759078803 Chr.1: 109507653-109507653 on Build GRCh38
rs764585983 Chr.1: 109507529-109507529 on Build GRCh38
rs749453465 Chr.1: 109507617-109507617 on Build GRCh38
rs769446069 Chr.1: 109507657-109507657 on Build GRCh38
rs757828757 Chr.1: 109507533-109507533 on Build GRCh38
rs747011267 Chr.1: 109507634-109507634 on Build GRCh38
rs765874668 Chr.1: 109507591-109507591 on Build GRCh38
rs779136835 Chr.1: 109507723-109507723 on Build GRCh38
rs747929337 Chr.1: 109507623-109507623 on Build GRCh38
rs139892756 Chr.1: 109507531-109507531 on Build GRCh38
rs749593046 Chr.1: 109507563-109507563 on Build GRCh38
rs113099691 Chr.1: 109507568-109507568 on Build GRCh38
rs752863643 Chr.1: 109507734-109507734 on Build GRCh38
rs148318675 Chr.1: 109507907-109507907 on Build GRCh38

View Full Product Details