Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 111792096-111792593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC1
Entrez Gene ID
64682
Gene Name
anaphase promoting complex subunit 1
Gene Aliases
APC1, MCPR, TSG24
UniGene
Hs.436527
Gene Chromosome Location
Chr.2: 111766147-111884196 on Build GRCh38
Transcripts
NM_022662.3 XM_011511633.2 XM_011511634.2 XM_017004710.1 XM_017004711.1 XM_017004712.1 XM_017004713.1 XM_017004714.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4927593 c.4618C>T p.R1540C Chr.2: 111792456-111792456 on Build GRCh38
1005411 c.4676A>G p.H1559R Chr.2: 111792398-111792398 on Build GRCh38
4582570 c.4551C>T p.S1517S Chr.2: 111792523-111792523 on Build GRCh38
1305547 c.4589C>T p.S1530L Chr.2: 111792485-111792485 on Build GRCh38
714759 c.4537G>T p.E1513* Chr.2: 111792537-111792537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148071783 Chr.2: 111792504-111792504 on Build GRCh38
rs899855963 Chr.2: 111792210-111792211 on Build GRCh38
rs971060655 Chr.2: 111792480-111792480 on Build GRCh38
rs776564319 Chr.2: 111792487-111792487 on Build GRCh38
rs773227100 Chr.2: 111792509-111792509 on Build GRCh38
rs767790819 Chr.2: 111792523-111792523 on Build GRCh38
rs774494210 Chr.2: 111792520-111792520 on Build GRCh38
rs754399096 Chr.2: 111792535-111792535 on Build GRCh38
rs576922362 Chr.2: 111792564-111792564 on Build GRCh38
rs375968322 Chr.2: 111792212-111792212 on Build GRCh38
rs762905195 Chr.2: 111792510-111792510 on Build GRCh38
rs967883864 Chr.2: 111792338-111792338 on Build GRCh38
rs755266662 Chr.2: 111792379-111792379 on Build GRCh38
rs201139301 Chr.2: 111792175-111792175 on Build GRCh38
rs566398029 Chr.2: 111792350-111792350 on Build GRCh38
rs1039676370 Chr.2: 111792195-111792195 on Build GRCh38
rs556988866 Chr.2: 111792529-111792529 on Build GRCh38
rs80125903 Chr.2: 111792116-111792116 on Build GRCh38
rs537157408 Chr.2: 111792501-111792501 on Build GRCh38
rs568329960 Chr.2: 111792478-111792478 on Build GRCh38
rs754649421 Chr.2: 111792553-111792553 on Build GRCh38
rs1047223673 Chr.2: 111792154-111792154 on Build GRCh38
rs778382016 Chr.2: 111792437-111792437 on Build GRCh38
rs751807425 Chr.2: 111792375-111792375 on Build GRCh38
rs878980413 Chr.2: 111792117-111792117 on Build GRCh38
rs752971771 Chr.2: 111792524-111792524 on Build GRCh38
rs748018684 Chr.2: 111792488-111792488 on Build GRCh38
rs757947467 Chr.2: 111792470-111792470 on Build GRCh38
rs745347333 Chr.2: 111792444-111792444 on Build GRCh38
rs570270118 Chr.2: 111792279-111792279 on Build GRCh38
rs763145132 Chr.2: 111792347-111792347 on Build GRCh38
rs888666555 Chr.2: 111792160-111792160 on Build GRCh38
rs767877268 Chr.2: 111792402-111792402 on Build GRCh38
rs773138508 Chr.2: 111792341-111792341 on Build GRCh38
rs371253262 Chr.2: 111792486-111792486 on Build GRCh38
rs368922137 Chr.2: 111792513-111792513 on Build GRCh38
rs534987123 Chr.2: 111792436-111792436 on Build GRCh38
rs1029717337 Chr.2: 111792334-111792334 on Build GRCh38
rs779700098 Chr.2: 111792538-111792538 on Build GRCh38
rs930138589 Chr.2: 111792152-111792152 on Build GRCh38
rs144263623 Chr.2: 111792505-111792505 on Build GRCh38
rs552797128 Chr.2: 111792298-111792298 on Build GRCh38
rs761679359 Chr.2: 111792351-111792351 on Build GRCh38
rs761643946 Chr.2: 111792318-111792318 on Build GRCh38
rs919016956 Chr.2: 111792120-111792120 on Build GRCh38
rs370295733 Chr.2: 111792522-111792522 on Build GRCh38
rs149586183 Chr.2: 111792477-111792477 on Build GRCh38
rs1005793619 Chr.2: 111792190-111792190 on Build GRCh38
rs532802832 Chr.2: 111792289-111792289 on Build GRCh38
rs765289790 Chr.2: 111792319-111792319 on Build GRCh38
rs761148732 Chr.2: 111792525-111792525 on Build GRCh38
rs757789549 Chr.2: 111792537-111792537 on Build GRCh38
rs779786301 Chr.2: 111792471-111792471 on Build GRCh38
rs766542051 Chr.2: 111792353-111792353 on Build GRCh38
rs764999241 Chr.2: 111792552-111792555 on Build GRCh38

View Full Product Details