Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132729658-132730162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKLE2
Entrez Gene ID
23141
Gene Name
ankyrin repeat and LEM domain containing 2
Gene Aliases
KIAA0692, LEMD7, Lem4, MCPH16
UniGene
Hs.654628
Gene Chromosome Location
Chr.12: 132725667-132761888 on Build GRCh38
Transcripts
NM_015114.2 XM_005266159.3 XM_005266160.1 XM_006719735.1 XM_017019076.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6988464 c.2203G>A p.E735K Chr.12: 132729959-132729959 on Build GRCh38
5983227 c.2189C>G p.S730W Chr.12: 132729973-132729973 on Build GRCh38
4979701 c.2472C>G p.L824L Chr.12: 132729690-132729690 on Build GRCh38
5980441 c.2078G>A p.G693D Chr.12: 132730084-132730084 on Build GRCh38
3398550 c.2044G>C p.E682Q Chr.12: 132730118-132730118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763836655 Chr.12: 132730085-132730085 on Build GRCh38
rs755447074 Chr.12: 132730074-132730074 on Build GRCh38
rs756910711 Chr.12: 132730036-132730036 on Build GRCh38
rs915949460 Chr.12: 132730105-132730105 on Build GRCh38
rs755196191 Chr.12: 132729699-132729699 on Build GRCh38
rs746368812 Chr.12: 132729920-132729920 on Build GRCh38
rs749797672 Chr.12: 132730063-132730063 on Build GRCh38
rs375055766 Chr.12: 132730009-132730009 on Build GRCh38
rs951686329 Chr.12: 132730016-132730016 on Build GRCh38
rs368204052 Chr.12: 132730004-132730004 on Build GRCh38
rs754976405 Chr.12: 132729860-132729860 on Build GRCh38
rs761024797 Chr.12: 132729740-132729740 on Build GRCh38
rs761438395 Chr.12: 132730107-132730107 on Build GRCh38
rs763948286 Chr.12: 132730097-132730097 on Build GRCh38
rs757033242 Chr.12: 132729988-132729988 on Build GRCh38
rs202225435 Chr.12: 132729972-132729972 on Build GRCh38
rs78905764 Chr.12: 132730124-132730124 on Build GRCh38
rs530881578 Chr.12: 132729768-132729768 on Build GRCh38
rs200721415 Chr.12: 132729813-132729813 on Build GRCh38
rs762852125 Chr.12: 132730026-132730026 on Build GRCh38
rs753763983 Chr.12: 132729859-132729859 on Build GRCh38
rs754464109 Chr.12: 132729786-132729786 on Build GRCh38
rs868061452 Chr.12: 132730041-132730041 on Build GRCh38
rs776806854 Chr.12: 132729749-132729749 on Build GRCh38
rs749265166 Chr.12: 132729782-132729782 on Build GRCh38
rs201843669 Chr.12: 132729913-132729913 on Build GRCh38
rs10781634 Chr.12: 132730003-132730003 on Build GRCh38
rs375841027 Chr.12: 132729702-132729702 on Build GRCh38
rs779987897 Chr.12: 132729772-132729772 on Build GRCh38
rs767298754 Chr.12: 132730047-132730047 on Build GRCh38
rs376360110 Chr.12: 132729767-132729767 on Build GRCh38
rs781726198 Chr.12: 132729910-132729910 on Build GRCh38
rs768247207 Chr.12: 132729728-132729728 on Build GRCh38
rs752269208 Chr.12: 132729876-132729876 on Build GRCh38
rs12809719 Chr.12: 132729722-132729722 on Build GRCh38
rs200992817 Chr.12: 132729825-132729825 on Build GRCh38
rs1010551641 Chr.12: 132730000-132730000 on Build GRCh38
rs367741360 Chr.12: 132730027-132730027 on Build GRCh38
rs773326383 Chr.12: 132730024-132730024 on Build GRCh38
rs369709746 Chr.12: 132730095-132730095 on Build GRCh38
rs376479500 Chr.12: 132730136-132730136 on Build GRCh38
rs568989665 Chr.12: 132729956-132729956 on Build GRCh38
rs755790390 Chr.12: 132730131-132730131 on Build GRCh38
rs200179300 Chr.12: 132729815-132729815 on Build GRCh38
rs905709173 Chr.12: 132730057-132730057 on Build GRCh38
rs200253935 Chr.12: 132729902-132729902 on Build GRCh38
rs376936921 Chr.12: 132729905-132729905 on Build GRCh38
rs779472244 Chr.12: 132729720-132729720 on Build GRCh38
rs767008737 Chr.12: 132729741-132729741 on Build GRCh38
rs371526496 Chr.12: 132729681-132729681 on Build GRCh38
rs545538645 Chr.12: 132729708-132729708 on Build GRCh38
rs773963548 Chr.12: 132729731-132729731 on Build GRCh38
rs372987812 Chr.12: 132729805-132729805 on Build GRCh38
rs750777480 Chr.12: 132730054-132730054 on Build GRCh38
rs775472298 Chr.12: 132730025-132730025 on Build GRCh38
rs199617196 Chr.12: 132730090-132730090 on Build GRCh38
rs768769425 Chr.12: 132730006-132730006 on Build GRCh38
rs758700836 Chr.12: 132729755-132729755 on Build GRCh38
rs764984991 Chr.12: 132730018-132730018 on Build GRCh38
rs199793994 Chr.12: 132730086-132730086 on Build GRCh38
rs1030597464 Chr.12: 132729982-132729982 on Build GRCh38
rs756469595 Chr.12: 132730056-132730056 on Build GRCh38
rs545290587 Chr.12: 132730098-132730098 on Build GRCh38
rs369320391 Chr.12: 132729679-132729679 on Build GRCh38
rs759717835 Chr.12: 132729750-132729750 on Build GRCh38
rs552320030 Chr.12: 132729921-132729921 on Build GRCh38
rs1037509093 Chr.12: 132729693-132729693 on Build GRCh38
rs186469877 Chr.12: 132729824-132729824 on Build GRCh38
rs753636070 Chr.12: 132729942-132729942 on Build GRCh38
rs1046239513 Chr.12: 132729962-132729962 on Build GRCh38
rs769880047 Chr.12: 132730003-132730004 on Build GRCh38
rs773697339 Chr.12: 132729733-132729733 on Build GRCh38
rs761791514 Chr.12: 132730003-132730003 on Build GRCh38
rs370985412 Chr.12: 132729977-132729977 on Build GRCh38
rs750134734 Chr.12: 132729845-132729845 on Build GRCh38
rs376530487 Chr.12: 132729812-132729812 on Build GRCh38
rs772299133 Chr.12: 132730008-132730008 on Build GRCh38
rs572111545 Chr.12: 132729753-132729753 on Build GRCh38
rs762082409 Chr.12: 132729931-132729931 on Build GRCh38
rs753976276 Chr.12: 132729695-132729695 on Build GRCh38
rs369591537 Chr.12: 132729796-132729796 on Build GRCh38
rs929596288 Chr.12: 132729923-132729923 on Build GRCh38
rs201785518 Chr.12: 132729818-132729818 on Build GRCh38
rs947870303 Chr.12: 132729937-132729937 on Build GRCh38
rs773946036 Chr.12: 132730089-132730089 on Build GRCh38
rs559941180 Chr.12: 132729678-132729678 on Build GRCh38
rs201453405 Chr.12: 132730007-132730007 on Build GRCh38
rs775374422 Chr.12: 132729925-132729925 on Build GRCh38
rs745775918 Chr.12: 132729709-132729709 on Build GRCh38
rs941368788 Chr.12: 132729919-132729919 on Build GRCh38
rs748808201 Chr.12: 132729721-132729721 on Build GRCh38
rs565063145 Chr.12: 132729754-132729754 on Build GRCh38
rs774611351 Chr.12: 132729927-132729927 on Build GRCh38
rs200454083 Chr.12: 132729704-132729704 on Build GRCh38
rs767176067 Chr.12: 132730111-132730111 on Build GRCh38
rs766368315 Chr.12: 132729694-132729694 on Build GRCh38
rs754168716 Chr.12: 132730137-132730137 on Build GRCh38
rs542928581 Chr.12: 132729677-132729677 on Build GRCh38
rs759361950 Chr.12: 132730010-132730010 on Build GRCh38
rs777516084 Chr.12: 132729893-132729893 on Build GRCh38
rs1000297366 Chr.12: 132729737-132729737 on Build GRCh38
rs528690169 Chr.12: 132729703-132729703 on Build GRCh38
rs373826499 Chr.12: 132729848-132729848 on Build GRCh38
rs771764666 Chr.12: 132729800-132729800 on Build GRCh38
rs766212388 Chr.12: 132729853-132729853 on Build GRCh38
rs775703008 Chr.12: 132729823-132729823 on Build GRCh38
rs746248008 Chr.12: 132729993-132729993 on Build GRCh38
rs373384820 Chr.12: 132729719-132729719 on Build GRCh38
rs542277356 Chr.12: 132729843-132729843 on Build GRCh38
rs769893376 Chr.12: 132730094-132730094 on Build GRCh38
rs772312045 Chr.12: 132729732-132729732 on Build GRCh38
rs759414994 Chr.12: 132729943-132729943 on Build GRCh38
rs963399121 Chr.12: 132729847-132729847 on Build GRCh38
rs762533467 Chr.12: 132730076-132730079 on Build GRCh38
rs781633125 Chr.12: 132729991-132729991 on Build GRCh38
rs201957832 Chr.12: 132729973-132729973 on Build GRCh38
rs776299184 Chr.12: 132729828-132729830 on Build GRCh38
rs757940974 Chr.12: 132729880-132729880 on Build GRCh38
rs766158600 Chr.12: 132729939-132729939 on Build GRCh38
rs867149912 Chr.12: 132730024-132730024 on Build GRCh38
rs747751622 Chr.12: 132729798-132729798 on Build GRCh38
rs780341126 Chr.12: 132730062-132730062 on Build GRCh38
rs777195065 Chr.12: 132729976-132729976 on Build GRCh38
rs778331687 Chr.12: 132729870-132729870 on Build GRCh38
rs767211770 Chr.12: 132729759-132729759 on Build GRCh38
rs371204974 Chr.12: 132729926-132729926 on Build GRCh38
rs983016578 Chr.12: 132730066-132730066 on Build GRCh38
rs371226098 Chr.12: 132730077-132730077 on Build GRCh38

View Full Product Details