Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10581521-10581927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1M2
Entrez Gene ID
10053
Gene Name
adaptor related protein complex 1 mu 2 subunit
Gene Aliases
AP1-mu2, HSMU1B, MU-1B, MU1B, mu2
UniGene
Hs.18894
Gene Chromosome Location
Chr.19: 10572671-10587315 on Build GRCh38
Transcripts
NM_001300887.1 NM_005498.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6601578 c.368C>T p.P123L Chr.19: 10581778-10581778 on Build GRCh38
990643 c.484G>A p.E162K Chr.19: 10581549-10581549 on Build GRCh38
6863402 c.458T>C p.V153A Chr.19: 10581575-10581575 on Build GRCh38
4421408 c.483C>G p.S161S Chr.19: 10581550-10581550 on Build GRCh38
990644 c.465C>T p.N155N Chr.19: 10581568-10581568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751855453 Chr.19: 10581891-10581892 on Build GRCh38
rs372188688 Chr.19: 10581555-10581555 on Build GRCh38
rs200149852 Chr.19: 10581822-10581822 on Build GRCh38
rs375907948 Chr.19: 10581582-10581582 on Build GRCh38
rs1055926839 Chr.19: 10581789-10581789 on Build GRCh38
rs551854027 Chr.19: 10581813-10581813 on Build GRCh38
rs760710394 Chr.19: 10581654-10581654 on Build GRCh38
rs373357882 Chr.19: 10581588-10581588 on Build GRCh38
rs751408678 Chr.19: 10581554-10581554 on Build GRCh38
rs992829194 Chr.19: 10581574-10581574 on Build GRCh38
rs755619204 Chr.19: 10581568-10581568 on Build GRCh38
rs541778207 Chr.19: 10581727-10581727 on Build GRCh38
rs529831141 Chr.19: 10581712-10581712 on Build GRCh38
rs764470341 Chr.19: 10581879-10581879 on Build GRCh38
rs752585662 Chr.19: 10581567-10581567 on Build GRCh38
rs910378573 Chr.19: 10581898-10581898 on Build GRCh38
rs771207751 Chr.19: 10581644-10581644 on Build GRCh38
rs371453571 Chr.19: 10581683-10581683 on Build GRCh38
rs777823743 Chr.19: 10581711-10581711 on Build GRCh38
rs559559157 Chr.19: 10581699-10581699 on Build GRCh38
rs528647825 Chr.19: 10581857-10581857 on Build GRCh38
rs772222226 Chr.19: 10581824-10581824 on Build GRCh38
rs542063701 Chr.19: 10581591-10581591 on Build GRCh38
rs773360506 Chr.19: 10581549-10581549 on Build GRCh38
rs370831236 Chr.19: 10581663-10581663 on Build GRCh38
rs574645437 Chr.19: 10581726-10581726 on Build GRCh38
rs377099427 Chr.19: 10581653-10581653 on Build GRCh38
rs369982695 Chr.19: 10581903-10581903 on Build GRCh38
rs761877447 Chr.19: 10581840-10581840 on Build GRCh38
rs775196307 Chr.19: 10581730-10581730 on Build GRCh38
rs756967644 Chr.19: 10581678-10581678 on Build GRCh38
rs770337159 Chr.19: 10581707-10581707 on Build GRCh38
rs1013037656 Chr.19: 10581805-10581805 on Build GRCh38
rs748612320 Chr.19: 10581600-10581600 on Build GRCh38
rs753436956 Chr.19: 10581674-10581674 on Build GRCh38
rs777455515 Chr.19: 10581569-10581569 on Build GRCh38
rs370107597 Chr.19: 10581602-10581602 on Build GRCh38
rs776644129 Chr.19: 10581836-10581836 on Build GRCh38
rs935846516 Chr.19: 10581630-10581630 on Build GRCh38
rs369613061 Chr.19: 10581558-10581558 on Build GRCh38
rs757498415 Chr.19: 10581892-10581895 on Build GRCh38
rs763673386 Chr.19: 10581665-10581665 on Build GRCh38
rs751224180 Chr.19: 10581871-10581871 on Build GRCh38
rs756399358 Chr.19: 10581872-10581872 on Build GRCh38
rs201611005 Chr.19: 10581870-10581870 on Build GRCh38
rs774562057 Chr.19: 10581723-10581723 on Build GRCh38
rs375300415 Chr.19: 10581828-10581828 on Build GRCh38
rs766115907 Chr.19: 10581552-10581552 on Build GRCh38
rs369732438 Chr.19: 10581801-10581801 on Build GRCh38
rs562958085 Chr.19: 10581735-10581735 on Build GRCh38
rs765319398 Chr.19: 10581547-10581547 on Build GRCh38
rs986409760 Chr.19: 10581894-10581894 on Build GRCh38
rs761350214 Chr.19: 10581764-10581764 on Build GRCh38
rs749424190 Chr.19: 10581721-10581721 on Build GRCh38
rs774001333 Chr.19: 10581647-10581647 on Build GRCh38
rs746080407 Chr.19: 10581818-10581818 on Build GRCh38
rs769031495 Chr.19: 10581590-10581590 on Build GRCh38
rs753387578 Chr.19: 10581754-10581754 on Build GRCh38
rs200971288 Chr.19: 10581682-10581682 on Build GRCh38
rs201917850 Chr.19: 10581648-10581648 on Build GRCh38
rs770998547 Chr.19: 10581812-10581812 on Build GRCh38
rs371468321 Chr.19: 10581768-10581768 on Build GRCh38
rs772650211 Chr.19: 10581869-10581869 on Build GRCh38
rs1031594050 Chr.19: 10581619-10581619 on Build GRCh38
rs373522742 Chr.19: 10581572-10581572 on Build GRCh38
rs373528455 Chr.19: 10581550-10581550 on Build GRCh38
rs1005693095 Chr.19: 10581560-10581560 on Build GRCh38
rs781602013 Chr.19: 10581771-10581771 on Build GRCh38
rs778192600 Chr.19: 10581791-10581791 on Build GRCh38
rs371169664 Chr.19: 10581649-10581649 on Build GRCh38
rs367841075 Chr.19: 10581750-10581750 on Build GRCh38
rs370546263 Chr.19: 10581589-10581589 on Build GRCh38
rs756597491 Chr.19: 10581778-10581778 on Build GRCh38
rs1013005526 Chr.19: 10581559-10581559 on Build GRCh38
rs746859368 Chr.19: 10581833-10581833 on Build GRCh38
rs547770953 Chr.19: 10581767-10581767 on Build GRCh38
rs185906682 Chr.19: 10581835-10581835 on Build GRCh38
rs762039768 Chr.19: 10581777-10581777 on Build GRCh38
rs371384839 Chr.19: 10581722-10581722 on Build GRCh38
rs533071729 Chr.19: 10581672-10581672 on Build GRCh38
rs375046722 Chr.19: 10581718-10581718 on Build GRCh38
rs748608035 Chr.19: 10581703-10581703 on Build GRCh38
rs375773149 Chr.19: 10581601-10581601 on Build GRCh38
rs764760419 Chr.19: 10581854-10581854 on Build GRCh38

View Full Product Details