Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 223765220-223765724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1S3
Entrez Gene ID
130340
Gene Name
adaptor related protein complex 1 sigma 3 subunit
Gene Aliases
PSORS15
UniGene
Hs.632555
Gene Chromosome Location
Chr.2: 223755330-223837602 on Build GRCh38
Transcripts
NM_001039569.1 NR_110905.1 NR_110906.1 XM_011510600.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6242046 c.401C>A p.A134D Chr.2: 223765241-223765241 on Build GRCh38
4784530 c.364G>A p.E122K Chr.2: 223765278-223765278 on Build GRCh38
4832346 c.406G>A p.E136K Chr.2: 223765236-223765236 on Build GRCh38
4662233 c.380C>T p.S127F Chr.2: 223765262-223765262 on Build GRCh38
4764656 c.346G>A p.E116K Chr.2: 223765296-223765296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61737679 Chr.2: 223765297-223765297 on Build GRCh38
rs371696885 Chr.2: 223765397-223765397 on Build GRCh38
rs560056066 Chr.2: 223765621-223765621 on Build GRCh38
rs564198697 Chr.2: 223765515-223765515 on Build GRCh38
rs193142483 Chr.2: 223765446-223765446 on Build GRCh38
rs780060463 Chr.2: 223765287-223765287 on Build GRCh38
rs753621986 Chr.2: 223765399-223765399 on Build GRCh38
rs1046639068 Chr.2: 223765533-223765533 on Build GRCh38
rs1018439652 Chr.2: 223765412-223765412 on Build GRCh38
rs267599222 Chr.2: 223765303-223765303 on Build GRCh38
rs776079279 Chr.2: 223765331-223765331 on Build GRCh38
rs376103346 Chr.2: 223765350-223765350 on Build GRCh38
rs745715122 Chr.2: 223765270-223765270 on Build GRCh38
rs893444184 Chr.2: 223765370-223765370 on Build GRCh38
rs769638454 Chr.2: 223765259-223765267 on Build GRCh38
rs763487787 Chr.2: 223765337-223765337 on Build GRCh38
rs575872347 Chr.2: 223765504-223765504 on Build GRCh38
rs774285632 Chr.2: 223765502-223765502 on Build GRCh38
rs533043587 Chr.2: 223765525-223765525 on Build GRCh38
rs759979517 Chr.2: 223765326-223765326 on Build GRCh38
rs779644579 Chr.2: 223765398-223765398 on Build GRCh38
rs79499688 Chr.2: 223765288-223765288 on Build GRCh38
rs74693185 Chr.2: 223765419-223765419 on Build GRCh38
rs773200040 Chr.2: 223765361-223765362 on Build GRCh38
rs749992364 Chr.2: 223765382-223765382 on Build GRCh38
rs975539082 Chr.2: 223765458-223765458 on Build GRCh38
rs947846884 Chr.2: 223765305-223765305 on Build GRCh38
rs964333580 Chr.2: 223765413-223765413 on Build GRCh38
rs755823263 Chr.2: 223765283-223765283 on Build GRCh38
rs991497821 Chr.2: 223765510-223765510 on Build GRCh38
rs949330986 Chr.2: 223765545-223765545 on Build GRCh38
rs369564336 Chr.2: 223765296-223765296 on Build GRCh38
rs540722054 Chr.2: 223765511-223765511 on Build GRCh38
rs909986292 Chr.2: 223765466-223765466 on Build GRCh38
rs183553035 Chr.2: 223765680-223765680 on Build GRCh38
rs968053768 Chr.2: 223765404-223765405 on Build GRCh38
rs775829407 Chr.2: 223765600-223765600 on Build GRCh38
rs16865177 Chr.2: 223765505-223765505 on Build GRCh38
rs767229570 Chr.2: 223765359-223765359 on Build GRCh38
rs919677183 Chr.2: 223765473-223765473 on Build GRCh38
rs779455942 Chr.2: 223765536-223765536 on Build GRCh38
rs755948015 Chr.2: 223765383-223765383 on Build GRCh38
rs112069986 Chr.2: 223765608-223765608 on Build GRCh38
rs573465549 Chr.2: 223765410-223765410 on Build GRCh38
rs541109964 Chr.2: 223765327-223765327 on Build GRCh38
rs951534816 Chr.2: 223765437-223765437 on Build GRCh38
rs931047566 Chr.2: 223765498-223765498 on Build GRCh38
rs776689811 Chr.2: 223765310-223765310 on Build GRCh38
rs1029856254 Chr.2: 223765423-223765423 on Build GRCh38
rs1024786155 Chr.2: 223765278-223765278 on Build GRCh38
rs771195596 Chr.2: 223765304-223765304 on Build GRCh38
rs935177119 Chr.2: 223765609-223765609 on Build GRCh38
rs765472298 Chr.2: 223765330-223765330 on Build GRCh38
rs916478946 Chr.2: 223765512-223765512 on Build GRCh38
rs764693754 Chr.2: 223765345-223765345 on Build GRCh38
rs906274018 Chr.2: 223765332-223765332 on Build GRCh38
rs893560228 Chr.2: 223765652-223765652 on Build GRCh38

View Full Product Details