Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86153653-86154166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0D2
Entrez Gene ID
245972
Gene Name
ATPase H+ transporting V0 subunit d2
Gene Aliases
ATP6D2, VMA6
UniGene
Hs.436360
Gene Chromosome Location
Chr.8: 86098910-86154225 on Build GRCh38
Transcripts
NM_152565.1

SNP Detail

SNP ID Chromosome Location
rs137972049 Chr.8: 86153831-86153831 on Build GRCh38
rs930224615 Chr.8: 86153814-86153814 on Build GRCh38
rs770010328 Chr.8: 86153836-86153836 on Build GRCh38
rs373178076 Chr.8: 86153839-86153839 on Build GRCh38
rs891441759 Chr.8: 86153931-86153931 on Build GRCh38
rs553474808 Chr.8: 86153960-86153960 on Build GRCh38
rs980270589 Chr.8: 86153768-86153768 on Build GRCh38
rs537642378 Chr.8: 86153852-86153852 on Build GRCh38
rs1007090888 Chr.8: 86153853-86153853 on Build GRCh38
rs901303429 Chr.8: 86153862-86153862 on Build GRCh38
rs59120440 Chr.8: 86153776-86153777 on Build GRCh38
rs919273264 Chr.8: 86154013-86154013 on Build GRCh38
rs989152889 Chr.8: 86153893-86153893 on Build GRCh38
rs371737573 Chr.8: 86153757-86153758 on Build GRCh38
rs16890863 Chr.8: 86153737-86153737 on Build GRCh38
rs968943925 Chr.8: 86153934-86153934 on Build GRCh38
rs955988334 Chr.8: 86153726-86153726 on Build GRCh38
rs779148875 Chr.8: 86153996-86153996 on Build GRCh38
rs560483204 Chr.8: 86153716-86153716 on Build GRCh38
rs765039048 Chr.8: 86153754-86153758 on Build GRCh38
rs570093580 Chr.8: 86153972-86153972 on Build GRCh38
rs191791814 Chr.8: 86154130-86154130 on Build GRCh38
rs973436408 Chr.8: 86154004-86154004 on Build GRCh38
rs918275789 Chr.8: 86154005-86154005 on Build GRCh38
rs1021428230 Chr.8: 86153920-86153920 on Build GRCh38
rs141838117 Chr.8: 86153753-86153754 on Build GRCh38
rs552484068 Chr.8: 86153809-86153809 on Build GRCh38
rs745895699 Chr.8: 86154000-86154000 on Build GRCh38
rs774925612 Chr.8: 86153856-86153856 on Build GRCh38
rs914390793 Chr.8: 86153738-86153738 on Build GRCh38
rs910162445 Chr.8: 86154053-86154053 on Build GRCh38
rs762375894 Chr.8: 86154023-86154023 on Build GRCh38
rs986310754 Chr.8: 86153977-86153977 on Build GRCh38
rs868453820 Chr.8: 86154033-86154033 on Build GRCh38
rs1027116968 Chr.8: 86153932-86153932 on Build GRCh38
rs772455185 Chr.8: 86154064-86154064 on Build GRCh38
rs955916691 Chr.8: 86153885-86153885 on Build GRCh38
rs1044577294 Chr.8: 86153846-86153846 on Build GRCh38
rs547785700 Chr.8: 86153758-86153759 on Build GRCh38
rs948593628 Chr.8: 86154103-86154103 on Build GRCh38
rs1031322388 Chr.8: 86153898-86153898 on Build GRCh38
rs867643454 Chr.8: 86153777-86153777 on Build GRCh38
rs952913286 Chr.8: 86154011-86154011 on Build GRCh38
rs187086145 Chr.8: 86154120-86154120 on Build GRCh38
rs959785339 Chr.8: 86153959-86153959 on Build GRCh38
rs539084308 Chr.8: 86154047-86154047 on Build GRCh38
rs979982852 Chr.8: 86153951-86153951 on Build GRCh38
rs548318843 Chr.8: 86153866-86153866 on Build GRCh38
rs80355395 Chr.8: 86154072-86154072 on Build GRCh38
rs927058579 Chr.8: 86153772-86153772 on Build GRCh38
rs1056852819 Chr.8: 86153778-86153778 on Build GRCh38
rs182761063 Chr.8: 86154079-86154079 on Build GRCh38
rs947133435 Chr.8: 86153767-86153767 on Build GRCh38
rs112122662 Chr.8: 86153958-86153958 on Build GRCh38
rs948541743 Chr.8: 86153793-86153794 on Build GRCh38
rs566985912 Chr.8: 86153869-86153869 on Build GRCh38
rs1057019600 Chr.8: 86153773-86153773 on Build GRCh38
rs568172692 Chr.8: 86153894-86153894 on Build GRCh38
rs1030967488 Chr.8: 86153871-86153871 on Build GRCh38
rs930216950 Chr.8: 86154014-86154014 on Build GRCh38

View Full Product Details