Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44777916-44778323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLC
Entrez Gene ID
23624
Gene Name
Cbl proto-oncogene C
Gene Aliases
CBL-3, CBL-SL, RNF57
UniGene
Hs.466907
Gene Chromosome Location
Chr.19: 44777855-44800646 on Build GRCh38
Transcripts
NM_001130852.1 NM_012116.3 XM_005258696.3 XM_011526688.2 XM_011526689.2 XM_011526690.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3535746 c.166C>T p.R56* Chr.19: 44778097-44778097 on Build GRCh38
6416321 c.325G>T p.E109* Chr.19: 44778256-44778256 on Build GRCh38
3990103 c.333C>G p.F111L Chr.19: 44778264-44778264 on Build GRCh38
5828885 c.187C>T p.R63W Chr.19: 44778118-44778118 on Build GRCh38
3422920 c.109C>T p.L37L Chr.19: 44778040-44778040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558512943 Chr.19: 44778045-44778045 on Build GRCh38
rs1039416820 Chr.19: 44778147-44778147 on Build GRCh38
rs866606097 Chr.19: 44778032-44778032 on Build GRCh38
rs1035887672 Chr.19: 44778242-44778242 on Build GRCh38
rs778906584 Chr.19: 44778022-44778022 on Build GRCh38
rs937514313 Chr.19: 44778123-44778123 on Build GRCh38
rs759152258 Chr.19: 44777981-44777981 on Build GRCh38
rs770372319 Chr.19: 44777971-44777971 on Build GRCh38
rs1030666838 Chr.19: 44778244-44778244 on Build GRCh38
rs761041888 Chr.19: 44778011-44778011 on Build GRCh38
rs772312798 Chr.19: 44777942-44777942 on Build GRCh38
rs768263588 Chr.19: 44778200-44778200 on Build GRCh38
rs996021174 Chr.19: 44778231-44778231 on Build GRCh38
rs572115097 Chr.19: 44778104-44778104 on Build GRCh38
rs115775900 Chr.19: 44778118-44778118 on Build GRCh38
rs1040883653 Chr.19: 44778116-44778116 on Build GRCh38
rs983411997 Chr.19: 44778056-44778056 on Build GRCh38
rs931299718 Chr.19: 44778064-44778064 on Build GRCh38
rs544784225 Chr.19: 44778036-44778036 on Build GRCh38
rs756006315 Chr.19: 44778042-44778042 on Build GRCh38
rs778923243 Chr.19: 44778143-44778143 on Build GRCh38
rs952747717 Chr.19: 44778275-44778275 on Build GRCh38
rs764374254 Chr.19: 44778013-44778013 on Build GRCh38
rs563733027 Chr.19: 44778255-44778255 on Build GRCh38
rs776955522 Chr.19: 44778080-44778080 on Build GRCh38
rs1014912119 Chr.19: 44778274-44778274 on Build GRCh38
rs949078334 Chr.19: 44778146-44778146 on Build GRCh38
rs773804002 Chr.19: 44778258-44778258 on Build GRCh38
rs770544741 Chr.19: 44778138-44778139 on Build GRCh38
rs568288126 Chr.19: 44778279-44778279 on Build GRCh38
rs750480822 Chr.19: 44778023-44778023 on Build GRCh38
rs1020514306 Chr.19: 44777997-44777997 on Build GRCh38
rs200348346 Chr.19: 44778008-44778008 on Build GRCh38
rs758611319 Chr.19: 44778025-44778025 on Build GRCh38
rs900367969 Chr.19: 44778195-44778195 on Build GRCh38
rs762946724 Chr.19: 44777968-44777968 on Build GRCh38
rs746650758 Chr.19: 44778115-44778116 on Build GRCh38
rs754372194 Chr.19: 44778018-44778018 on Build GRCh38
rs749090451 Chr.19: 44778131-44778131 on Build GRCh38
rs774931320 Chr.19: 44778171-44778171 on Build GRCh38
rs1000666675 Chr.19: 44777999-44777999 on Build GRCh38
rs543965825 Chr.19: 44778207-44778207 on Build GRCh38
rs766618955 Chr.19: 44778294-44778294 on Build GRCh38
rs773799426 Chr.19: 44778054-44778054 on Build GRCh38
rs772943938 Chr.19: 44778100-44778101 on Build GRCh38
rs1032038079 Chr.19: 44778179-44778179 on Build GRCh38
rs749288188 Chr.19: 44778046-44778046 on Build GRCh38
rs530433110 Chr.19: 44778164-44778164 on Build GRCh38
rs1053937701 Chr.19: 44778150-44778150 on Build GRCh38
rs757201148 Chr.19: 44778142-44778142 on Build GRCh38
rs943884863 Chr.19: 44778100-44778100 on Build GRCh38
rs747946824 Chr.19: 44777952-44777952 on Build GRCh38
rs975094314 Chr.19: 44778030-44778030 on Build GRCh38
rs35701940 Chr.19: 44778291-44778292 on Build GRCh38
rs757709044 Chr.19: 44778019-44778019 on Build GRCh38
rs983465843 Chr.19: 44778263-44778263 on Build GRCh38
rs1032029318 Chr.19: 44777947-44777947 on Build GRCh38
rs775969330 Chr.19: 44778142-44778142 on Build GRCh38
rs777753616 Chr.19: 44778119-44778119 on Build GRCh38
rs773891788 Chr.19: 44777980-44777980 on Build GRCh38
rs548486447 Chr.19: 44778199-44778199 on Build GRCh38
rs541721336 Chr.19: 44778107-44778107 on Build GRCh38
rs902236013 Chr.19: 44777973-44777973 on Build GRCh38
rs775127808 Chr.19: 44778076-44778076 on Build GRCh38
rs967594513 Chr.19: 44778232-44778232 on Build GRCh38
rs767195128 Chr.19: 44778005-44778005 on Build GRCh38
rs140237650 Chr.19: 44777942-44777943 on Build GRCh38
rs771859187 Chr.19: 44778013-44778014 on Build GRCh38
rs367849742 Chr.19: 44778246-44778246 on Build GRCh38
rs780884827 Chr.19: 44777944-44777944 on Build GRCh38
rs762221327 Chr.19: 44778213-44778213 on Build GRCh38
rs762265193 Chr.19: 44778083-44778083 on Build GRCh38
rs919974224 Chr.19: 44778260-44778260 on Build GRCh38
rs147885389 Chr.19: 44778055-44778055 on Build GRCh38
rs34932144 Chr.19: 44777954-44777954 on Build GRCh38
rs148948759 Chr.19: 44778077-44778077 on Build GRCh38
rs1000593338 Chr.19: 44778172-44778172 on Build GRCh38
rs777043372 Chr.19: 44778205-44778205 on Build GRCh38
rs765766542 Chr.19: 44778098-44778098 on Build GRCh38
rs765456312 Chr.19: 44778235-44778235 on Build GRCh38
rs746098134 Chr.19: 44777941-44777941 on Build GRCh38
rs746709240 Chr.19: 44778190-44778190 on Build GRCh38
rs773289730 Chr.19: 44777965-44777965 on Build GRCh38
rs780339606 Chr.19: 44778031-44778031 on Build GRCh38

View Full Product Details