Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65155030-65155568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLPX
Entrez Gene ID
10845
Gene Name
caseinolytic mitochondrial matrix peptidase chaperone subunit
Gene Aliases
-
UniGene
Hs.113823
Gene Chromosome Location
Chr.15: 65148219-65185420 on Build GRCh38
Transcripts
NM_006660.4 NR_133680.1 XM_011521164.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6329327 c.1342G>A p.G448R Chr.15: 65155051-65155051 on Build GRCh38
1749269 c.1355G>T p.R452M Chr.15: 65155038-65155038 on Build GRCh38
6330746 c.1324G>A p.G442R Chr.15: 65155069-65155069 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs551962246 Chr.15: 65155050-65155050 on Build GRCh38
rs957014804 Chr.15: 65155438-65155438 on Build GRCh38
rs186647987 Chr.15: 65155522-65155522 on Build GRCh38
rs918142842 Chr.15: 65155218-65155218 on Build GRCh38
rs1000049853 Chr.15: 65155423-65155423 on Build GRCh38
rs758135098 Chr.15: 65155106-65155106 on Build GRCh38
rs35262213 Chr.15: 65155105-65155106 on Build GRCh38
rs745747684 Chr.15: 65155105-65155105 on Build GRCh38
rs377233548 Chr.15: 65155217-65155218 on Build GRCh38
rs779692253 Chr.15: 65155537-65155537 on Build GRCh38
rs535103569 Chr.15: 65155297-65155297 on Build GRCh38
rs112513983 Chr.15: 65155066-65155066 on Build GRCh38
rs988561293 Chr.15: 65155182-65155184 on Build GRCh38
rs549822242 Chr.15: 65155079-65155079 on Build GRCh38
rs904337682 Chr.15: 65155420-65155420 on Build GRCh38
rs911394920 Chr.15: 65155178-65155178 on Build GRCh38
rs1008308225 Chr.15: 65155465-65155465 on Build GRCh38
rs1020292470 Chr.15: 65155441-65155441 on Build GRCh38
rs981087805 Chr.15: 65155243-65155243 on Build GRCh38
rs1047408061 Chr.15: 65155271-65155271 on Build GRCh38
rs1032452472 Chr.15: 65155125-65155125 on Build GRCh38
rs372984639 Chr.15: 65155169-65155169 on Build GRCh38
rs571769966 Chr.15: 65155317-65155317 on Build GRCh38
rs543636678 Chr.15: 65155505-65155505 on Build GRCh38
rs529233183 Chr.15: 65155529-65155529 on Build GRCh38
rs758614832 Chr.15: 65155528-65155528 on Build GRCh38
rs974332378 Chr.15: 65155186-65155186 on Build GRCh38
rs549387944 Chr.15: 65155539-65155539 on Build GRCh38
rs574306694 Chr.15: 65155212-65155212 on Build GRCh38
rs557382932 Chr.15: 65155200-65155200 on Build GRCh38
rs1010540653 Chr.15: 65155421-65155421 on Build GRCh38
rs986815969 Chr.15: 65155151-65155151 on Build GRCh38
rs757598939 Chr.15: 65155097-65155097 on Build GRCh38
rs1048106497 Chr.15: 65155295-65155295 on Build GRCh38
rs2414876 Chr.15: 65155495-65155495 on Build GRCh38
rs776160262 Chr.15: 65155130-65155130 on Build GRCh38
rs930327608 Chr.15: 65155242-65155242 on Build GRCh38
rs376399690 Chr.15: 65155181-65155181 on Build GRCh38
rs1045468665 Chr.15: 65155326-65155326 on Build GRCh38
rs879324440 Chr.15: 65155503-65155503 on Build GRCh38
rs1020681618 Chr.15: 65155141-65155141 on Build GRCh38
rs779841803 Chr.15: 65155113-65155113 on Build GRCh38
rs891084910 Chr.15: 65155459-65155459 on Build GRCh38
rs1018308259 Chr.15: 65155487-65155487 on Build GRCh38
rs778028615 Chr.15: 65155069-65155069 on Build GRCh38
rs756636936 Chr.15: 65155061-65155061 on Build GRCh38
rs955497525 Chr.15: 65155148-65155148 on Build GRCh38
rs201869759 Chr.15: 65155373-65155373 on Build GRCh38
rs554986520 Chr.15: 65155306-65155306 on Build GRCh38
rs376949994 Chr.15: 65155406-65155406 on Build GRCh38
rs767853852 Chr.15: 65155058-65155058 on Build GRCh38
rs1032398853 Chr.15: 65155425-65155425 on Build GRCh38
rs926787913 Chr.15: 65155272-65155272 on Build GRCh38
rs746560956 Chr.15: 65155538-65155538 on Build GRCh38
rs997157772 Chr.15: 65155383-65155383 on Build GRCh38
rs753157584 Chr.15: 65155060-65155060 on Build GRCh38
rs746944472 Chr.15: 65155114-65155114 on Build GRCh38
rs781588969 Chr.15: 65155279-65155279 on Build GRCh38
rs931512583 Chr.15: 65155274-65155274 on Build GRCh38
rs184100973 Chr.15: 65155351-65155351 on Build GRCh38
rs569637818 Chr.15: 65155355-65155355 on Build GRCh38
rs907501492 Chr.15: 65155287-65155287 on Build GRCh38
rs756449518 Chr.15: 65155347-65155347 on Build GRCh38
rs920175280 Chr.15: 65155198-65155198 on Build GRCh38
rs11635595 Chr.15: 65155381-65155381 on Build GRCh38
rs768541749 Chr.15: 65155127-65155127 on Build GRCh38
rs1027541357 Chr.15: 65155523-65155523 on Build GRCh38
rs747643005 Chr.15: 65155131-65155131 on Build GRCh38

View Full Product Details