Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45846708-45847248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRY2
Entrez Gene ID
1408
Gene Name
cryptochrome circadian clock 2
Gene Aliases
HCRY2, PHLL2
UniGene
Hs.532491
Gene Chromosome Location
Chr.11: 45847118-45883248 on Build GRCh38
Transcripts
NM_001127457.2

SNP Detail

SNP ID Chromosome Location
rs920934379 Chr.11: 45846980-45846980 on Build GRCh38
rs549459670 Chr.11: 45847158-45847158 on Build GRCh38
rs752192327 Chr.11: 45847200-45847201 on Build GRCh38
rs912103564 Chr.11: 45846902-45846902 on Build GRCh38
rs1002113064 Chr.11: 45846993-45846993 on Build GRCh38
rs919580181 Chr.11: 45847083-45847083 on Build GRCh38
rs1038412095 Chr.11: 45846748-45846753 on Build GRCh38
rs762570390 Chr.11: 45847201-45847217 on Build GRCh38
rs939800438 Chr.11: 45846743-45846743 on Build GRCh38
rs529410313 Chr.11: 45847071-45847071 on Build GRCh38
rs968079116 Chr.11: 45846988-45846988 on Build GRCh38
rs1050264260 Chr.11: 45847104-45847119 on Build GRCh38
rs1033088233 Chr.11: 45846799-45846799 on Build GRCh38
rs905860167 Chr.11: 45847123-45847123 on Build GRCh38
rs745816902 Chr.11: 45846898-45846898 on Build GRCh38
rs1050669643 Chr.11: 45846809-45846809 on Build GRCh38
rs1044740995 Chr.11: 45847122-45847122 on Build GRCh38
rs1040563716 Chr.11: 45846939-45846939 on Build GRCh38
rs140500010 Chr.11: 45846749-45846749 on Build GRCh38
rs939811176 Chr.11: 45847059-45847059 on Build GRCh38
rs1033624420 Chr.11: 45847016-45847016 on Build GRCh38
rs932337710 Chr.11: 45846981-45846981 on Build GRCh38
rs944971586 Chr.11: 45847117-45847117 on Build GRCh38
rs1010061164 Chr.11: 45847129-45847129 on Build GRCh38
rs949996514 Chr.11: 45847106-45847106 on Build GRCh38
rs796514430 Chr.11: 45846892-45846892 on Build GRCh38
rs769389529 Chr.11: 45846956-45846956 on Build GRCh38
rs892515197 Chr.11: 45846950-45846950 on Build GRCh38
rs976784513 Chr.11: 45846975-45846975 on Build GRCh38
rs988959066 Chr.11: 45846888-45846888 on Build GRCh38
rs781128443 Chr.11: 45846785-45846785 on Build GRCh38
rs780961899 Chr.11: 45847230-45847230 on Build GRCh38
rs1000229609 Chr.11: 45846783-45846783 on Build GRCh38
rs754735174 Chr.11: 45847225-45847225 on Build GRCh38
rs908380679 Chr.11: 45847039-45847039 on Build GRCh38
rs1048158771 Chr.11: 45847019-45847019 on Build GRCh38
rs909656153 Chr.11: 45847032-45847032 on Build GRCh38
rs571040377 Chr.11: 45846974-45846974 on Build GRCh38
rs1033035937 Chr.11: 45847201-45847201 on Build GRCh38
rs966325563 Chr.11: 45846949-45846949 on Build GRCh38
rs1022675904 Chr.11: 45846957-45846957 on Build GRCh38
rs533207223 Chr.11: 45846847-45846847 on Build GRCh38
rs970071350 Chr.11: 45846751-45846751 on Build GRCh38
rs111440493 Chr.11: 45846880-45846880 on Build GRCh38
rs1000178855 Chr.11: 45847150-45847150 on Build GRCh38
rs895685024 Chr.11: 45847089-45847089 on Build GRCh38
rs563679950 Chr.11: 45846976-45846976 on Build GRCh38
rs961379380 Chr.11: 45847067-45847067 on Build GRCh38
rs1010788956 Chr.11: 45847214-45847214 on Build GRCh38
rs891896386 Chr.11: 45847203-45847203 on Build GRCh38

View Full Product Details