Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 149546585-149547086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf40A
Entrez Gene ID
91966
Gene Name
chromosome X open reading frame 40A
Gene Aliases
CXorf40, EOLA1, FLJ16423
UniGene
Hs.458216
Gene Chromosome Location
Chr.X: 149540630-149555338 on Build GRCh38
Transcripts
NM_001171907.2 NM_001171908.2 NM_001171909.3 NM_001324274.1 NM_001324275.1 NM_001324276.1 NM_001324277.1 NM_001324278.1 NM_001324279.1 NM_001324280.1 NM_178124.5 XM_005262349.4

SNP Detail

SNP ID Chromosome Location
rs141148310 Chr.X: 149546789-149546789 on Build GRCh38
rs781914229 Chr.X: 149546779-149546779 on Build GRCh38
rs781881848 Chr.X: 149546848-149546848 on Build GRCh38
rs781937700 Chr.X: 149546987-149546987 on Build GRCh38
rs781852659 Chr.X: 149546995-149546995 on Build GRCh38
rs782263934 Chr.X: 149546888-149546888 on Build GRCh38
rs146921818 Chr.X: 149546943-149546943 on Build GRCh38
rs782512155 Chr.X: 149547005-149547005 on Build GRCh38
rs781882682 Chr.X: 149546694-149546694 on Build GRCh38
rs782154220 Chr.X: 149546991-149546991 on Build GRCh38
rs782562069 Chr.X: 149546881-149546881 on Build GRCh38
rs147733282 Chr.X: 149546774-149546774 on Build GRCh38
rs782610552 Chr.X: 149546727-149546727 on Build GRCh38
rs782325989 Chr.X: 149546731-149546731 on Build GRCh38
rs782731115 Chr.X: 149546807-149546807 on Build GRCh38
rs781990675 Chr.X: 149546791-149546791 on Build GRCh38
rs782178952 Chr.X: 149546935-149546935 on Build GRCh38
rs150927010 Chr.X: 149546949-149546949 on Build GRCh38
rs782671234 Chr.X: 149546736-149546736 on Build GRCh38
rs782624190 Chr.X: 149546884-149546884 on Build GRCh38
rs782732288 Chr.X: 149546845-149546845 on Build GRCh38
rs782295406 Chr.X: 149546744-149546744 on Build GRCh38
rs781920871 Chr.X: 149546667-149546667 on Build GRCh38
rs509250 Chr.X: 149547062-149547062 on Build GRCh38
rs782466220 Chr.X: 149546897-149546897 on Build GRCh38
rs782102377 Chr.X: 149546977-149546977 on Build GRCh38
rs782141941 Chr.X: 149546804-149546804 on Build GRCh38
rs782610819 Chr.X: 149546921-149546921 on Build GRCh38
rs374116725 Chr.X: 149546714-149546714 on Build GRCh38
rs35210802 Chr.X: 149547062-149547063 on Build GRCh38
rs7378 Chr.X: 149547013-149547013 on Build GRCh38
rs782401602 Chr.X: 149546966-149546966 on Build GRCh38
rs781957366 Chr.X: 149546971-149546971 on Build GRCh38
rs782440502 Chr.X: 149546830-149546830 on Build GRCh38
rs374770539 Chr.X: 149546767-149546767 on Build GRCh38
rs11987 Chr.X: 149546820-149546820 on Build GRCh38
rs782702459 Chr.X: 149546978-149546978 on Build GRCh38
rs782799138 Chr.X: 149546994-149546994 on Build GRCh38
rs782752836 Chr.X: 149546788-149546788 on Build GRCh38
rs12116111 Chr.X: 149546944-149546944 on Build GRCh38
rs782003395 Chr.X: 149546769-149546769 on Build GRCh38
rs7379 Chr.X: 149547032-149547032 on Build GRCh38
rs782469876 Chr.X: 149546721-149546721 on Build GRCh38
rs7380 Chr.X: 149547050-149547050 on Build GRCh38
rs145671628 Chr.X: 149546773-149546773 on Build GRCh38
rs138997261 Chr.X: 149547036-149547036 on Build GRCh38

View Full Product Details