Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5724203-5724615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM208B
Entrez Gene ID
54906
Gene Name
family with sequence similarity 208 member B
Gene Aliases
C10orf18, bA318E3.2
UniGene
Hs.610717
Gene Chromosome Location
Chr.10: 5684759-5763740 on Build GRCh38
Transcripts
NM_001321783.1 NM_001321784.1 NM_001321785.1 NM_017782.4 XM_005252474.3 XM_005252475.3 XM_005252476.4 XM_005252480.4 XM_005252481.4 XM_005252482.2 XM_011519515.2 XM_011519516.2 XM_011519517.2 XM_011519518.2 XM_011519519.2 XM_011519520.2 XM_011519521.2 XM_011519522.2 XM_011519525.2 XM_017016361.1 XM_017016362.1 XM_017016363.1 XM_017016364.1 XM_017016365.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5749463 c.335T>C p.I112T Chr.10: 5724517-5724517 on Build GRCh38
6567724 c.248-6C>T p.? Chr.10: 5724424-5724424 on Build GRCh38
6338384 c.276G>T p.L92F Chr.10: 5724458-5724458 on Build GRCh38
5359956 c.303A>G p.Q101Q Chr.10: 5724485-5724485 on Build GRCh38
1289390 c.348A>G p.Q116Q Chr.10: 5724530-5724530 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754111881 Chr.10: 5724312-5724312 on Build GRCh38
rs763657169 Chr.10: 5724444-5724444 on Build GRCh38
rs772335984 Chr.10: 5724390-5724390 on Build GRCh38
rs749759371 Chr.10: 5724458-5724458 on Build GRCh38
rs982009942 Chr.10: 5724500-5724500 on Build GRCh38
rs750741806 Chr.10: 5724446-5724446 on Build GRCh38
rs777459288 Chr.10: 5724522-5724524 on Build GRCh38
rs373569620 Chr.10: 5724477-5724477 on Build GRCh38
rs540201661 Chr.10: 5724454-5724454 on Build GRCh38
rs770821955 Chr.10: 5724527-5724528 on Build GRCh38
rs916224024 Chr.10: 5724240-5724240 on Build GRCh38
rs759077360 Chr.10: 5724429-5724429 on Build GRCh38
rs1019635125 Chr.10: 5724408-5724408 on Build GRCh38
rs572525463 Chr.10: 5724432-5724432 on Build GRCh38
rs778184237 Chr.10: 5724259-5724259 on Build GRCh38
rs10737018 Chr.10: 5724374-5724374 on Build GRCh38
rs759314567 Chr.10: 5724559-5724561 on Build GRCh38
rs1008193959 Chr.10: 5724394-5724394 on Build GRCh38
rs961727287 Chr.10: 5724555-5724560 on Build GRCh38
rs776598455 Chr.10: 5724557-5724560 on Build GRCh38
rs949145103 Chr.10: 5724592-5724592 on Build GRCh38
rs775072233 Chr.10: 5724559-5724559 on Build GRCh38
rs72774076 Chr.10: 5724389-5724389 on Build GRCh38
rs972693853 Chr.10: 5724448-5724448 on Build GRCh38
rs377622420 Chr.10: 5724482-5724482 on Build GRCh38
rs773059022 Chr.10: 5724393-5724393 on Build GRCh38
rs371027335 Chr.10: 5724496-5724496 on Build GRCh38
rs774449878 Chr.10: 5724382-5724382 on Build GRCh38
rs1009074915 Chr.10: 5724254-5724254 on Build GRCh38
rs778947171 Chr.10: 5724468-5724468 on Build GRCh38
rs1046047012 Chr.10: 5724301-5724301 on Build GRCh38
rs758111912 Chr.10: 5724438-5724438 on Build GRCh38
rs375095868 Chr.10: 5724509-5724509 on Build GRCh38
rs770664519 Chr.10: 5724492-5724492 on Build GRCh38
rs766285568 Chr.10: 5724414-5724414 on Build GRCh38
rs772273627 Chr.10: 5724474-5724474 on Build GRCh38
rs780566495 Chr.10: 5724455-5724455 on Build GRCh38
rs143897907 Chr.10: 5724381-5724381 on Build GRCh38
rs746470796 Chr.10: 5724526-5724527 on Build GRCh38
rs949093321 Chr.10: 5724242-5724242 on Build GRCh38
rs769617674 Chr.10: 5724575-5724578 on Build GRCh38
rs764809933 Chr.10: 5724431-5724431 on Build GRCh38
rs561665364 Chr.10: 5724589-5724589 on Build GRCh38
rs771783933 Chr.10: 5724395-5724397 on Build GRCh38
rs377480980 Chr.10: 5724437-5724437 on Build GRCh38
rs907820712 Chr.10: 5724519-5724519 on Build GRCh38
rs1053671785 Chr.10: 5724353-5724353 on Build GRCh38
rs193221853 Chr.10: 5724350-5724350 on Build GRCh38
rs763753727 Chr.10: 5724585-5724585 on Build GRCh38
rs369992459 Chr.10: 5724471-5724471 on Build GRCh38
rs907428880 Chr.10: 5724333-5724333 on Build GRCh38
rs762564239 Chr.10: 5724566-5724566 on Build GRCh38
rs760393686 Chr.10: 5724401-5724401 on Build GRCh38
rs184442211 Chr.10: 5724426-5724426 on Build GRCh38
rs765325194 Chr.10: 5724530-5724530 on Build GRCh38

View Full Product Details