Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154987936-154988444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLAD1
Entrez Gene ID
80308
Gene Name
flavin adenine dinucleotide synthetase 1
Gene Aliases
FAD1, FADS, LSMFLAD, PP591
UniGene
Hs.118666
Gene Chromosome Location
Chr.1: 154983294-154993111 on Build GRCh38
Transcripts
NM_001184891.1 NM_001184892.1 NM_025207.4 NM_201398.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4622135 c.121C>T p.R41C Chr.1: 154988150-154988150 on Build GRCh38
269179 c.618A>G p.E206E Chr.1: 154988350-154988350 on Build GRCh38
4398173 c.541C>T p.P181S Chr.1: 154988273-154988273 on Build GRCh38
7195729 c.344G>A p.G115E Chr.1: 154988373-154988373 on Build GRCh38
6664362 c.475A>G p.I159V Chr.1: 154988207-154988207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750853103 Chr.1: 154988098-154988099 on Build GRCh38
rs370120170 Chr.1: 154988099-154988099 on Build GRCh38
rs767091112 Chr.1: 154988269-154988269 on Build GRCh38
rs769578156 Chr.1: 154988106-154988106 on Build GRCh38
rs368257835 Chr.1: 154988213-154988213 on Build GRCh38
rs867813327 Chr.1: 154988119-154988119 on Build GRCh38
rs1009078855 Chr.1: 154988389-154988389 on Build GRCh38
rs377032800 Chr.1: 154988173-154988173 on Build GRCh38
rs753948492 Chr.1: 154988079-154988079 on Build GRCh38
rs140409443 Chr.1: 154988175-154988175 on Build GRCh38
rs558598160 Chr.1: 154987983-154987983 on Build GRCh38
rs375023842 Chr.1: 154988102-154988102 on Build GRCh38
rs1026708401 Chr.1: 154988054-154988054 on Build GRCh38
rs764231231 Chr.1: 154988069-154988069 on Build GRCh38
rs776874328 Chr.1: 154988341-154988341 on Build GRCh38
rs369915979 Chr.1: 154988293-154988293 on Build GRCh38
rs138843652 Chr.1: 154988377-154988377 on Build GRCh38
rs1037960547 Chr.1: 154988237-154988237 on Build GRCh38
rs767252057 Chr.1: 154988128-154988130 on Build GRCh38
rs771503176 Chr.1: 154988042-154988042 on Build GRCh38
rs920886455 Chr.1: 154988264-154988264 on Build GRCh38
rs376637647 Chr.1: 154988270-154988270 on Build GRCh38
rs780578888 Chr.1: 154988284-154988284 on Build GRCh38
rs762410845 Chr.1: 154988069-154988070 on Build GRCh38
rs780088175 Chr.1: 154988091-154988091 on Build GRCh38
rs765635608 Chr.1: 154988094-154988095 on Build GRCh38
rs145054820 Chr.1: 154988372-154988372 on Build GRCh38
rs763964020 Chr.1: 154988262-154988262 on Build GRCh38
rs876661315 Chr.1: 154988230-154988230 on Build GRCh38
rs777621144 Chr.1: 154988183-154988183 on Build GRCh38
rs745306436 Chr.1: 154988221-154988221 on Build GRCh38
rs764022484 Chr.1: 154988408-154988408 on Build GRCh38
rs748865800 Chr.1: 154988198-154988198 on Build GRCh38
rs747895040 Chr.1: 154988105-154988105 on Build GRCh38
rs192454315 Chr.1: 154987973-154987973 on Build GRCh38
rs769489679 Chr.1: 154987994-154987994 on Build GRCh38
rs778295669 Chr.1: 154988210-154988210 on Build GRCh38
rs768977246 Chr.1: 154988238-154988238 on Build GRCh38
rs774985299 Chr.1: 154988128-154988128 on Build GRCh38
rs751572707 Chr.1: 154988166-154988166 on Build GRCh38
rs374803472 Chr.1: 154988414-154988414 on Build GRCh38
rs765228219 Chr.1: 154988162-154988162 on Build GRCh38
rs372170239 Chr.1: 154988151-154988151 on Build GRCh38
rs761928040 Chr.1: 154988240-154988240 on Build GRCh38
rs879171776 Chr.1: 154988338-154988338 on Build GRCh38
rs765442744 Chr.1: 154988245-154988245 on Build GRCh38
rs776464003 Chr.1: 154987990-154987990 on Build GRCh38
rs755926549 Chr.1: 154988177-154988177 on Build GRCh38
rs750044923 Chr.1: 154988421-154988421 on Build GRCh38
rs757880504 Chr.1: 154988280-154988280 on Build GRCh38
rs775833539 Chr.1: 154988148-154988148 on Build GRCh38
rs150372864 Chr.1: 154988235-154988235 on Build GRCh38
rs148134209 Chr.1: 154988379-154988379 on Build GRCh38
rs879119618 Chr.1: 154988417-154988417 on Build GRCh38
rs556272462 Chr.1: 154988152-154988152 on Build GRCh38
rs144986636 Chr.1: 154988415-154988415 on Build GRCh38
rs145148009 Chr.1: 154988115-154988115 on Build GRCh38
rs895264052 Chr.1: 154987984-154987984 on Build GRCh38
rs876661312 Chr.1: 154988258-154988269 on Build GRCh38
rs876661310 Chr.1: 154988301-154988302 on Build GRCh38
rs776977748 Chr.1: 154988239-154988239 on Build GRCh38
rs757233939 Chr.1: 154988087-154988087 on Build GRCh38
rs1041500319 Chr.1: 154988392-154988392 on Build GRCh38
rs772596666 Chr.1: 154988222-154988222 on Build GRCh38
rs372048892 Chr.1: 154988254-154988254 on Build GRCh38
rs773171190 Chr.1: 154988247-154988247 on Build GRCh38
rs754886240 Chr.1: 154988168-154988168 on Build GRCh38
rs769942679 Chr.1: 154988369-154988369 on Build GRCh38
rs770329270 Chr.1: 154988125-154988125 on Build GRCh38
rs921109401 Chr.1: 154988155-154988155 on Build GRCh38
rs572645053 Chr.1: 154988336-154988336 on Build GRCh38
rs752137994 Chr.1: 154988402-154988402 on Build GRCh38
rs754559231 Chr.1: 154988110-154988113 on Build GRCh38
rs764845268 Chr.1: 154988416-154988416 on Build GRCh38
rs112420699 Chr.1: 154987979-154987979 on Build GRCh38
rs772863652 Chr.1: 154988023-154988023 on Build GRCh38
rs747798837 Chr.1: 154987991-154987991 on Build GRCh38
rs756018287 Chr.1: 154988256-154988256 on Build GRCh38
rs763089278 Chr.1: 154988250-154988250 on Build GRCh38
rs876661313 Chr.1: 154988133-154988136 on Build GRCh38
rs150683998 Chr.1: 154988402-154988402 on Build GRCh38
rs1057518160 Chr.1: 154988140-154988140 on Build GRCh38
rs766539386 Chr.1: 154988378-154988378 on Build GRCh38
rs748480567 Chr.1: 154988359-154988359 on Build GRCh38
rs370022149 Chr.1: 154988056-154988056 on Build GRCh38
rs752662832 Chr.1: 154988255-154988255 on Build GRCh38
rs760898739 Chr.1: 154988068-154988068 on Build GRCh38
rs367959633 Chr.1: 154988141-154988141 on Build GRCh38
rs1004152015 Chr.1: 154987975-154987975 on Build GRCh38
rs61741145 Chr.1: 154988142-154988142 on Build GRCh38
rs770528228 Chr.1: 154988208-154988208 on Build GRCh38
rs566506048 Chr.1: 154987972-154987972 on Build GRCh38
rs544231293 Chr.1: 154988150-154988150 on Build GRCh38
rs560748872 Chr.1: 154988277-154988277 on Build GRCh38
rs527449937 Chr.1: 154988048-154988048 on Build GRCh38
rs775640575 Chr.1: 154988380-154988380 on Build GRCh38
rs576901302 Chr.1: 154988030-154988030 on Build GRCh38
rs752970809 Chr.1: 154988061-154988061 on Build GRCh38
rs765661425 Chr.1: 154988058-154988058 on Build GRCh38
rs753608799 Chr.1: 154988267-154988267 on Build GRCh38
rs751421255 Chr.1: 154988097-154988097 on Build GRCh38

View Full Product Details