Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RUNX1T1
- Entrez Gene ID
-
862
- Gene Name
- RUNX1 translocation partner 1
- Gene Aliases
-
AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2
- Chromosome Location
-
Chr.8: 91954967 - 92103365 on Build GRCh38
View Full Product Details