Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 144007058-144007598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYMAI
Entrez Gene ID
57061
Gene Name
hydatidiform mole associated and imprinted (non-protein coding)
Gene Aliases
NCRNA00020
UniGene
Hs.657760
Gene Chromosome Location
Chr.6: 144004916-144008730 on Build GRCh38
Transcripts
NR_002768.2 NM_001080951.2 NM_001080952.2 NM_001080953.2 NM_001080954.2 NM_001080955.2 NM_001080956.2 NM_001289037.1 NM_001289038.1 NM_001289039.1 NM_001289040.1 NM_001289041.1 NM_001289042.1 NM_001289043.1 NM_001289044.1 NM_001289045.1 NM_001289046.1 NM_001289047.1 NM_001289048.1 NM_001289049.1 NM_001317159.1 NM_001317160.1 NM_001317161.1 NM_001317162.1 NM_006718.4

Gene Symbol
PLAGL1
Entrez Gene ID
5325
Gene Name
PLAG1 like zinc finger 1
Gene Aliases
LOT1, ZAC, ZAC1
UniGene
Hs.743225
Gene Chromosome Location
Chr.6: 143940300-144064599 on Build GRCh38
Transcripts
NR_002768.2 NM_001080951.2 NM_001080952.2 NM_001080953.2 NM_001080954.2 NM_001080955.2 NM_001080956.2 NM_001289037.1 NM_001289038.1 NM_001289039.1 NM_001289040.1 NM_001289041.1 NM_001289042.1 NM_001289043.1 NM_001289044.1 NM_001289045.1 NM_001289046.1 NM_001289047.1 NM_001289048.1 NM_001289049.1 NM_001317159.1 NM_001317160.1 NM_001317161.1 NM_001317162.1 NM_006718.4

SNP Detail

SNP ID Chromosome Location
rs968728092 Chr.6: 144007300-144007300 on Build GRCh38
rs551130635 Chr.6: 144007359-144007359 on Build GRCh38
rs941514502 Chr.6: 144007164-144007164 on Build GRCh38
rs905773288 Chr.6: 144007249-144007249 on Build GRCh38
rs776020539 Chr.6: 144007451-144007451 on Build GRCh38
rs73006277 Chr.6: 144007534-144007534 on Build GRCh38
rs929662794 Chr.6: 144007118-144007118 on Build GRCh38
rs887421348 Chr.6: 144007210-144007210 on Build GRCh38
rs768981654 Chr.6: 144007324-144007324 on Build GRCh38
rs998677973 Chr.6: 144007205-144007205 on Build GRCh38
rs1005868187 Chr.6: 144007224-144007224 on Build GRCh38
rs566556921 Chr.6: 144007398-144007398 on Build GRCh38
rs911457091 Chr.6: 144007079-144007079 on Build GRCh38
rs934208706 Chr.6: 144007373-144007373 on Build GRCh38
rs916917601 Chr.6: 144007090-144007090 on Build GRCh38
rs891892507 Chr.6: 144007286-144007286 on Build GRCh38
rs958605691 Chr.6: 144007287-144007287 on Build GRCh38
rs951123783 Chr.6: 144007535-144007535 on Build GRCh38
rs568685211 Chr.6: 144007434-144007434 on Build GRCh38
rs1034305112 Chr.6: 144007446-144007446 on Build GRCh38
rs983187937 Chr.6: 144007386-144007386 on Build GRCh38
rs561567714 Chr.6: 144007229-144007229 on Build GRCh38
rs529142858 Chr.6: 144007231-144007231 on Build GRCh38
rs1040287745 Chr.6: 144007171-144007171 on Build GRCh38
rs760312498 Chr.6: 144007340-144007340 on Build GRCh38
rs776174915 Chr.6: 144007133-144007133 on Build GRCh38
rs965620075 Chr.6: 144007322-144007322 on Build GRCh38
rs770204868 Chr.6: 144007294-144007294 on Build GRCh38
rs968622331 Chr.6: 144007284-144007284 on Build GRCh38
rs938577492 Chr.6: 144007485-144007485 on Build GRCh38
rs540019338 Chr.6: 144007228-144007228 on Build GRCh38
rs148450783 Chr.6: 144007568-144007568 on Build GRCh38
rs944231174 Chr.6: 144007082-144007082 on Build GRCh38
rs74968697 Chr.6: 144007189-144007189 on Build GRCh38
rs761256735 Chr.6: 144007366-144007366 on Build GRCh38
rs1041216072 Chr.6: 144007085-144007085 on Build GRCh38
rs759023237 Chr.6: 144007493-144007493 on Build GRCh38
rs951008184 Chr.6: 144007307-144007307 on Build GRCh38
rs1021651556 Chr.6: 144007298-144007298 on Build GRCh38
rs929078960 Chr.6: 144007365-144007365 on Build GRCh38
rs533903323 Chr.6: 144007399-144007399 on Build GRCh38
rs539805658 Chr.6: 144007512-144007512 on Build GRCh38
rs144569241 Chr.6: 144007433-144007433 on Build GRCh38
rs143105242 Chr.6: 144007130-144007130 on Build GRCh38
rs1039832560 Chr.6: 144007346-144007346 on Build GRCh38
rs904401155 Chr.6: 144007436-144007436 on Build GRCh38
rs772889042 Chr.6: 144007326-144007326 on Build GRCh38
rs562296792 Chr.6: 144007280-144007280 on Build GRCh38
rs1003158090 Chr.6: 144007266-144007266 on Build GRCh38
rs1000518458 Chr.6: 144007441-144007441 on Build GRCh38
rs535732467 Chr.6: 144007443-144007443 on Build GRCh38
rs918729604 Chr.6: 144007332-144007332 on Build GRCh38
rs557631847 Chr.6: 144007487-144007487 on Build GRCh38
rs575601817 Chr.6: 144007078-144007078 on Build GRCh38
rs1000000069 Chr.6: 144007285-144007285 on Build GRCh38
rs776024082 Chr.6: 144007304-144007304 on Build GRCh38
rs982409964 Chr.6: 144007559-144007559 on Build GRCh38
rs765742987 Chr.6: 144007135-144007135 on Build GRCh38
rs1020137549 Chr.6: 144007273-144007273 on Build GRCh38
rs532938058 Chr.6: 144007358-144007358 on Build GRCh38
rs749633762 Chr.6: 144007314-144007314 on Build GRCh38
rs564233848 Chr.6: 144007163-144007163 on Build GRCh38
rs550862530 Chr.6: 144007271-144007271 on Build GRCh38
rs929024896 Chr.6: 144007136-144007136 on Build GRCh38
rs569490803 Chr.6: 144007498-144007498 on Build GRCh38
rs1014111088 Chr.6: 144007524-144007524 on Build GRCh38
rs1018973830 Chr.6: 144007308-144007308 on Build GRCh38

View Full Product Details