Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49440171-49440702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUT
Entrez Gene ID
4594
Gene Name
methylmalonyl-CoA mutase
Gene Aliases
MCM
UniGene
Hs.485527
Gene Chromosome Location
Chr.6: 49430360-49463328 on Build GRCh38
Transcripts
NM_000255.3 XM_005249143.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
742692 c.1904C>A p.A635D Chr.6: 49440258-49440258 on Build GRCh38
1292337 c.1829G>A p.R610H Chr.6: 49440333-49440333 on Build GRCh38
3628247 c.1837C>T p.R613C Chr.6: 49440325-49440325 on Build GRCh38
4955328 c.1902T>A p.I634I Chr.6: 49440260-49440260 on Build GRCh38
223094 c.1834G>A p.G612S Chr.6: 49440328-49440328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371665665 Chr.6: 49440374-49440374 on Build GRCh38
rs893711847 Chr.6: 49440584-49440584 on Build GRCh38
rs746563084 Chr.6: 49440474-49440474 on Build GRCh38
rs962978567 Chr.6: 49440589-49440589 on Build GRCh38
rs756743196 Chr.6: 49440353-49440353 on Build GRCh38
rs760692554 Chr.6: 49440215-49440216 on Build GRCh38
rs1042380782 Chr.6: 49440488-49440488 on Build GRCh38
rs865815395 Chr.6: 49440244-49440244 on Build GRCh38
rs778617168 Chr.6: 49440262-49440262 on Build GRCh38
rs946689006 Chr.6: 49440486-49440486 on Build GRCh38
rs889618893 Chr.6: 49440532-49440532 on Build GRCh38
rs143023066 Chr.6: 49440273-49440273 on Build GRCh38
rs753123265 Chr.6: 49440352-49440352 on Build GRCh38
rs938214569 Chr.6: 49440447-49440447 on Build GRCh38
rs368139265 Chr.6: 49440376-49440376 on Build GRCh38
rs927077779 Chr.6: 49440666-49440666 on Build GRCh38
rs372486357 Chr.6: 49440282-49440282 on Build GRCh38
rs573727541 Chr.6: 49440325-49440325 on Build GRCh38
rs750619189 Chr.6: 49440204-49440204 on Build GRCh38
rs767966332 Chr.6: 49440349-49440349 on Build GRCh38
rs115741934 Chr.6: 49440638-49440638 on Build GRCh38
rs766529896 Chr.6: 49440333-49440333 on Build GRCh38
rs758674900 Chr.6: 49440218-49440218 on Build GRCh38
rs200055428 Chr.6: 49440264-49440264 on Build GRCh38
rs1023427200 Chr.6: 49440633-49440633 on Build GRCh38
rs368690932 Chr.6: 49440324-49440324 on Build GRCh38
rs770978452 Chr.6: 49440334-49440334 on Build GRCh38
rs938175848 Chr.6: 49440672-49440672 on Build GRCh38
rs981150971 Chr.6: 49440648-49440648 on Build GRCh38
rs201536536 Chr.6: 49440297-49440297 on Build GRCh38
rs982110849 Chr.6: 49440286-49440286 on Build GRCh38
rs781095765 Chr.6: 49440398-49440398 on Build GRCh38
rs200996892 Chr.6: 49440320-49440320 on Build GRCh38
rs894074192 Chr.6: 49440463-49440463 on Build GRCh38
rs1055381041 Chr.6: 49440459-49440459 on Build GRCh38
rs755256821 Chr.6: 49440348-49440348 on Build GRCh38
rs1007174916 Chr.6: 49440558-49440558 on Build GRCh38
rs943752009 Chr.6: 49440478-49440478 on Build GRCh38
rs906616202 Chr.6: 49440637-49440637 on Build GRCh38
rs569669261 Chr.6: 49440256-49440256 on Build GRCh38
rs370761357 Chr.6: 49440488-49440489 on Build GRCh38
rs1012507905 Chr.6: 49440599-49440599 on Build GRCh38
rs879253847 Chr.6: 49440288-49440288 on Build GRCh38
rs765284825 Chr.6: 49440316-49440316 on Build GRCh38
rs746708475 Chr.6: 49440388-49440388 on Build GRCh38
rs78100773 Chr.6: 49440529-49440529 on Build GRCh38
rs184057724 Chr.6: 49440649-49440649 on Build GRCh38
rs755382704 Chr.6: 49440224-49440224 on Build GRCh38
rs201489323 Chr.6: 49440408-49440408 on Build GRCh38
rs778562196 Chr.6: 49440364-49440364 on Build GRCh38
rs121918254 Chr.6: 49440295-49440295 on Build GRCh38
rs863224898 Chr.6: 49440271-49440271 on Build GRCh38
rs751828125 Chr.6: 49440345-49440345 on Build GRCh38
rs756798250 Chr.6: 49440261-49440261 on Build GRCh38
rs563690091 Chr.6: 49440573-49440573 on Build GRCh38
rs775291072 Chr.6: 49440276-49440276 on Build GRCh38
rs546095473 Chr.6: 49440531-49440531 on Build GRCh38
rs986286327 Chr.6: 49440401-49440401 on Build GRCh38
rs781501004 Chr.6: 49440253-49440253 on Build GRCh38
rs747897332 Chr.6: 49440238-49440238 on Build GRCh38
rs1025225144 Chr.6: 49440631-49440631 on Build GRCh38
rs768521956 Chr.6: 49440291-49440291 on Build GRCh38
rs766721811 Chr.6: 49440219-49440219 on Build GRCh38
rs981159200 Chr.6: 49440373-49440373 on Build GRCh38
rs902532966 Chr.6: 49440489-49440489 on Build GRCh38
rs751804284 Chr.6: 49440220-49440220 on Build GRCh38
rs777532992 Chr.6: 49440440-49440440 on Build GRCh38
rs554196010 Chr.6: 49440458-49440458 on Build GRCh38
rs879253846 Chr.6: 49440309-49440309 on Build GRCh38
rs893670326 Chr.6: 49440231-49440231 on Build GRCh38
rs745503019 Chr.6: 49440268-49440268 on Build GRCh38
rs374494276 Chr.6: 49440556-49440556 on Build GRCh38
rs398123277 Chr.6: 49440308-49440310 on Build GRCh38
rs770331518 Chr.6: 49440484-49440484 on Build GRCh38
rs757999518 Chr.6: 49440375-49440375 on Build GRCh38
rs973689274 Chr.6: 49440362-49440362 on Build GRCh38
rs972644072 Chr.6: 49440615-49440615 on Build GRCh38
rs532855260 Chr.6: 49440577-49440577 on Build GRCh38
rs796052004 Chr.6: 49440277-49440277 on Build GRCh38
rs199592843 Chr.6: 49440390-49440390 on Build GRCh38
rs1040695915 Chr.6: 49440519-49440519 on Build GRCh38
rs779526951 Chr.6: 49440382-49440382 on Build GRCh38

View Full Product Details