Product Details

Assay Reference Genome
Location

Chr.19:16226464 on build GRCh38
Cytoband
19p13.11
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
8907
Gene Symbol
AP1M1
Gene Name
adaptor related protein complex 1 mu 1 subunit
Gene Aliases
AP47, CLAPM2, CLTNM, MU-1A
Location
Chr.19:16197854-16235345 on build GRCh38
Assay Gene Location
Within Exon 8
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
AP1M1 NM_001130524.1 7 799 NP_001123996.1
NM_032493.3 6 763 NP_115882.1
XM_017027420.1 7 739 XP_016882909.1
XM_017027421.1 6 703 XP_016882910.1
AB209808.1 7 719 BAD93045.1
AF290613.1 6 666 AAK28024.1
AK027528.1 6 671
AK293188.1 5 547
AK297824.1 6 706
BC017469.2 6 708 AAH17469.1
DQ059565.1 7 626 AAY54246.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv833767 Chr19:16121391 - 16347760 on Build GRCh38 Loss CIB3 HSH2D FAM32A KLF2 AP1M1 RAB8A

View Full Product Details