Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107642266-107642759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPS1
Entrez Gene ID
5631
Gene Name
phosphoribosyl pyrophosphate synthetase 1
Gene Aliases
ARTS, CMTX5, DFN2, DFNX1, PPRibP, PRS-I, PRSI
UniGene
Hs.56
Gene Chromosome Location
Chr.X: 107628424-107651026 on Build GRCh38
Transcripts
NM_001204402.1 NM_002764.3

SNP Detail

SNP ID Chromosome Location
rs775344370 Chr.X: 107642514-107642514 on Build GRCh38
rs759133840 Chr.X: 107642459-107642459 on Build GRCh38
rs765582060 Chr.X: 107642497-107642497 on Build GRCh38
rs768454424 Chr.X: 107642393-107642393 on Build GRCh38
rs1015778376 Chr.X: 107642550-107642550 on Build GRCh38
rs201285459 Chr.X: 107642404-107642404 on Build GRCh38
rs764641895 Chr.X: 107642517-107642517 on Build GRCh38
rs890471190 Chr.X: 107642499-107642500 on Build GRCh38
rs374847355 Chr.X: 107642505-107642505 on Build GRCh38
rs61752962 Chr.X: 107642437-107642437 on Build GRCh38
rs61735617 Chr.X: 107642416-107642416 on Build GRCh38
rs776308065 Chr.X: 107642452-107642452 on Build GRCh38
rs961757497 Chr.X: 107642557-107642557 on Build GRCh38
rs398122855 Chr.X: 107642384-107642384 on Build GRCh38
rs757533724 Chr.X: 107642674-107642674 on Build GRCh38
rs377525200 Chr.X: 107642508-107642508 on Build GRCh38
rs760421564 Chr.X: 107642603-107642603 on Build GRCh38
rs754926002 Chr.X: 107642338-107642338 on Build GRCh38
rs888601981 Chr.X: 107642457-107642457 on Build GRCh38
rs80338676 Chr.X: 107642415-107642415 on Build GRCh38
rs747278443 Chr.X: 107642640-107642640 on Build GRCh38
rs751616994 Chr.X: 107642529-107642529 on Build GRCh38
rs757366976 Chr.X: 107642537-107642537 on Build GRCh38
rs1021888722 Chr.X: 107642714-107642714 on Build GRCh38
rs80338730 Chr.X: 107642407-107642407 on Build GRCh38
rs966273444 Chr.X: 107642597-107642597 on Build GRCh38
rs373971736 Chr.X: 107642380-107642380 on Build GRCh38
rs768359844 Chr.X: 107642673-107642673 on Build GRCh38
rs370001788 Chr.X: 107642329-107642329 on Build GRCh38
rs781524630 Chr.X: 107642346-107642346 on Build GRCh38
rs372046005 Chr.X: 107642320-107642320 on Build GRCh38
rs772838760 Chr.X: 107642414-107642414 on Build GRCh38
rs993249305 Chr.X: 107642605-107642605 on Build GRCh38

View Full Product Details