Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 166199380-166199879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929680
Entrez Gene ID
101929680
Gene Name
uncharacterized LOC101929680
Gene Aliases
-
UniGene
Hs.662210
Gene Chromosome Location
Chr.2: 166081531-166301784 on Build GRCh38
Transcripts
NR_110260.1 NM_002977.3 XM_005246757.2 XM_011511616.2 XM_011511617.2 XM_011511618.2 XM_011511619.2 XM_017004668.1 XM_017004669.1

Gene Symbol
SCN9A
Entrez Gene ID
6335
Gene Name
sodium voltage-gated channel alpha subunit 9
Gene Aliases
ETHA, FEB3B, GEFSP7, HSAN2D, NE-NA, NENA, Nav1.7, PN1, SFNP
UniGene
Hs.439145
Gene Chromosome Location
Chr.2: 166195185-166375987 on Build GRCh38
Transcripts
NR_110260.1 NM_002977.3 XM_005246757.2 XM_011511616.2 XM_011511617.2 XM_011511618.2 XM_011511619.2 XM_017004668.1 XM_017004669.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4571656 c.5121T>A p.P1707P Chr.2: 166199521-166199521 on Build GRCh38
1008765 c.5026A>G p.S1676G Chr.2: 166199580-166199580 on Build GRCh38
6041339 c.5243C>A p.S1748Y Chr.2: 166199399-166199399 on Build GRCh38
3569168 c.4817C>T p.S1606F Chr.2: 166199825-166199825 on Build GRCh38
4571657 c.5085T>A p.P1695P Chr.2: 166199521-166199521 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202208945 Chr.2: 166199452-166199452 on Build GRCh38
rs372887865 Chr.2: 166199478-166199478 on Build GRCh38
rs867544697 Chr.2: 166199580-166199580 on Build GRCh38
rs751948774 Chr.2: 166199631-166199631 on Build GRCh38
rs370267820 Chr.2: 166199530-166199530 on Build GRCh38
rs976620881 Chr.2: 166199689-166199689 on Build GRCh38
rs794727440 Chr.2: 166199404-166199404 on Build GRCh38
rs938958211 Chr.2: 166199498-166199498 on Build GRCh38
rs879254053 Chr.2: 166199735-166199735 on Build GRCh38
rs200266625 Chr.2: 166199468-166199468 on Build GRCh38
rs762076834 Chr.2: 166199823-166199823 on Build GRCh38
rs1026039008 Chr.2: 166199754-166199754 on Build GRCh38
rs267598971 Chr.2: 166199535-166199535 on Build GRCh38
rs201554947 Chr.2: 166199520-166199520 on Build GRCh38
rs775277644 Chr.2: 166199416-166199416 on Build GRCh38
rs767879707 Chr.2: 166199594-166199594 on Build GRCh38
rs763972874 Chr.2: 166199427-166199427 on Build GRCh38
rs200042812 Chr.2: 166199624-166199624 on Build GRCh38
rs375946140 Chr.2: 166199840-166199840 on Build GRCh38
rs202155356 Chr.2: 166199748-166199748 on Build GRCh38
rs201537698 Chr.2: 166199444-166199444 on Build GRCh38
rs1027061303 Chr.2: 166199482-166199482 on Build GRCh38
rs200364146 Chr.2: 166199762-166199762 on Build GRCh38
rs371341018 Chr.2: 166199837-166199837 on Build GRCh38
rs777688318 Chr.2: 166199803-166199803 on Build GRCh38
rs768953526 Chr.2: 166199815-166199815 on Build GRCh38
rs757858235 Chr.2: 166199665-166199665 on Build GRCh38
rs779325771 Chr.2: 166199661-166199661 on Build GRCh38
rs746833579 Chr.2: 166199512-166199512 on Build GRCh38
rs200637125 Chr.2: 166199546-166199546 on Build GRCh38
rs200338455 Chr.2: 166199836-166199836 on Build GRCh38
rs770424172 Chr.2: 166199401-166199401 on Build GRCh38
rs973063452 Chr.2: 166199743-166199743 on Build GRCh38
rs757798263 Chr.2: 166199652-166199652 on Build GRCh38
rs759618043 Chr.2: 166199590-166199590 on Build GRCh38
rs376892319 Chr.2: 166199523-166199523 on Build GRCh38
rs768193278 Chr.2: 166199680-166199680 on Build GRCh38
rs751081867 Chr.2: 166199852-166199852 on Build GRCh38
rs768548713 Chr.2: 166199517-166199517 on Build GRCh38
rs769660085 Chr.2: 166199797-166199797 on Build GRCh38
rs779521437 Chr.2: 166199671-166199671 on Build GRCh38
rs764559105 Chr.2: 166199629-166199629 on Build GRCh38
rs547418120 Chr.2: 166199794-166199794 on Build GRCh38
rs748002341 Chr.2: 166199793-166199793 on Build GRCh38
rs201079869 Chr.2: 166199747-166199747 on Build GRCh38
rs200470541 Chr.2: 166199819-166199819 on Build GRCh38
rs201371960 Chr.2: 166199664-166199664 on Build GRCh38
rs751682235 Chr.2: 166199442-166199442 on Build GRCh38
rs200993217 Chr.2: 166199773-166199773 on Build GRCh38
rs199806034 Chr.2: 166199487-166199487 on Build GRCh38
rs765068868 Chr.2: 166199572-166199572 on Build GRCh38
rs750980200 Chr.2: 166199770-166199770 on Build GRCh38
rs200817449 Chr.2: 166199419-166199419 on Build GRCh38
rs142201175 Chr.2: 166199783-166199783 on Build GRCh38
rs200553720 Chr.2: 166199659-166199659 on Build GRCh38
rs776586182 Chr.2: 166199432-166199432 on Build GRCh38
rs761637677 Chr.2: 166199437-166199437 on Build GRCh38
rs925260428 Chr.2: 166199727-166199727 on Build GRCh38
rs761022368 Chr.2: 166199744-166199744 on Build GRCh38
rs747598487 Chr.2: 166199707-166199707 on Build GRCh38
rs200166620 Chr.2: 166199674-166199674 on Build GRCh38
rs149207258 Chr.2: 166199827-166199827 on Build GRCh38
rs1055143422 Chr.2: 166199844-166199844 on Build GRCh38
rs765755704 Chr.2: 166199769-166199769 on Build GRCh38
rs199567943 Chr.2: 166199534-166199534 on Build GRCh38
rs201989809 Chr.2: 166199789-166199789 on Build GRCh38
rs200733722 Chr.2: 166199499-166199499 on Build GRCh38
rs775654130 Chr.2: 166199811-166199811 on Build GRCh38
rs778164639 Chr.2: 166199480-166199480 on Build GRCh38
rs199550149 Chr.2: 166199716-166199716 on Build GRCh38
rs766522690 Chr.2: 166199441-166199441 on Build GRCh38
rs368487219 Chr.2: 166199673-166199673 on Build GRCh38
rs903242507 Chr.2: 166199578-166199578 on Build GRCh38
rs61029356 Chr.2: 166199668-166199668 on Build GRCh38
rs199910483 Chr.2: 166199718-166199718 on Build GRCh38
rs866205197 Chr.2: 166199575-166199575 on Build GRCh38
rs754141856 Chr.2: 166199636-166199636 on Build GRCh38
rs752918053 Chr.2: 166199620-166199620 on Build GRCh38
rs879253994 Chr.2: 166199711-166199711 on Build GRCh38
rs747837371 Chr.2: 166199698-166199698 on Build GRCh38
rs774583593 Chr.2: 166199573-166199573 on Build GRCh38
rs894505761 Chr.2: 166199554-166199554 on Build GRCh38
rs760606370 Chr.2: 166199812-166199812 on Build GRCh38
rs770986636 Chr.2: 166199809-166199809 on Build GRCh38
rs779609081 Chr.2: 166199784-166199784 on Build GRCh38
rs572623538 Chr.2: 166199801-166199801 on Build GRCh38
rs567322169 Chr.2: 166199806-166199806 on Build GRCh38
rs201186665 Chr.2: 166199707-166199707 on Build GRCh38
rs754670164 Chr.2: 166199792-166199792 on Build GRCh38
rs771718677 Chr.2: 166199500-166199500 on Build GRCh38
rs773240225 Chr.2: 166199702-166199702 on Build GRCh38
rs771700555 Chr.2: 166199413-166199413 on Build GRCh38
rs201421963 Chr.2: 166199842-166199842 on Build GRCh38
rs769855186 Chr.2: 166199699-166199699 on Build GRCh38
rs866369977 Chr.2: 166199704-166199707 on Build GRCh38
rs749848631 Chr.2: 166199482-166199482 on Build GRCh38
rs199675949 Chr.2: 166199828-166199828 on Build GRCh38
rs201783312 Chr.2: 166199849-166199849 on Build GRCh38
rs201850617 Chr.2: 166199569-166199569 on Build GRCh38
rs776545037 Chr.2: 166199686-166199686 on Build GRCh38
rs879254140 Chr.2: 166199553-166199553 on Build GRCh38
rs201264153 Chr.2: 166199475-166199475 on Build GRCh38
rs759253467 Chr.2: 166199410-166199410 on Build GRCh38
rs761880427 Chr.2: 166199543-166199543 on Build GRCh38

View Full Product Details