Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68803760-68804256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANP32A
Entrez Gene ID
8125
Gene Name
acidic nuclear phosphoprotein 32 family member A
Gene Aliases
C15orf1, HPPCn, I1PP2A, LANP, MAPM, PHAP1, PHAPI, PP32
UniGene
Hs.458747
Gene Chromosome Location
Chr.15: 68778535-68820922 on Build GRCh38
Transcripts
NM_006305.3 NR_026808.1

Gene Symbol
ANP32A-IT1
Entrez Gene ID
80035
Gene Name
ANP32A intronic transcript 1
Gene Aliases
C15orf28, HsT18971, NCRNA00321
UniGene
Hs.662150
Gene Chromosome Location
Chr.15: 68803821-68807101 on Build GRCh38
Transcripts
NM_006305.3 NR_026808.1

SNP Detail

SNP ID Chromosome Location
rs557272835 Chr.15: 68804169-68804169 on Build GRCh38
rs775768311 Chr.15: 68803946-68803946 on Build GRCh38
rs869167212 Chr.15: 68803799-68803799 on Build GRCh38
rs540480675 Chr.15: 68804064-68804064 on Build GRCh38
rs745640113 Chr.15: 68803799-68803801 on Build GRCh38
rs924374876 Chr.15: 68803878-68803878 on Build GRCh38
rs748743398 Chr.15: 68803798-68803799 on Build GRCh38
rs924378569 Chr.15: 68804122-68804122 on Build GRCh38
rs967312807 Chr.15: 68804095-68804095 on Build GRCh38
rs116945256 Chr.15: 68804199-68804199 on Build GRCh38
rs1010600365 Chr.15: 68803947-68803947 on Build GRCh38
rs62008191 Chr.15: 68803985-68803985 on Build GRCh38
rs185118351 Chr.15: 68804065-68804065 on Build GRCh38
rs71402289 Chr.15: 68803842-68803842 on Build GRCh38
rs1004015435 Chr.15: 68803994-68803994 on Build GRCh38
rs982102798 Chr.15: 68803831-68803831 on Build GRCh38
rs902298401 Chr.15: 68803953-68803953 on Build GRCh38
rs926904391 Chr.15: 68803833-68803833 on Build GRCh38
rs772382584 Chr.15: 68803924-68803924 on Build GRCh38
rs943745447 Chr.15: 68803884-68803884 on Build GRCh38
rs983520917 Chr.15: 68804147-68804147 on Build GRCh38
rs1004494697 Chr.15: 68804227-68804227 on Build GRCh38
rs920670520 Chr.15: 68804138-68804138 on Build GRCh38
rs566117691 Chr.15: 68804029-68804029 on Build GRCh38
rs978281323 Chr.15: 68804096-68804096 on Build GRCh38
rs145474475 Chr.15: 68804204-68804204 on Build GRCh38
rs190242545 Chr.15: 68803983-68803983 on Build GRCh38
rs944568783 Chr.15: 68804158-68804158 on Build GRCh38
rs1048285309 Chr.15: 68803950-68803950 on Build GRCh38
rs915411322 Chr.15: 68803854-68803854 on Build GRCh38
rs746964742 Chr.15: 68803800-68803801 on Build GRCh38
rs945523955 Chr.15: 68803868-68803868 on Build GRCh38
rs1022050715 Chr.15: 68803952-68803952 on Build GRCh38
rs537333910 Chr.15: 68804148-68804148 on Build GRCh38
rs943776346 Chr.15: 68804129-68804129 on Build GRCh38
rs764413865 Chr.15: 68804052-68804052 on Build GRCh38
rs35905951 Chr.15: 68804043-68804044 on Build GRCh38
rs1040669216 Chr.15: 68803887-68803887 on Build GRCh38
rs746207964 Chr.15: 68803888-68803888 on Build GRCh38
rs776813187 Chr.15: 68804139-68804139 on Build GRCh38
rs970836721 Chr.15: 68804032-68804032 on Build GRCh38
rs3985625 Chr.15: 68803819-68803820 on Build GRCh38
rs900154802 Chr.15: 68803942-68803942 on Build GRCh38
rs915461275 Chr.15: 68804092-68804092 on Build GRCh38
rs989732180 Chr.15: 68804091-68804091 on Build GRCh38
rs530795971 Chr.15: 68803841-68803841 on Build GRCh38
rs567148649 Chr.15: 68804182-68804182 on Build GRCh38
rs181943134 Chr.15: 68804051-68804051 on Build GRCh38
rs187046225 Chr.15: 68803871-68803871 on Build GRCh38
rs767277392 Chr.15: 68804220-68804220 on Build GRCh38
rs146452520 Chr.15: 68803835-68803835 on Build GRCh38

View Full Product Details