Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30922231-30922723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL19-AS1
Entrez Gene ID
283932
Gene Name
FBXL19 antisense RNA 1 (head to head)
Gene Aliases
NCRNA00095
UniGene
Hs.711535
Gene Chromosome Location
Chr.16: 30919319-30923269 on Build GRCh38
Transcripts
NR_024348.1

SNP Detail

SNP ID Chromosome Location
rs1019603739 Chr.16: 30922537-30922537 on Build GRCh38
rs965923797 Chr.16: 30922386-30922386 on Build GRCh38
rs953375612 Chr.16: 30922575-30922575 on Build GRCh38
rs995687757 Chr.16: 30922524-30922524 on Build GRCh38
rs944938220 Chr.16: 30922319-30922319 on Build GRCh38
rs983738353 Chr.16: 30922308-30922308 on Build GRCh38
rs965354474 Chr.16: 30922543-30922543 on Build GRCh38
rs967128754 Chr.16: 30922589-30922589 on Build GRCh38
rs1027062308 Chr.16: 30922329-30922329 on Build GRCh38
rs921787302 Chr.16: 30922388-30922388 on Build GRCh38
rs1055957128 Chr.16: 30922487-30922487 on Build GRCh38
rs1054501411 Chr.16: 30922632-30922632 on Build GRCh38
rs1025391336 Chr.16: 30922525-30922525 on Build GRCh38
rs894680289 Chr.16: 30922494-30922494 on Build GRCh38
rs1010183334 Chr.16: 30922389-30922389 on Build GRCh38
rs759490863 Chr.16: 30922630-30922631 on Build GRCh38
rs765447318 Chr.16: 30922263-30922263 on Build GRCh38
rs767144129 Chr.16: 30922544-30922544 on Build GRCh38
rs763040109 Chr.16: 30922387-30922387 on Build GRCh38
rs61678005 Chr.16: 30922483-30922483 on Build GRCh38
rs755603269 Chr.16: 30922695-30922695 on Build GRCh38
rs913313972 Chr.16: 30922633-30922633 on Build GRCh38
rs907427168 Chr.16: 30922259-30922259 on Build GRCh38
rs370935618 Chr.16: 30922613-30922613 on Build GRCh38
rs929610323 Chr.16: 30922303-30922303 on Build GRCh38
rs907183160 Chr.16: 30922694-30922694 on Build GRCh38
rs1004635674 Chr.16: 30922532-30922532 on Build GRCh38
rs141345629 Chr.16: 30922581-30922581 on Build GRCh38
rs751298355 Chr.16: 30922448-30922448 on Build GRCh38
rs931481027 Chr.16: 30922666-30922666 on Build GRCh38
rs1054528617 Chr.16: 30922385-30922385 on Build GRCh38
rs962551371 Chr.16: 30922507-30922507 on Build GRCh38
rs1034454920 Chr.16: 30922265-30922265 on Build GRCh38
rs887209414 Chr.16: 30922683-30922683 on Build GRCh38
rs1056011445 Chr.16: 30922651-30922651 on Build GRCh38
rs977120245 Chr.16: 30922600-30922601 on Build GRCh38
rs914327978 Chr.16: 30922394-30922394 on Build GRCh38
rs1039619512 Chr.16: 30922338-30922338 on Build GRCh38
rs188206247 Chr.16: 30922638-30922638 on Build GRCh38
rs912199538 Chr.16: 30922541-30922541 on Build GRCh38
rs975933236 Chr.16: 30922387-30922387 on Build GRCh38
rs907129527 Chr.16: 30922423-30922423 on Build GRCh38
rs900642106 Chr.16: 30922361-30922361 on Build GRCh38
rs986468487 Chr.16: 30922531-30922531 on Build GRCh38
rs946094029 Chr.16: 30922636-30922636 on Build GRCh38
rs912147151 Chr.16: 30922310-30922310 on Build GRCh38
rs945708524 Chr.16: 30922398-30922398 on Build GRCh38
rs775544771 Chr.16: 30922370-30922370 on Build GRCh38
rs894627828 Chr.16: 30922286-30922286 on Build GRCh38
rs1014858264 Chr.16: 30922376-30922376 on Build GRCh38
rs951501143 Chr.16: 30922345-30922345 on Build GRCh38
rs750087630 Chr.16: 30922551-30922551 on Build GRCh38
rs142592691 Chr.16: 30922587-30922587 on Build GRCh38
rs1048527856 Chr.16: 30922667-30922667 on Build GRCh38
rs75930472 Chr.16: 30922457-30922457 on Build GRCh38
rs866045321 Chr.16: 30922620-30922620 on Build GRCh38
rs147834303 Chr.16: 30922431-30922431 on Build GRCh38
rs928906901 Chr.16: 30922454-30922454 on Build GRCh38
rs1028908314 Chr.16: 30922562-30922562 on Build GRCh38
rs753589125 Chr.16: 30922609-30922609 on Build GRCh38
rs1045714636 Chr.16: 30922693-30922693 on Build GRCh38
rs916125091 Chr.16: 30922663-30922663 on Build GRCh38
rs76919728 Chr.16: 30922530-30922530 on Build GRCh38
rs1004324585 Chr.16: 30922685-30922685 on Build GRCh38
rs989963123 Chr.16: 30922578-30922578 on Build GRCh38
rs1045662562 Chr.16: 30922393-30922393 on Build GRCh38
rs116566908 Chr.16: 30922584-30922584 on Build GRCh38
rs569089776 Chr.16: 30922390-30922390 on Build GRCh38
rs938340195 Chr.16: 30922642-30922642 on Build GRCh38
rs151069683 Chr.16: 30922692-30922692 on Build GRCh38

View Full Product Details