Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 16438867-16439359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC75A-AS1
Entrez Gene ID
125144
Gene Name
LRRC75A antisense RNA 1
Gene Aliases
C17orf45, C17orf76-AS1, FAM211A-AS1, NCRNA00188, TSAP19
UniGene
Hs.368934
Gene Chromosome Location
Chr.17: 16438987-16470648 on Build GRCh38
Transcripts
NR_027158.1 NR_027159.1 NR_027160.1 NR_027161.1 NR_027162.1 NR_027163.1 NR_027164.1 NR_027165.1 NR_027166.1 NR_027167.1 NR_027168.2 NR_027169.1 NR_027170.1 NR_027171.1 NR_027172.2 NR_027173.1 NR_027174.1 NR_027175.1 NR_027176.1 NR_027177.1 NR_027178.1 NR_027179.1 NR_027667.1 NR_045021.1 NR_045022.1 NR_045023.1 NR_045024.1 NR_045025.1 NR_045026.1 NR_045027.1 NR_045028.1 NR_045029.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
975921 c.167G>T p.G56V Chr.17: 16439308-16439308 on Build GRCh38
4837589 c.192A>G p.T64T Chr.17: 16439333-16439333 on Build GRCh38
975920 c.18G>A p.L6L Chr.17: 16439159-16439159 on Build GRCh38
4667093 c.186G>A p.R62R Chr.17: 16439327-16439327 on Build GRCh38
7243322 c.153G>T p.V51V Chr.17: 16439294-16439294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897178994 Chr.17: 16438974-16438974 on Build GRCh38
rs554769527 Chr.17: 16439055-16439055 on Build GRCh38
rs772567328 Chr.17: 16439291-16439291 on Build GRCh38
rs758904846 Chr.17: 16439152-16439152 on Build GRCh38
rs895863963 Chr.17: 16439290-16439290 on Build GRCh38
rs921656103 Chr.17: 16438919-16438919 on Build GRCh38
rs772059224 Chr.17: 16439268-16439268 on Build GRCh38
rs886085278 Chr.17: 16439030-16439030 on Build GRCh38
rs745844844 Chr.17: 16439192-16439192 on Build GRCh38
rs78578055 Chr.17: 16439339-16439339 on Build GRCh38
rs529388324 Chr.17: 16439080-16439080 on Build GRCh38
rs981450501 Chr.17: 16438895-16438895 on Build GRCh38
rs766513647 Chr.17: 16439135-16439136 on Build GRCh38
rs1001416138 Chr.17: 16439012-16439012 on Build GRCh38
rs767720637 Chr.17: 16439227-16439227 on Build GRCh38
rs772867384 Chr.17: 16439285-16439285 on Build GRCh38
rs780178748 Chr.17: 16438971-16438971 on Build GRCh38
rs749737521 Chr.17: 16439065-16439065 on Build GRCh38
rs565144098 Chr.17: 16439122-16439122 on Build GRCh38
rs370371402 Chr.17: 16439226-16439226 on Build GRCh38
rs558045180 Chr.17: 16438917-16438917 on Build GRCh38
rs897378420 Chr.17: 16439103-16439103 on Build GRCh38
rs759314862 Chr.17: 16439136-16439136 on Build GRCh38
rs547182395 Chr.17: 16438923-16438923 on Build GRCh38
rs112215248 Chr.17: 16439324-16439324 on Build GRCh38
rs776211049 Chr.17: 16439208-16439208 on Build GRCh38
rs1020174765 Chr.17: 16439027-16439027 on Build GRCh38
rs762819998 Chr.17: 16439129-16439129 on Build GRCh38
rs749562826 Chr.17: 16439241-16439241 on Build GRCh38
rs559176760 Chr.17: 16439032-16439032 on Build GRCh38
rs74356387 Chr.17: 16439077-16439077 on Build GRCh38
rs751875284 Chr.17: 16439216-16439224 on Build GRCh38
rs753111304 Chr.17: 16439232-16439232 on Build GRCh38
rs891416226 Chr.17: 16438939-16438939 on Build GRCh38
rs1041527991 Chr.17: 16439316-16439316 on Build GRCh38
rs759897048 Chr.17: 16439292-16439292 on Build GRCh38
rs1048456796 Chr.17: 16439254-16439254 on Build GRCh38
rs374779510 Chr.17: 16439196-16439196 on Build GRCh38
rs117030628 Chr.17: 16439029-16439029 on Build GRCh38
rs187334792 Chr.17: 16439070-16439070 on Build GRCh38
rs560689683 Chr.17: 16439139-16439139 on Build GRCh38
rs947159460 Chr.17: 16438891-16438891 on Build GRCh38
rs180957165 Chr.17: 16439036-16439036 on Build GRCh38
rs928590642 Chr.17: 16438897-16438897 on Build GRCh38
rs932828688 Chr.17: 16438933-16438933 on Build GRCh38
rs574985574 Chr.17: 16439068-16439068 on Build GRCh38
rs762961422 Chr.17: 16439092-16439093 on Build GRCh38
rs764793846 Chr.17: 16439278-16439278 on Build GRCh38
rs763802927 Chr.17: 16439202-16439202 on Build GRCh38
rs761391025 Chr.17: 16439211-16439211 on Build GRCh38
rs752258634 Chr.17: 16439141-16439141 on Build GRCh38
rs756147319 Chr.17: 16439025-16439025 on Build GRCh38
rs769207648 Chr.17: 16439098-16439098 on Build GRCh38
rs756481718 Chr.17: 16439323-16439323 on Build GRCh38
rs144209033 Chr.17: 16438934-16438934 on Build GRCh38
rs777648572 Chr.17: 16439168-16439168 on Build GRCh38
rs748445703 Chr.17: 16439107-16439107 on Build GRCh38
rs1003345672 Chr.17: 16439124-16439124 on Build GRCh38
rs756375803 Chr.17: 16439166-16439166 on Build GRCh38
rs746810858 Chr.17: 16439273-16439273 on Build GRCh38
rs749366591 Chr.17: 16439171-16439171 on Build GRCh38
rs139940264 Chr.17: 16439256-16439256 on Build GRCh38
rs570047084 Chr.17: 16439007-16439007 on Build GRCh38
rs34602775 Chr.17: 16439163-16439163 on Build GRCh38
rs914490223 Chr.17: 16439145-16439145 on Build GRCh38
rs944278898 Chr.17: 16438946-16438946 on Build GRCh38
rs771058680 Chr.17: 16439177-16439177 on Build GRCh38
rs199930566 Chr.17: 16439148-16439148 on Build GRCh38
rs767391142 Chr.17: 16439316-16439317 on Build GRCh38
rs748476291 Chr.17: 16439155-16439155 on Build GRCh38
rs138791234 Chr.17: 16439161-16439161 on Build GRCh38
rs527445769 Chr.17: 16438918-16438918 on Build GRCh38
rs552605147 Chr.17: 16439235-16439235 on Build GRCh38
rs768391740 Chr.17: 16439274-16439274 on Build GRCh38
rs1035927024 Chr.17: 16439023-16439023 on Build GRCh38
rs35517418 Chr.17: 16439248-16439248 on Build GRCh38
rs370765922 Chr.17: 16439127-16439127 on Build GRCh38
rs931728695 Chr.17: 16439037-16439037 on Build GRCh38
rs763462332 Chr.17: 16439143-16439143 on Build GRCh38
rs572720413 Chr.17: 16439024-16439024 on Build GRCh38
rs529687795 Chr.17: 16438966-16438966 on Build GRCh38
rs780700097 Chr.17: 16439153-16439153 on Build GRCh38
rs1037064373 Chr.17: 16438968-16438968 on Build GRCh38
rs753202805 Chr.17: 16439304-16439304 on Build GRCh38
rs544120910 Chr.17: 16439130-16439130 on Build GRCh38
rs776398476 Chr.17: 16439275-16439275 on Build GRCh38
rs755366009 Chr.17: 16439156-16439156 on Build GRCh38
rs148808845 Chr.17: 16439132-16439132 on Build GRCh38
rs576435026 Chr.17: 16439084-16439084 on Build GRCh38
rs190554319 Chr.17: 16439095-16439095 on Build GRCh38
rs761591619 Chr.17: 16439276-16439276 on Build GRCh38
rs995669209 Chr.17: 16439089-16439089 on Build GRCh38
rs995478585 Chr.17: 16438995-16438995 on Build GRCh38
rs933095766 Chr.17: 16439026-16439026 on Build GRCh38
rs914499931 Chr.17: 16438942-16438942 on Build GRCh38
rs780504115 Chr.17: 16439094-16439094 on Build GRCh38
rs540226461 Chr.17: 16439225-16439225 on Build GRCh38
rs148434726 Chr.17: 16439333-16439333 on Build GRCh38
rs181849171 Chr.17: 16439181-16439181 on Build GRCh38
rs115485976 Chr.17: 16439003-16439003 on Build GRCh38
rs144509918 Chr.17: 16439301-16439301 on Build GRCh38
rs953185472 Chr.17: 16439182-16439182 on Build GRCh38
rs778044610 Chr.17: 16439236-16439236 on Build GRCh38
rs760187092 Chr.17: 16439198-16439198 on Build GRCh38
rs757491538 Chr.17: 16439338-16439338 on Build GRCh38
rs764883574 Chr.17: 16439212-16439212 on Build GRCh38
rs759663891 Chr.17: 16439228-16439228 on Build GRCh38
rs775077520 Chr.17: 16439270-16439270 on Build GRCh38
rs911617332 Chr.17: 16439206-16439206 on Build GRCh38
rs781251268 Chr.17: 16439105-16439105 on Build GRCh38
rs535179522 Chr.17: 16439049-16439049 on Build GRCh38
rs757294091 Chr.17: 16439243-16439243 on Build GRCh38
rs868245645 Chr.17: 16439146-16439146 on Build GRCh38
rs760406845 Chr.17: 16439142-16439142 on Build GRCh38

View Full Product Details