Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 12145061-12145559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM66D
Entrez Gene ID
100132923
Gene Name
family with sequence similarity 66 member D
Gene Aliases
-
UniGene
Hs.546711
Gene Chromosome Location
Chr.8: 12115782-12151189 on Build GRCh38
Transcripts
NR_027425.1

SNP Detail

SNP ID Chromosome Location
rs778180472 Chr.8: 12145231-12145231 on Build GRCh38
rs188722852 Chr.8: 12145192-12145192 on Build GRCh38
rs55786797 Chr.8: 12145230-12145230 on Build GRCh38
rs138618341 Chr.8: 12145390-12145390 on Build GRCh38
rs551673783 Chr.8: 12145324-12145324 on Build GRCh38
rs768260605 Chr.8: 12145341-12145341 on Build GRCh38
rs577869002 Chr.8: 12145249-12145249 on Build GRCh38
rs79789435 Chr.8: 12145312-12145312 on Build GRCh38
rs544390857 Chr.8: 12145210-12145210 on Build GRCh38
rs557748633 Chr.8: 12145141-12145141 on Build GRCh38
rs1036607197 Chr.8: 12145534-12145534 on Build GRCh38
rs4841760 Chr.8: 12145208-12145208 on Build GRCh38
rs747362542 Chr.8: 12145238-12145238 on Build GRCh38
rs542691650 Chr.8: 12145294-12145294 on Build GRCh38
rs1010165725 Chr.8: 12145125-12145125 on Build GRCh38
rs966019434 Chr.8: 12145226-12145226 on Build GRCh38
rs984106086 Chr.8: 12145444-12145444 on Build GRCh38
rs543006151 Chr.8: 12145535-12145535 on Build GRCh38
rs975658253 Chr.8: 12145298-12145298 on Build GRCh38
rs569172537 Chr.8: 12145114-12145114 on Build GRCh38
rs537550402 Chr.8: 12145402-12145402 on Build GRCh38
rs4841759 Chr.8: 12145127-12145127 on Build GRCh38
rs921512063 Chr.8: 12145354-12145354 on Build GRCh38
rs117247188 Chr.8: 12145271-12145271 on Build GRCh38
rs952711304 Chr.8: 12145374-12145374 on Build GRCh38
rs555107133 Chr.8: 12145387-12145387 on Build GRCh38
rs572691521 Chr.8: 12145479-12145479 on Build GRCh38
rs566679358 Chr.8: 12145334-12145334 on Build GRCh38
rs561426238 Chr.8: 12145299-12145299 on Build GRCh38
rs569084480 Chr.8: 12145303-12145303 on Build GRCh38
rs1019759232 Chr.8: 12145162-12145162 on Build GRCh38
rs908638789 Chr.8: 12145515-12145515 on Build GRCh38
rs550750231 Chr.8: 12145302-12145302 on Build GRCh38
rs554431986 Chr.8: 12145471-12145471 on Build GRCh38
rs533621298 Chr.8: 12145190-12145190 on Build GRCh38
rs140824001 Chr.8: 12145300-12145300 on Build GRCh38
rs940095729 Chr.8: 12145529-12145529 on Build GRCh38
rs772121004 Chr.8: 12145517-12145517 on Build GRCh38
rs577831949 Chr.8: 12145439-12145439 on Build GRCh38
rs527663809 Chr.8: 12145290-12145290 on Build GRCh38
rs146950892 Chr.8: 12145177-12145177 on Build GRCh38
rs73663093 Chr.8: 12145279-12145279 on Build GRCh38
rs773703047 Chr.8: 12145418-12145418 on Build GRCh38
rs181201171 Chr.8: 12145383-12145383 on Build GRCh38
rs183826096 Chr.8: 12145426-12145426 on Build GRCh38
rs188226421 Chr.8: 12145440-12145440 on Build GRCh38

View Full Product Details