Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5157509-5157910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1C8P
Entrez Gene ID
340811
Gene Name
aldo-keto reductase family 1 member C8, pseudogene
Gene Aliases
AKR1CL1
UniGene
Hs.282513
Gene Chromosome Location
Chr.10: 5154460-5185187 on Build GRCh38
Transcripts
NR_027916.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1603504 c.249C>G p.V83V Chr.10: 5157716-5157716 on Build GRCh38
5939987 c.272G>A p.R91Q Chr.10: 5157693-5157693 on Build GRCh38
7305683 c.174C>T p.P58P Chr.10: 5157791-5157791 on Build GRCh38
5899005 c.189C>T p.L63L Chr.10: 5157776-5157776 on Build GRCh38
7243827 c.226C>T p.H76Y Chr.10: 5157739-5157739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994780944 Chr.10: 5157531-5157531 on Build GRCh38
rs1012806089 Chr.10: 5157738-5157738 on Build GRCh38
rs935332089 Chr.10: 5157616-5157616 on Build GRCh38
rs782655097 Chr.10: 5157727-5157727 on Build GRCh38
rs140602420 Chr.10: 5157739-5157739 on Build GRCh38
rs782224782 Chr.10: 5157723-5157723 on Build GRCh38
rs782013397 Chr.10: 5157764-5157764 on Build GRCh38
rs1761601 Chr.10: 5157858-5157858 on Build GRCh38
rs551206646 Chr.10: 5157553-5157553 on Build GRCh38
rs912071066 Chr.10: 5157838-5157838 on Build GRCh38
rs371654797 Chr.10: 5157615-5157615 on Build GRCh38
rs944892183 Chr.10: 5157856-5157856 on Build GRCh38
rs971235232 Chr.10: 5157741-5157741 on Build GRCh38
rs560631999 Chr.10: 5157559-5157559 on Build GRCh38
rs1027563582 Chr.10: 5157541-5157541 on Build GRCh38
rs782151837 Chr.10: 5157840-5157840 on Build GRCh38
rs782133624 Chr.10: 5157797-5157797 on Build GRCh38
rs782685472 Chr.10: 5157685-5157685 on Build GRCh38
rs1781935 Chr.10: 5157733-5157733 on Build GRCh38
rs782059942 Chr.10: 5157766-5157766 on Build GRCh38
rs11253019 Chr.10: 5157875-5157875 on Build GRCh38
rs782101260 Chr.10: 5157607-5157607 on Build GRCh38
rs1050688763 Chr.10: 5157826-5157826 on Build GRCh38
rs894133224 Chr.10: 5157876-5157876 on Build GRCh38
rs7912150 Chr.10: 5157584-5157584 on Build GRCh38
rs187531326 Chr.10: 5157544-5157544 on Build GRCh38
rs782334385 Chr.10: 5157726-5157726 on Build GRCh38
rs539095699 Chr.10: 5157781-5157781 on Build GRCh38
rs782101158 Chr.10: 5157791-5157791 on Build GRCh38
rs782369971 Chr.10: 5157767-5157767 on Build GRCh38
rs72776976 Chr.10: 5157812-5157812 on Build GRCh38
rs1009093530 Chr.10: 5157693-5157693 on Build GRCh38
rs367970206 Chr.10: 5157545-5157545 on Build GRCh38
rs897579280 Chr.10: 5157859-5157859 on Build GRCh38
rs897526910 Chr.10: 5157709-5157709 on Build GRCh38
rs915219968 Chr.10: 5157803-5157803 on Build GRCh38
rs982204516 Chr.10: 5157602-5157602 on Build GRCh38
rs142921071 Chr.10: 5157874-5157874 on Build GRCh38
rs782557901 Chr.10: 5157721-5157721 on Build GRCh38
rs958627462 Chr.10: 5157734-5157734 on Build GRCh38
rs1051053639 Chr.10: 5157669-5157669 on Build GRCh38
rs565058124 Chr.10: 5157712-5157712 on Build GRCh38
rs115744677 Chr.10: 5157715-5157715 on Build GRCh38
rs781878747 Chr.10: 5157653-5157653 on Build GRCh38
rs782506271 Chr.10: 5157680-5157680 on Build GRCh38
rs114988413 Chr.10: 5157799-5157799 on Build GRCh38
rs915501912 Chr.10: 5157629-5157629 on Build GRCh38
rs958576589 Chr.10: 5157548-5157548 on Build GRCh38
rs782530875 Chr.10: 5157873-5157873 on Build GRCh38
rs113877453 Chr.10: 5157583-5157583 on Build GRCh38

View Full Product Details