Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5161558-5162107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1C8P
Entrez Gene ID
340811
Gene Name
aldo-keto reductase family 1 member C8, pseudogene
Gene Aliases
AKR1CL1
UniGene
Hs.282513
Gene Chromosome Location
Chr.10: 5154460-5185187 on Build GRCh38
Transcripts
NR_027916.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5086603 c.378+2T>C p.? Chr.10: 5161854-5161854 on Build GRCh38
918802 c.272C>G p.T91S Chr.10: 5161962-5161962 on Build GRCh38
3438554 c.311G>A p.R104K Chr.10: 5161923-5161923 on Build GRCh38
918801 c.277T>G p.F93V Chr.10: 5161957-5161957 on Build GRCh38
3438553 c.334G>A p.D112N Chr.10: 5161900-5161900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781890026 Chr.10: 5161874-5161874 on Build GRCh38
rs782515793 Chr.10: 5162023-5162023 on Build GRCh38
rs781956278 Chr.10: 5161771-5161771 on Build GRCh38
rs782812658 Chr.10: 5161943-5161943 on Build GRCh38
rs367949592 Chr.10: 5161845-5161845 on Build GRCh38
rs958219633 Chr.10: 5161662-5161662 on Build GRCh38
rs1046573117 Chr.10: 5162049-5162049 on Build GRCh38
rs782467293 Chr.10: 5161876-5161876 on Build GRCh38
rs370608265 Chr.10: 5161751-5161751 on Build GRCh38
rs569529340 Chr.10: 5161600-5161600 on Build GRCh38
rs369301573 Chr.10: 5162013-5162013 on Build GRCh38
rs376085953 Chr.10: 5161790-5161790 on Build GRCh38
rs551105957 Chr.10: 5161992-5161992 on Build GRCh38
rs192359311 Chr.10: 5161835-5161835 on Build GRCh38
rs782391837 Chr.10: 5161832-5161832 on Build GRCh38
rs529025383 Chr.10: 5161991-5161991 on Build GRCh38
rs201532847 Chr.10: 5161716-5161716 on Build GRCh38
rs545709193 Chr.10: 5161851-5161851 on Build GRCh38
rs782217472 Chr.10: 5161827-5161827 on Build GRCh38
rs782126074 Chr.10: 5161778-5161778 on Build GRCh38
rs568506750 Chr.10: 5161802-5161802 on Build GRCh38
rs782647738 Chr.10: 5161899-5161899 on Build GRCh38
rs574094554 Chr.10: 5161734-5161734 on Build GRCh38
rs782258501 Chr.10: 5161582-5161582 on Build GRCh38
rs782029100 Chr.10: 5161623-5161623 on Build GRCh38
rs782443283 Chr.10: 5161813-5161813 on Build GRCh38
rs782592748 Chr.10: 5161963-5161963 on Build GRCh38
rs142886454 Chr.10: 5161682-5161682 on Build GRCh38
rs781802842 Chr.10: 5161855-5161855 on Build GRCh38
rs11253020 Chr.10: 5161616-5161616 on Build GRCh38
rs782202833 Chr.10: 5161810-5161810 on Build GRCh38
rs902678845 Chr.10: 5162053-5162053 on Build GRCh38
rs782432769 Chr.10: 5161896-5161896 on Build GRCh38
rs782740447 Chr.10: 5161999-5161999 on Build GRCh38
rs17134472 Chr.10: 5161640-5161640 on Build GRCh38
rs546222845 Chr.10: 5161908-5161908 on Build GRCh38
rs371026070 Chr.10: 5162005-5162005 on Build GRCh38
rs782659030 Chr.10: 5161838-5161838 on Build GRCh38
rs376446369 Chr.10: 5161998-5161998 on Build GRCh38
rs375029592 Chr.10: 5161725-5161725 on Build GRCh38
rs782474467 Chr.10: 5161898-5161898 on Build GRCh38
rs954525473 Chr.10: 5161690-5161690 on Build GRCh38
rs781863427 Chr.10: 5162001-5162001 on Build GRCh38
rs782773337 Chr.10: 5161779-5161779 on Build GRCh38
rs782303060 Chr.10: 5161905-5161905 on Build GRCh38
rs782178456 Chr.10: 5161968-5161968 on Build GRCh38
rs555885461 Chr.10: 5161713-5161713 on Build GRCh38
rs782410368 Chr.10: 5161746-5161746 on Build GRCh38
rs782408705 Chr.10: 5161958-5161958 on Build GRCh38
rs1046522398 Chr.10: 5161872-5161872 on Build GRCh38
rs782149443 Chr.10: 5161919-5161919 on Build GRCh38
rs782028306 Chr.10: 5161780-5161780 on Build GRCh38
rs782694106 Chr.10: 5161882-5161882 on Build GRCh38
rs78245564 Chr.10: 5161983-5161983 on Build GRCh38
rs916846407 Chr.10: 5162030-5162030 on Build GRCh38
rs782820201 Chr.10: 5161855-5161855 on Build GRCh38
rs781950859 Chr.10: 5161988-5161988 on Build GRCh38
rs782462971 Chr.10: 5162056-5162056 on Build GRCh38
rs148533013 Chr.10: 5161586-5161586 on Build GRCh38
rs782208201 Chr.10: 5161909-5161909 on Build GRCh38
rs376200113 Chr.10: 5161953-5161953 on Build GRCh38
rs782381901 Chr.10: 5161912-5161912 on Build GRCh38
rs782573822 Chr.10: 5161728-5161728 on Build GRCh38
rs1013319832 Chr.10: 5161601-5161601 on Build GRCh38
rs11253021 Chr.10: 5161901-5161901 on Build GRCh38
rs367590462 Chr.10: 5161965-5161965 on Build GRCh38
rs558141746 Chr.10: 5161631-5161631 on Build GRCh38
rs782131299 Chr.10: 5161990-5161990 on Build GRCh38
rs552121710 Chr.10: 5162080-5162080 on Build GRCh38
rs782401730 Chr.10: 5161735-5161735 on Build GRCh38
rs371778963 Chr.10: 5161916-5161916 on Build GRCh38
rs374383879 Chr.10: 5161841-5161841 on Build GRCh38
rs782362504 Chr.10: 5161985-5161985 on Build GRCh38
rs782396787 Chr.10: 5161844-5161844 on Build GRCh38
rs781941150 Chr.10: 5161747-5161747 on Build GRCh38
rs370199611 Chr.10: 5161794-5161794 on Build GRCh38
rs782671902 Chr.10: 5162009-5162009 on Build GRCh38
rs782630320 Chr.10: 5161740-5161740 on Build GRCh38
rs782442873 Chr.10: 5162015-5162015 on Build GRCh38
rs188902432 Chr.10: 5161653-5161653 on Build GRCh38
rs1055724242 Chr.10: 5161583-5161583 on Build GRCh38
rs781953357 Chr.10: 5161773-5161773 on Build GRCh38
rs11325738 Chr.10: 5161982-5161982 on Build GRCh38
rs781839150 Chr.10: 5161894-5161894 on Build GRCh38
rs374458601 Chr.10: 5161795-5161795 on Build GRCh38
rs543081605 Chr.10: 5161711-5161711 on Build GRCh38
rs782328101 Chr.10: 5161923-5161923 on Build GRCh38
rs782039404 Chr.10: 5161637-5161637 on Build GRCh38
rs999267044 Chr.10: 5161896-5161896 on Build GRCh38
rs782556138 Chr.10: 5161944-5161944 on Build GRCh38
rs781862761 Chr.10: 5161806-5161806 on Build GRCh38
rs782776445 Chr.10: 5161804-5161804 on Build GRCh38
rs780130478 Chr.10: 5161820-5161820 on Build GRCh38
rs3750573 Chr.10: 5161785-5161785 on Build GRCh38
rs747954344 Chr.10: 5162018-5162018 on Build GRCh38
rs782303649 Chr.10: 5161741-5161741 on Build GRCh38
rs77303730 Chr.10: 5161809-5161809 on Build GRCh38
rs782548198 Chr.10: 5161807-5161807 on Build GRCh38
rs782116293 Chr.10: 5161942-5161942 on Build GRCh38
rs949618105 Chr.10: 5162044-5162044 on Build GRCh38
rs782741030 Chr.10: 5161846-5161846 on Build GRCh38
rs931584317 Chr.10: 5161811-5161811 on Build GRCh38
rs61729616 Chr.10: 5161902-5161902 on Build GRCh38
rs781921165 Chr.10: 5161930-5161930 on Build GRCh38
rs921795031 Chr.10: 5161684-5161684 on Build GRCh38
rs373454628 Chr.10: 5161983-5161983 on Build GRCh38
rs782287749 Chr.10: 5161839-5161839 on Build GRCh38
rs987263908 Chr.10: 5161696-5161696 on Build GRCh38
rs781873814 Chr.10: 5161935-5161935 on Build GRCh38
rs79777776 Chr.10: 5162004-5162004 on Build GRCh38

View Full Product Details