Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 43897648-43898196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH3
Entrez Gene ID
221120
Gene Name
alkB homolog 3, alpha-ketoglutaratedependent dioxygenase
Gene Aliases
ABH3, DEPC-1, DEPC1, PCA1, hABH3
UniGene
Hs.720708
Gene Chromosome Location
Chr.11: 43880807-43920275 on Build GRCh38
Transcripts
NM_139178.3 NR_026952.1

Gene Symbol
SEC14L1P1
Entrez Gene ID
729799
Gene Name
SEC14 like 1 pseudogene 1
Gene Aliases
-
UniGene
Hs.368920
Gene Chromosome Location
Chr.11: 43897303-43899888 on Build GRCh38
Transcripts
NM_139178.3 NR_026952.1

SNP Detail

SNP ID Chromosome Location
rs941144223 Chr.11: 43897738-43897738 on Build GRCh38
rs921483999 Chr.11: 43897777-43897777 on Build GRCh38
rs975981132 Chr.11: 43897749-43897749 on Build GRCh38
rs1056249906 Chr.11: 43898136-43898136 on Build GRCh38
rs763806732 Chr.11: 43898141-43898141 on Build GRCh38
rs904166904 Chr.11: 43897820-43897820 on Build GRCh38
rs77207849 Chr.11: 43897707-43897707 on Build GRCh38
rs955581030 Chr.11: 43898044-43898044 on Build GRCh38
rs772329155 Chr.11: 43898176-43898176 on Build GRCh38
rs960783195 Chr.11: 43898163-43898163 on Build GRCh38
rs964941430 Chr.11: 43897912-43897912 on Build GRCh38
rs934199970 Chr.11: 43897809-43897809 on Build GRCh38
rs974998201 Chr.11: 43897934-43897934 on Build GRCh38
rs921401605 Chr.11: 43898031-43898031 on Build GRCh38
rs567023996 Chr.11: 43898125-43898125 on Build GRCh38
rs1008569437 Chr.11: 43898058-43898058 on Build GRCh38
rs951025795 Chr.11: 43897979-43897979 on Build GRCh38
rs114924068 Chr.11: 43898112-43898112 on Build GRCh38
rs568085520 Chr.11: 43898078-43898078 on Build GRCh38
rs751171260 Chr.11: 43897895-43897895 on Build GRCh38
rs866709837 Chr.11: 43898017-43898017 on Build GRCh38
rs749403213 Chr.11: 43898117-43898117 on Build GRCh38
rs920604210 Chr.11: 43897935-43897935 on Build GRCh38
rs537881865 Chr.11: 43898159-43898159 on Build GRCh38
rs535509133 Chr.11: 43898087-43898087 on Build GRCh38
rs907955703 Chr.11: 43897970-43897970 on Build GRCh38
rs1011136545 Chr.11: 43897676-43897676 on Build GRCh38
rs895726493 Chr.11: 43897720-43897720 on Build GRCh38
rs909652205 Chr.11: 43897721-43897721 on Build GRCh38
rs1053320734 Chr.11: 43898047-43898047 on Build GRCh38
rs879241711 Chr.11: 43897773-43897773 on Build GRCh38
rs760213311 Chr.11: 43898039-43898039 on Build GRCh38
rs146850479 Chr.11: 43897989-43897989 on Build GRCh38
rs753176015 Chr.11: 43898163-43898164 on Build GRCh38
rs964818526 Chr.11: 43898096-43898096 on Build GRCh38
rs749937139 Chr.11: 43897715-43897715 on Build GRCh38
rs558998580 Chr.11: 43897687-43897687 on Build GRCh38
rs143752599 Chr.11: 43897686-43897686 on Build GRCh38
rs1049426 Chr.11: 43897804-43897804 on Build GRCh38
rs891541826 Chr.11: 43898049-43898049 on Build GRCh38
rs1003394201 Chr.11: 43898134-43898134 on Build GRCh38
rs1031406543 Chr.11: 43897857-43897857 on Build GRCh38
rs527586643 Chr.11: 43897888-43897888 on Build GRCh38
rs1044292390 Chr.11: 43898010-43898010 on Build GRCh38
rs755338421 Chr.11: 43897929-43897929 on Build GRCh38
rs939869996 Chr.11: 43897974-43897974 on Build GRCh38
rs559029075 Chr.11: 43898151-43898151 on Build GRCh38
rs11037720 Chr.11: 43897861-43897861 on Build GRCh38
rs570721933 Chr.11: 43897973-43897973 on Build GRCh38
rs866110701 Chr.11: 43897968-43897968 on Build GRCh38
rs748531849 Chr.11: 43898006-43898006 on Build GRCh38
rs1016590440 Chr.11: 43897992-43897992 on Build GRCh38
rs1054451472 Chr.11: 43898109-43898109 on Build GRCh38
rs541641752 Chr.11: 43897860-43897860 on Build GRCh38
rs184421794 Chr.11: 43897906-43897906 on Build GRCh38
rs116511955 Chr.11: 43897703-43897703 on Build GRCh38
rs1013035405 Chr.11: 43897927-43897927 on Build GRCh38
rs1023057132 Chr.11: 43897805-43897805 on Build GRCh38
rs1019087988 Chr.11: 43898077-43898077 on Build GRCh38
rs541395893 Chr.11: 43898157-43898157 on Build GRCh38
rs535611085 Chr.11: 43897990-43897990 on Build GRCh38
rs757880281 Chr.11: 43897782-43897782 on Build GRCh38
rs1012804843 Chr.11: 43897748-43897748 on Build GRCh38
rs745864562 Chr.11: 43897727-43897727 on Build GRCh38
rs2460130 Chr.11: 43898144-43898144 on Build GRCh38
rs999847148 Chr.11: 43898034-43898034 on Build GRCh38
rs530186070 Chr.11: 43897847-43897847 on Build GRCh38
rs992221042 Chr.11: 43898164-43898164 on Build GRCh38
rs562877661 Chr.11: 43897772-43897772 on Build GRCh38
rs983963296 Chr.11: 43897969-43897969 on Build GRCh38
rs1044491586 Chr.11: 43898120-43898120 on Build GRCh38
rs34844726 Chr.11: 43898069-43898071 on Build GRCh38
rs775095622 Chr.11: 43897993-43897993 on Build GRCh38
rs1049424 Chr.11: 43897758-43897758 on Build GRCh38
rs778322298 Chr.11: 43898106-43898106 on Build GRCh38
rs549734071 Chr.11: 43898011-43898011 on Build GRCh38

View Full Product Details