Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2369981-2370537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA17P
Entrez Gene ID
650655
Gene Name
ATP binding cassette subfamily A member 17, pseudogene
Gene Aliases
ABCA17
UniGene
Hs.376281
Gene Chromosome Location
Chr.16: 2340922-2426699 on Build GRCh38
Transcripts
NR_003574.1

SNP Detail

SNP ID Chromosome Location
rs750256436 Chr.16: 2370198-2370198 on Build GRCh38
rs1009978769 Chr.16: 2370156-2370156 on Build GRCh38
rs753235737 Chr.16: 2370402-2370402 on Build GRCh38
rs942896197 Chr.16: 2370408-2370408 on Build GRCh38
rs945724576 Chr.16: 2370440-2370440 on Build GRCh38
rs1023192803 Chr.16: 2370385-2370385 on Build GRCh38
rs561995272 Chr.16: 2370454-2370454 on Build GRCh38
rs1016883515 Chr.16: 2370334-2370334 on Build GRCh38
rs371364275 Chr.16: 2370382-2370382 on Build GRCh38
rs766084792 Chr.16: 2370356-2370356 on Build GRCh38
rs921375976 Chr.16: 2370222-2370227 on Build GRCh38
rs1033081171 Chr.16: 2370080-2370080 on Build GRCh38
rs988347103 Chr.16: 2370133-2370133 on Build GRCh38
rs944216866 Chr.16: 2370188-2370188 on Build GRCh38
rs972824680 Chr.16: 2370338-2370338 on Build GRCh38
rs907015675 Chr.16: 2370172-2370172 on Build GRCh38
rs775321725 Chr.16: 2370496-2370496 on Build GRCh38
rs564709416 Chr.16: 2370410-2370410 on Build GRCh38
rs958336296 Chr.16: 2370086-2370086 on Build GRCh38
rs185396884 Chr.16: 2370396-2370396 on Build GRCh38
rs934102634 Chr.16: 2370390-2370390 on Build GRCh38
rs962724310 Chr.16: 2370335-2370335 on Build GRCh38
rs1014064259 Chr.16: 2370005-2370005 on Build GRCh38
rs767478531 Chr.16: 2370149-2370149 on Build GRCh38
rs28402683 Chr.16: 2370249-2370249 on Build GRCh38
rs1014116759 Chr.16: 2370357-2370357 on Build GRCh38
rs905642789 Chr.16: 2370393-2370393 on Build GRCh38
rs1021163433 Chr.16: 2370473-2370473 on Build GRCh38
rs918697381 Chr.16: 2370351-2370351 on Build GRCh38
rs988401194 Chr.16: 2370425-2370425 on Build GRCh38
rs1054084126 Chr.16: 2370060-2370060 on Build GRCh38
rs909238631 Chr.16: 2370092-2370092 on Build GRCh38
rs958579370 Chr.16: 2370265-2370265 on Build GRCh38
rs761697483 Chr.16: 2370145-2370145 on Build GRCh38
rs555607784 Chr.16: 2370185-2370185 on Build GRCh38
rs745827509 Chr.16: 2370073-2370074 on Build GRCh38
rs1036959209 Chr.16: 2370313-2370313 on Build GRCh38
rs987349486 Chr.16: 2370394-2370394 on Build GRCh38
rs942950042 Chr.16: 2370039-2370039 on Build GRCh38
rs539898619 Chr.16: 2370395-2370395 on Build GRCh38
rs754563857 Chr.16: 2370477-2370477 on Build GRCh38
rs115525912 Chr.16: 2370331-2370331 on Build GRCh38
rs537837744 Chr.16: 2370288-2370288 on Build GRCh38
rs914104453 Chr.16: 2370170-2370170 on Build GRCh38
rs767775296 Chr.16: 2370073-2370073 on Build GRCh38
rs796458956 Chr.16: 2370073-2370073 on Build GRCh38
rs563183874 Chr.16: 2370433-2370433 on Build GRCh38
rs1054743848 Chr.16: 2370430-2370430 on Build GRCh38
rs145430005 Chr.16: 2370424-2370424 on Build GRCh38
rs371728641 Chr.16: 2370130-2370130 on Build GRCh38
rs970190075 Chr.16: 2370050-2370050 on Build GRCh38
rs531955470 Chr.16: 2370444-2370444 on Build GRCh38
rs557377241 Chr.16: 2370293-2370293 on Build GRCh38
rs553464507 Chr.16: 2370417-2370417 on Build GRCh38
rs199635624 Chr.16: 2370077-2370077 on Build GRCh38
rs1020491663 Chr.16: 2370168-2370168 on Build GRCh38
rs939954072 Chr.16: 2370304-2370304 on Build GRCh38
rs978466413 Chr.16: 2370071-2370073 on Build GRCh38
rs574147252 Chr.16: 2370419-2370419 on Build GRCh38
rs545385041 Chr.16: 2370478-2370478 on Build GRCh38
rs868285601 Chr.16: 2370486-2370486 on Build GRCh38
rs932698969 Chr.16: 2370239-2370239 on Build GRCh38
rs199555354 Chr.16: 2370069-2370070 on Build GRCh38

View Full Product Details