Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2406494-2407043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA17P
Entrez Gene ID
650655
Gene Name
ATP binding cassette subfamily A member 17, pseudogene
Gene Aliases
ABCA17
UniGene
Hs.376281
Gene Chromosome Location
Chr.16: 2340922-2426699 on Build GRCh38
Transcripts
NR_003574.1

SNP Detail

SNP ID Chromosome Location
rs547607434 Chr.16: 2406960-2406960 on Build GRCh38
rs1057133501 Chr.16: 2407013-2407013 on Build GRCh38
rs780592320 Chr.16: 2406580-2406583 on Build GRCh38
rs1038786337 Chr.16: 2406859-2406859 on Build GRCh38
rs546714333 Chr.16: 2406680-2406680 on Build GRCh38
rs777900176 Chr.16: 2406821-2406821 on Build GRCh38
rs942483082 Chr.16: 2406923-2406923 on Build GRCh38
rs377393959 Chr.16: 2406713-2406713 on Build GRCh38
rs191034850 Chr.16: 2406675-2406675 on Build GRCh38
rs528571039 Chr.16: 2406882-2406882 on Build GRCh38
rs778523117 Chr.16: 2406901-2406901 on Build GRCh38
rs556224746 Chr.16: 2406595-2406595 on Build GRCh38
rs559847069 Chr.16: 2406860-2406860 on Build GRCh38
rs752274316 Chr.16: 2406956-2406956 on Build GRCh38
rs950939424 Chr.16: 2406780-2406780 on Build GRCh38
rs991591256 Chr.16: 2406759-2406759 on Build GRCh38
rs541656520 Chr.16: 2406683-2406683 on Build GRCh38
rs745435584 Chr.16: 2406621-2406622 on Build GRCh38
rs1042687244 Chr.16: 2406902-2406902 on Build GRCh38
rs987114544 Chr.16: 2406840-2406840 on Build GRCh38
rs984044858 Chr.16: 2406791-2406791 on Build GRCh38
rs57562614 Chr.16: 2406855-2406855 on Build GRCh38
rs1011224907 Chr.16: 2406658-2406658 on Build GRCh38
rs929554454 Chr.16: 2406828-2406828 on Build GRCh38
rs935161491 Chr.16: 2406672-2406672 on Build GRCh38
rs919558506 Chr.16: 2406822-2406822 on Build GRCh38
rs1023110748 Chr.16: 2406794-2406794 on Build GRCh38
rs765938291 Chr.16: 2406764-2406764 on Build GRCh38
rs142182396 Chr.16: 2406853-2406853 on Build GRCh38
rs1025240096 Chr.16: 2406993-2406993 on Build GRCh38
rs574825300 Chr.16: 2406793-2406793 on Build GRCh38
rs979395250 Chr.16: 2406858-2406858 on Build GRCh38
rs554855409 Chr.16: 2406687-2406687 on Build GRCh38
rs915055352 Chr.16: 2406884-2406884 on Build GRCh38
rs764509013 Chr.16: 2406790-2406790 on Build GRCh38
rs890169549 Chr.16: 2406564-2406564 on Build GRCh38
rs538755441 Chr.16: 2406599-2406599 on Build GRCh38
rs890952313 Chr.16: 2406919-2406919 on Build GRCh38
rs57499237 Chr.16: 2406798-2406798 on Build GRCh38
rs1020893075 Chr.16: 2406677-2406677 on Build GRCh38
rs1039430063 Chr.16: 2406941-2406941 on Build GRCh38
rs536535240 Chr.16: 2406549-2406549 on Build GRCh38
rs567595052 Chr.16: 2406520-2406520 on Build GRCh38
rs995094815 Chr.16: 2406734-2406734 on Build GRCh38
rs898162541 Chr.16: 2406731-2406731 on Build GRCh38
rs893653527 Chr.16: 2406617-2406617 on Build GRCh38
rs186564557 Chr.16: 2406598-2406598 on Build GRCh38
rs1051949308 Chr.16: 2406563-2406563 on Build GRCh38
rs995147228 Chr.16: 2406992-2406992 on Build GRCh38
rs182410171 Chr.16: 2406845-2406845 on Build GRCh38

View Full Product Details