Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42868838-42869387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOC4P
Entrez Gene ID
90586
Gene Name
amine oxidase, copper containing 4, pseudogene
Gene Aliases
AOC4, UPAT
UniGene
Hs.367781
Gene Chromosome Location
Chr.17: 42867145-42869217 on Build GRCh38
Transcripts
NR_002773.1

SNP Detail

SNP ID Chromosome Location
rs945719579 Chr.17: 42869310-42869310 on Build GRCh38
rs112249308 Chr.17: 42869230-42869231 on Build GRCh38
rs576857057 Chr.17: 42869063-42869063 on Build GRCh38
rs764786842 Chr.17: 42868958-42868958 on Build GRCh38
rs746965913 Chr.17: 42869064-42869064 on Build GRCh38
rs929346936 Chr.17: 42869056-42869056 on Build GRCh38
rs536535468 Chr.17: 42868891-42868891 on Build GRCh38
rs548442066 Chr.17: 42868940-42868940 on Build GRCh38
rs1018161766 Chr.17: 42869099-42869099 on Build GRCh38
rs867434264 Chr.17: 42869227-42869227 on Build GRCh38
rs569336001 Chr.17: 42869004-42869004 on Build GRCh38
rs191438372 Chr.17: 42869124-42869124 on Build GRCh38
rs917455179 Chr.17: 42868973-42868973 on Build GRCh38
rs1027694523 Chr.17: 42869161-42869161 on Build GRCh38
rs1007154981 Chr.17: 42869097-42869097 on Build GRCh38
rs950036905 Chr.17: 42869086-42869086 on Build GRCh38
rs905085838 Chr.17: 42869102-42869102 on Build GRCh38
rs905132195 Chr.17: 42868911-42868911 on Build GRCh38
rs764512754 Chr.17: 42869043-42869043 on Build GRCh38
rs771629023 Chr.17: 42868950-42868950 on Build GRCh38
rs879514482 Chr.17: 42869231-42869231 on Build GRCh38
rs773321386 Chr.17: 42869004-42869004 on Build GRCh38
rs757012477 Chr.17: 42869066-42869066 on Build GRCh38
rs775609143 Chr.17: 42869342-42869342 on Build GRCh38
rs1047795134 Chr.17: 42869074-42869074 on Build GRCh38
rs552874547 Chr.17: 42869172-42869172 on Build GRCh38
rs780917604 Chr.17: 42869083-42869083 on Build GRCh38
rs950269883 Chr.17: 42868990-42868990 on Build GRCh38
rs1020699337 Chr.17: 42869332-42869332 on Build GRCh38
rs1009444044 Chr.17: 42869285-42869285 on Build GRCh38
rs752249240 Chr.17: 42869005-42869005 on Build GRCh38
rs1044403793 Chr.17: 42869092-42869092 on Build GRCh38
rs566393688 Chr.17: 42868857-42868857 on Build GRCh38
rs1024811656 Chr.17: 42869184-42869184 on Build GRCh38
rs749351202 Chr.17: 42869026-42869026 on Build GRCh38
rs113330268 Chr.17: 42869123-42869123 on Build GRCh38
rs746651874 Chr.17: 42868949-42868949 on Build GRCh38
rs767844777 Chr.17: 42868860-42868860 on Build GRCh38
rs763459522 Chr.17: 42869034-42869034 on Build GRCh38
rs747655629 Chr.17: 42868931-42868931 on Build GRCh38
rs545787683 Chr.17: 42869277-42869277 on Build GRCh38
rs778424583 Chr.17: 42868917-42868917 on Build GRCh38
rs954360916 Chr.17: 42868983-42868983 on Build GRCh38
rs78121341 Chr.17: 42869231-42869231 on Build GRCh38
rs11871375 Chr.17: 42869006-42869006 on Build GRCh38
rs779750396 Chr.17: 42868903-42868903 on Build GRCh38
rs950768601 Chr.17: 42869167-42869167 on Build GRCh38
rs920774376 Chr.17: 42869017-42869017 on Build GRCh38
rs954475204 Chr.17: 42869346-42869346 on Build GRCh38
rs80177250 Chr.17: 42869230-42869230 on Build GRCh38
rs574650465 Chr.17: 42869228-42869228 on Build GRCh38
rs754887910 Chr.17: 42868916-42868916 on Build GRCh38
rs558629322 Chr.17: 42869027-42869027 on Build GRCh38
rs912866102 Chr.17: 42869300-42869300 on Build GRCh38
rs771855248 Chr.17: 42869316-42869316 on Build GRCh38
rs777641998 Chr.17: 42869007-42869007 on Build GRCh38
rs772831648 Chr.17: 42868965-42868965 on Build GRCh38
rs568233501 Chr.17: 42868898-42868898 on Build GRCh38
rs755948843 Chr.17: 42868897-42868897 on Build GRCh38
rs971563931 Chr.17: 42869189-42869189 on Build GRCh38
rs749204505 Chr.17: 42868912-42868912 on Build GRCh38
rs529537449 Chr.17: 42868963-42868963 on Build GRCh38
rs1020751736 Chr.17: 42868978-42868978 on Build GRCh38

View Full Product Details