Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 64974653-64975207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMZ2P1
Entrez Gene ID
201283
Gene Name
archaelysin family metallopeptidase 2 pseudogene 1
Gene Aliases
-
UniGene
Hs.396447
Gene Chromosome Location
Chr.17: 64966550-64975585 on Build GRCh38
Transcripts
NR_026903.1

SNP Detail

SNP ID Chromosome Location
rs879555680 Chr.17: 64975099-64975099 on Build GRCh38
rs1016768002 Chr.17: 64975054-64975054 on Build GRCh38
rs953901932 Chr.17: 64974723-64974723 on Build GRCh38
rs1020653019 Chr.17: 64975003-64975003 on Build GRCh38
rs973186894 Chr.17: 64974834-64974834 on Build GRCh38
rs576706423 Chr.17: 64974718-64974718 on Build GRCh38
rs755194630 Chr.17: 64974945-64974945 on Build GRCh38
rs893544619 Chr.17: 64974680-64974680 on Build GRCh38
rs1011044803 Chr.17: 64974687-64974687 on Build GRCh38
rs1054643965 Chr.17: 64974981-64974981 on Build GRCh38
rs950026322 Chr.17: 64974871-64974871 on Build GRCh38
rs556803271 Chr.17: 64974711-64974711 on Build GRCh38
rs532744958 Chr.17: 64975037-64975037 on Build GRCh38
rs543084152 Chr.17: 64975070-64975070 on Build GRCh38
rs966311254 Chr.17: 64974699-64974699 on Build GRCh38
rs985755967 Chr.17: 64975040-64975040 on Build GRCh38
rs756619624 Chr.17: 64975143-64975143 on Build GRCh38
rs751794025 Chr.17: 64974698-64974698 on Build GRCh38
rs937627400 Chr.17: 64975153-64975153 on Build GRCh38
rs927251593 Chr.17: 64975131-64975131 on Build GRCh38
rs1046055890 Chr.17: 64974941-64974941 on Build GRCh38
rs981428846 Chr.17: 64975114-64975114 on Build GRCh38
rs918675201 Chr.17: 64975093-64975093 on Build GRCh38
rs752723625 Chr.17: 64974984-64974984 on Build GRCh38
rs902226365 Chr.17: 64975018-64975018 on Build GRCh38
rs985297395 Chr.17: 64974729-64974729 on Build GRCh38
rs1029562650 Chr.17: 64975035-64975035 on Build GRCh38
rs147366503 Chr.17: 64974866-64974866 on Build GRCh38
rs997787545 Chr.17: 64975019-64975019 on Build GRCh38
rs555635566 Chr.17: 64974857-64974857 on Build GRCh38
rs12948019 Chr.17: 64975178-64975178 on Build GRCh38
rs150027633 Chr.17: 64975177-64975177 on Build GRCh38
rs547629802 Chr.17: 64974925-64974925 on Build GRCh38
rs879803343 Chr.17: 64974796-64974796 on Build GRCh38
rs962825279 Chr.17: 64975079-64975079 on Build GRCh38
rs1010658719 Chr.17: 64974988-64974988 on Build GRCh38
rs540690722 Chr.17: 64974947-64974947 on Build GRCh38
rs563588149 Chr.17: 64975090-64975090 on Build GRCh38
rs140092768 Chr.17: 64974968-64974968 on Build GRCh38
rs542406719 Chr.17: 64974815-64974815 on Build GRCh38
rs182046659 Chr.17: 64975167-64975167 on Build GRCh38
rs941471780 Chr.17: 64974829-64974829 on Build GRCh38

View Full Product Details