Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200730925-200731474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOX3P-AOX2P
Entrez Gene ID
107161151
Gene Name
AOX3P-AOX2P readthrough
Gene Aliases
AOX3/AOX3L1
UniGene
Hs.148269
Gene Chromosome Location
Chr.2: 200695723-200794915 on Build GRCh38
Transcripts
NR_135011.1 NR_135012.1 NR_037886.1

Gene Symbol
LOC100507140
Entrez Gene ID
100507140
Gene Name
uncharacterized LOC100507140
Gene Aliases
-
UniGene
Hs.382116
Gene Chromosome Location
Chr.2: 200712305-200735177 on Build GRCh38
Transcripts
NR_135011.1 NR_135012.1 NR_037886.1

SNP Detail

SNP ID Chromosome Location
rs34853557 Chr.2: 200731236-200731237 on Build GRCh38
rs529604456 Chr.2: 200731088-200731088 on Build GRCh38
rs781082607 Chr.2: 200731058-200731058 on Build GRCh38
rs991392328 Chr.2: 200731403-200731403 on Build GRCh38
rs994128501 Chr.2: 200731247-200731247 on Build GRCh38
rs192710909 Chr.2: 200731311-200731311 on Build GRCh38
rs966795156 Chr.2: 200731187-200731187 on Build GRCh38
rs920930671 Chr.2: 200731026-200731026 on Build GRCh38
rs193222706 Chr.2: 200731041-200731041 on Build GRCh38
rs749550068 Chr.2: 200731063-200731063 on Build GRCh38
rs912710812 Chr.2: 200731045-200731045 on Build GRCh38
rs949700110 Chr.2: 200731092-200731092 on Build GRCh38
rs1024217172 Chr.2: 200731164-200731164 on Build GRCh38
rs148403115 Chr.2: 200731358-200731358 on Build GRCh38
rs990479561 Chr.2: 200731306-200731306 on Build GRCh38
rs895152670 Chr.2: 200731165-200731165 on Build GRCh38
rs901069084 Chr.2: 200731244-200731244 on Build GRCh38
rs1025529982 Chr.2: 200731308-200731308 on Build GRCh38
rs970384664 Chr.2: 200731186-200731186 on Build GRCh38
rs781491695 Chr.2: 200731110-200731110 on Build GRCh38
rs986487849 Chr.2: 200731342-200731342 on Build GRCh38
rs940150337 Chr.2: 200731049-200731049 on Build GRCh38
rs746941856 Chr.2: 200730954-200730954 on Build GRCh38
rs1031505014 Chr.2: 200731231-200731231 on Build GRCh38
rs551886613 Chr.2: 200731216-200731216 on Build GRCh38
rs1013649189 Chr.2: 200731171-200731171 on Build GRCh38
rs534354853 Chr.2: 200731366-200731366 on Build GRCh38
rs747869513 Chr.2: 200731093-200731093 on Build GRCh38
rs957272154 Chr.2: 200731278-200731278 on Build GRCh38
rs565308806 Chr.2: 200731411-200731411 on Build GRCh38
rs185231618 Chr.2: 200731154-200731154 on Build GRCh38
rs918155090 Chr.2: 200731065-200731065 on Build GRCh38
rs548081507 Chr.2: 200731148-200731148 on Build GRCh38
rs889553415 Chr.2: 200731168-200731168 on Build GRCh38
rs953860864 Chr.2: 200731319-200731319 on Build GRCh38
rs188497315 Chr.2: 200731261-200731261 on Build GRCh38
rs1042056245 Chr.2: 200731104-200731104 on Build GRCh38
rs996166920 Chr.2: 200731133-200731133 on Build GRCh38
rs1026298276 Chr.2: 200730956-200730956 on Build GRCh38
rs116483221 Chr.2: 200731367-200731367 on Build GRCh38
rs989829225 Chr.2: 200730990-200730990 on Build GRCh38
rs768582146 Chr.2: 200731040-200731040 on Build GRCh38
rs918113623 Chr.2: 200731426-200731426 on Build GRCh38
rs950790188 Chr.2: 200730994-200730994 on Build GRCh38
rs10179459 Chr.2: 200731053-200731053 on Build GRCh38
rs10203498 Chr.2: 200731179-200731179 on Build GRCh38
rs774621572 Chr.2: 200731227-200731227 on Build GRCh38
rs549758096 Chr.2: 200731341-200731341 on Build GRCh38

View Full Product Details