Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200762001-200762546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOX3P-AOX2P
Entrez Gene ID
107161151
Gene Name
AOX3P-AOX2P readthrough
Gene Aliases
AOX3/AOX3L1
UniGene
Hs.148269
Gene Chromosome Location
Chr.2: 200695723-200794915 on Build GRCh38
Transcripts
NR_135011.1 NR_135012.1

SNP Detail

SNP ID Chromosome Location
rs890624268 Chr.2: 200762286-200762286 on Build GRCh38
rs893966779 Chr.2: 200762051-200762051 on Build GRCh38
rs568623355 Chr.2: 200762064-200762064 on Build GRCh38
rs1050602335 Chr.2: 200762083-200762083 on Build GRCh38
rs976843913 Chr.2: 200762063-200762063 on Build GRCh38
rs911250254 Chr.2: 200762029-200762029 on Build GRCh38
rs748067970 Chr.2: 200762195-200762195 on Build GRCh38
rs185009529 Chr.2: 200762154-200762154 on Build GRCh38
rs760680438 Chr.2: 200762452-200762452 on Build GRCh38
rs541515748 Chr.2: 200762269-200762269 on Build GRCh38
rs755892562 Chr.2: 200762205-200762205 on Build GRCh38
rs61746360 Chr.2: 200762206-200762206 on Build GRCh38
rs148958689 Chr.2: 200762285-200762285 on Build GRCh38
rs576485309 Chr.2: 200762150-200762150 on Build GRCh38
rs184906234 Chr.2: 200762040-200762040 on Build GRCh38
rs563982833 Chr.2: 200762303-200762303 on Build GRCh38
rs916427937 Chr.2: 200762224-200762224 on Build GRCh38
rs13427812 Chr.2: 200762271-200762271 on Build GRCh38
rs189679419 Chr.2: 200762387-200762387 on Build GRCh38
rs539929346 Chr.2: 200762130-200762130 on Build GRCh38
rs562336102 Chr.2: 200762121-200762121 on Build GRCh38
rs1053516143 Chr.2: 200762261-200762261 on Build GRCh38
rs386654184 Chr.2: 200762129-200762130 on Build GRCh38
rs538480211 Chr.2: 200762347-200762347 on Build GRCh38
rs971005666 Chr.2: 200762399-200762399 on Build GRCh38
rs754415091 Chr.2: 200762147-200762147 on Build GRCh38
rs923627857 Chr.2: 200762250-200762250 on Build GRCh38
rs890684749 Chr.2: 200762090-200762090 on Build GRCh38
rs922765414 Chr.2: 200762103-200762104 on Build GRCh38
rs536167343 Chr.2: 200762089-200762089 on Build GRCh38
rs894533007 Chr.2: 200762371-200762371 on Build GRCh38
rs867535237 Chr.2: 200762116-200762116 on Build GRCh38
rs751134117 Chr.2: 200762054-200762054 on Build GRCh38
rs762283357 Chr.2: 200762260-200762260 on Build GRCh38
rs554186918 Chr.2: 200762101-200762101 on Build GRCh38
rs990587813 Chr.2: 200762219-200762219 on Build GRCh38
rs994819861 Chr.2: 200762334-200762334 on Build GRCh38
rs982438934 Chr.2: 200762239-200762239 on Build GRCh38
rs187661960 Chr.2: 200762120-200762120 on Build GRCh38
rs763187640 Chr.2: 200762398-200762398 on Build GRCh38
rs541753020 Chr.2: 200762230-200762230 on Build GRCh38
rs1027722923 Chr.2: 200762183-200762183 on Build GRCh38
rs6725741 Chr.2: 200762178-200762178 on Build GRCh38
rs914970984 Chr.2: 200762266-200762266 on Build GRCh38
rs375169407 Chr.2: 200762231-200762231 on Build GRCh38
rs1031361237 Chr.2: 200762514-200762514 on Build GRCh38
rs982042527 Chr.2: 200762406-200762406 on Build GRCh38
rs1053572877 Chr.2: 200762049-200762049 on Build GRCh38
rs192102008 Chr.2: 200762143-200762143 on Build GRCh38
rs531449091 Chr.2: 200762309-200762309 on Build GRCh38
rs956628420 Chr.2: 200762518-200762518 on Build GRCh38
rs529313903 Chr.2: 200762061-200762061 on Build GRCh38
rs747100610 Chr.2: 200762492-200762492 on Build GRCh38
rs61744314 Chr.2: 200762287-200762287 on Build GRCh38
rs919108243 Chr.2: 200762467-200762467 on Build GRCh38
rs930098820 Chr.2: 200762120-200762121 on Build GRCh38
rs941592520 Chr.2: 200762446-200762446 on Build GRCh38
rs1037056936 Chr.2: 200762290-200762290 on Build GRCh38
rs73991105 Chr.2: 200762359-200762359 on Build GRCh38

View Full Product Details