Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 16675330-16675884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD62P1-PARP4P3
Entrez Gene ID
23783
Gene Name
ANKRD62P1-PARP4P3 readthrough, transcribed pseudogene
Gene Aliases
VWFP1-ANKRD62P1-PARP4P3
UniGene
Hs.627452
Gene Chromosome Location
Chr.22: 16653708-16675540 on Build GRCh38
Transcripts
NR_040115.1

SNP Detail

SNP ID Chromosome Location
rs921125732 Chr.22: 16675528-16675528 on Build GRCh38
rs1004254553 Chr.22: 16675859-16675859 on Build GRCh38
rs777261520 Chr.22: 16675364-16675364 on Build GRCh38
rs962501756 Chr.22: 16675523-16675523 on Build GRCh38
rs143927867 Chr.22: 16675671-16675671 on Build GRCh38
rs530664504 Chr.22: 16675424-16675425 on Build GRCh38
rs1041727764 Chr.22: 16675715-16675715 on Build GRCh38
rs375633895 Chr.22: 16675604-16675604 on Build GRCh38
rs932558578 Chr.22: 16675356-16675356 on Build GRCh38
rs1042191172 Chr.22: 16675389-16675389 on Build GRCh38
rs936116693 Chr.22: 16675718-16675718 on Build GRCh38
rs982453360 Chr.22: 16675500-16675500 on Build GRCh38
rs535751598 Chr.22: 16675462-16675462 on Build GRCh38
rs564191948 Chr.22: 16675678-16675678 on Build GRCh38
rs572294394 Chr.22: 16675638-16675638 on Build GRCh38
rs564467946 Chr.22: 16675839-16675839 on Build GRCh38
rs1050993427 Chr.22: 16675611-16675611 on Build GRCh38
rs6518594 Chr.22: 16675390-16675390 on Build GRCh38
rs971021489 Chr.22: 16675496-16675496 on Build GRCh38
rs181871949 Chr.22: 16675831-16675831 on Build GRCh38
rs908284192 Chr.22: 16675510-16675510 on Build GRCh38
rs868674141 Chr.22: 16675524-16675524 on Build GRCh38
rs903226759 Chr.22: 16675716-16675716 on Build GRCh38
rs185101216 Chr.22: 16675407-16675407 on Build GRCh38
rs866963653 Chr.22: 16675431-16675431 on Build GRCh38
rs1024539429 Chr.22: 16675494-16675498 on Build GRCh38
rs1033183895 Chr.22: 16675423-16675423 on Build GRCh38
rs1008931861 Chr.22: 16675371-16675372 on Build GRCh38
rs894632797 Chr.22: 16675811-16675811 on Build GRCh38
rs200630794 Chr.22: 16675633-16675633 on Build GRCh38
rs1055001451 Chr.22: 16675746-16675746 on Build GRCh38
rs189520269 Chr.22: 16675493-16675493 on Build GRCh38
rs199872086 Chr.22: 16675420-16675420 on Build GRCh38
rs912522273 Chr.22: 16675667-16675667 on Build GRCh38
rs576135008 Chr.22: 16675704-16675704 on Build GRCh38
rs547886276 Chr.22: 16675365-16675365 on Build GRCh38
rs1013132325 Chr.22: 16675458-16675458 on Build GRCh38
rs1050394931 Chr.22: 16675363-16675363 on Build GRCh38
rs907500659 Chr.22: 16675851-16675851 on Build GRCh38
rs201716493 Chr.22: 16675651-16675651 on Build GRCh38

View Full Product Details