Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51819082-51819631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCF4
Entrez Gene ID
100506840
Gene Name
IQ motif containing F5 pseudogene
Gene Aliases
-
UniGene
Hs.254808
Gene Chromosome Location
Chr.3: 51817603-51819635 on Build GRCh38
Transcripts
NR_038213.1 NR_038214.1

SNP Detail

SNP ID Chromosome Location
rs943673390 Chr.3: 51819410-51819410 on Build GRCh38
rs930019071 Chr.3: 51819138-51819138 on Build GRCh38
rs928861569 Chr.3: 51819584-51819584 on Build GRCh38
rs554261279 Chr.3: 51819444-51819444 on Build GRCh38
rs534897016 Chr.3: 51819372-51819372 on Build GRCh38
rs918812576 Chr.3: 51819109-51819109 on Build GRCh38
rs778344344 Chr.3: 51819337-51819337 on Build GRCh38
rs573030456 Chr.3: 51819452-51819452 on Build GRCh38
rs982453196 Chr.3: 51819378-51819378 on Build GRCh38
rs1009599250 Chr.3: 51819224-51819224 on Build GRCh38
rs1034236329 Chr.3: 51819261-51819261 on Build GRCh38
rs912851126 Chr.3: 51819127-51819127 on Build GRCh38
rs568443070 Chr.3: 51819411-51819411 on Build GRCh38
rs540048292 Chr.3: 51819478-51819478 on Build GRCh38
rs754342310 Chr.3: 51819249-51819249 on Build GRCh38
rs143090258 Chr.3: 51819448-51819448 on Build GRCh38
rs997437034 Chr.3: 51819458-51819458 on Build GRCh38
rs557537348 Chr.3: 51819451-51819451 on Build GRCh38
rs928640280 Chr.3: 51819425-51819425 on Build GRCh38
rs757833738 Chr.3: 51819432-51819432 on Build GRCh38
rs764530193 Chr.3: 51819195-51819195 on Build GRCh38
rs570985308 Chr.3: 51819353-51819353 on Build GRCh38
rs951128005 Chr.3: 51819371-51819371 on Build GRCh38
rs117077796 Chr.3: 51819500-51819500 on Build GRCh38
rs1030221407 Chr.3: 51819468-51819468 on Build GRCh38
rs535880120 Chr.3: 51819419-51819419 on Build GRCh38
rs865948903 Chr.3: 51819277-51819277 on Build GRCh38
rs965034475 Chr.3: 51819599-51819599 on Build GRCh38
rs555414612 Chr.3: 51819472-51819472 on Build GRCh38
rs963447396 Chr.3: 51819495-51819495 on Build GRCh38
rs1016042221 Chr.3: 51819493-51819493 on Build GRCh38
rs140866737 Chr.3: 51819308-51819308 on Build GRCh38
rs952986659 Chr.3: 51819311-51819311 on Build GRCh38
rs757775847 Chr.3: 51819288-51819288 on Build GRCh38
rs937359868 Chr.3: 51819585-51819585 on Build GRCh38
rs908927483 Chr.3: 51819348-51819348 on Build GRCh38
rs575186842 Chr.3: 51819440-51819440 on Build GRCh38
rs79492384 Chr.3: 51819156-51819156 on Build GRCh38
rs892535289 Chr.3: 51819223-51819223 on Build GRCh38
rs981665613 Chr.3: 51819524-51819524 on Build GRCh38
rs544240712 Chr.3: 51819510-51819510 on Build GRCh38
rs951179817 Chr.3: 51819518-51819518 on Build GRCh38
rs1024970564 Chr.3: 51819245-51819245 on Build GRCh38
rs772051194 Chr.3: 51819497-51819497 on Build GRCh38
rs556904897 Chr.3: 51819424-51819424 on Build GRCh38
rs531241424 Chr.3: 51819594-51819594 on Build GRCh38
rs561848847 Chr.3: 51819480-51819480 on Build GRCh38
rs774120661 Chr.3: 51819396-51819396 on Build GRCh38
rs553478520 Chr.3: 51819407-51819407 on Build GRCh38
rs1054300870 Chr.3: 51819218-51819218 on Build GRCh38
rs182733680 Chr.3: 51819200-51819200 on Build GRCh38
rs561616413 Chr.3: 51819208-51819208 on Build GRCh38
rs563909748 Chr.3: 51819152-51819152 on Build GRCh38
rs1007211102 Chr.3: 51819479-51819479 on Build GRCh38
rs937307310 Chr.3: 51819426-51819426 on Build GRCh38
rs2271068 Chr.3: 51819256-51819256 on Build GRCh38
rs550572468 Chr.3: 51819274-51819274 on Build GRCh38
rs562422826 Chr.3: 51819523-51819523 on Build GRCh38
rs552809257 Chr.3: 51819328-51819328 on Build GRCh38
rs973909488 Chr.3: 51819275-51819275 on Build GRCh38
rs570396587 Chr.3: 51819303-51819303 on Build GRCh38
rs1027172655 Chr.3: 51819347-51819347 on Build GRCh38
rs974631128 Chr.3: 51819393-51819393 on Build GRCh38
rs1011795356 Chr.3: 51819250-51819250 on Build GRCh38

View Full Product Details