Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26428545-26429092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN2A3P
Entrez Gene ID
54718
Gene Name
butyrophilin subfamily 2 member A3, pseudogene
Gene Aliases
BTN2.3, BTN2A3
UniGene
Hs.729655
Gene Chromosome Location
Chr.6: 26421391-26430588 on Build GRCh38
Transcripts
NR_027795.1 XM_011514268.2

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
NR_027795.1 XM_011514268.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3948891 c.847A>G p.N283D Chr.6: 26428737-26428737 on Build GRCh38
1621298 c.838A>G p.M280V Chr.6: 26428728-26428728 on Build GRCh38
3623515 c.834T>A p.H278Q Chr.6: 26428724-26428724 on Build GRCh38
6471006 c.846C>T p.F282F Chr.6: 26428736-26428736 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767691802 Chr.6: 26428691-26428691 on Build GRCh38
rs754218877 Chr.6: 26429026-26429026 on Build GRCh38
rs764815831 Chr.6: 26428744-26428744 on Build GRCh38
rs929695805 Chr.6: 26428584-26428584 on Build GRCh38
rs190608739 Chr.6: 26428827-26428827 on Build GRCh38
rs538820552 Chr.6: 26429047-26429047 on Build GRCh38
rs1036447260 Chr.6: 26428594-26428594 on Build GRCh38
rs1022216962 Chr.6: 26428995-26428995 on Build GRCh38
rs773530496 Chr.6: 26428736-26428736 on Build GRCh38
rs780053695 Chr.6: 26428739-26428740 on Build GRCh38
rs183639019 Chr.6: 26428857-26428857 on Build GRCh38
rs369075865 Chr.6: 26428685-26428685 on Build GRCh38
rs571534420 Chr.6: 26428739-26428739 on Build GRCh38
rs555461539 Chr.6: 26428847-26428847 on Build GRCh38
rs1002565216 Chr.6: 26428891-26428891 on Build GRCh38
rs749331499 Chr.6: 26428681-26428681 on Build GRCh38
rs879024342 Chr.6: 26428822-26428822 on Build GRCh38
rs191311184 Chr.6: 26428868-26428868 on Build GRCh38
rs762103969 Chr.6: 26428727-26428727 on Build GRCh38
rs1034451885 Chr.6: 26428673-26428673 on Build GRCh38
rs546249133 Chr.6: 26428981-26428981 on Build GRCh38
rs1036005849 Chr.6: 26428869-26428869 on Build GRCh38
rs921747430 Chr.6: 26428795-26428795 on Build GRCh38
rs757113449 Chr.6: 26428768-26428768 on Build GRCh38
rs887439679 Chr.6: 26428591-26428591 on Build GRCh38
rs965396669 Chr.6: 26429054-26429054 on Build GRCh38
rs766012028 Chr.6: 26428885-26428885 on Build GRCh38
rs186722245 Chr.6: 26429015-26429015 on Build GRCh38
rs753620472 Chr.6: 26428802-26428802 on Build GRCh38
rs182661741 Chr.6: 26428945-26428945 on Build GRCh38
rs754768689 Chr.6: 26429067-26429067 on Build GRCh38
rs62619242 Chr.6: 26428786-26428786 on Build GRCh38
rs911473864 Chr.6: 26428933-26428933 on Build GRCh38
rs186433588 Chr.6: 26428860-26428860 on Build GRCh38
rs1741740 Chr.6: 26428614-26428614 on Build GRCh38
rs752212493 Chr.6: 26428751-26428751 on Build GRCh38
rs768730362 Chr.6: 26428826-26428826 on Build GRCh38
rs756398140 Chr.6: 26428848-26428848 on Build GRCh38
rs1002596490 Chr.6: 26428660-26428660 on Build GRCh38
rs759248513 Chr.6: 26428740-26428740 on Build GRCh38
rs941567572 Chr.6: 26428934-26428934 on Build GRCh38
rs1045023294 Chr.6: 26429051-26429051 on Build GRCh38
rs763964323 Chr.6: 26428764-26428764 on Build GRCh38
rs748725769 Chr.6: 26428828-26428828 on Build GRCh38
rs767916028 Chr.6: 26428728-26428728 on Build GRCh38
rs1012179369 Chr.6: 26428642-26428643 on Build GRCh38
rs181936484 Chr.6: 26428592-26428592 on Build GRCh38
rs528247068 Chr.6: 26428696-26428696 on Build GRCh38
rs201780313 Chr.6: 26428738-26428738 on Build GRCh38
rs755769752 Chr.6: 26428736-26428738 on Build GRCh38
rs1011733918 Chr.6: 26428991-26428991 on Build GRCh38
rs967774195 Chr.6: 26428750-26428750 on Build GRCh38
rs757888177 Chr.6: 26428753-26428753 on Build GRCh38
rs745644071 Chr.6: 26428788-26428788 on Build GRCh38
rs372000188 Chr.6: 26428790-26428790 on Build GRCh38
rs544576912 Chr.6: 26428569-26428569 on Build GRCh38
rs376817277 Chr.6: 26428737-26428737 on Build GRCh38
rs542495004 Chr.6: 26428666-26428666 on Build GRCh38

View Full Product Details