Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240682533-240682875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP12B
Entrez Gene ID
653437
Gene Name
aquaporin 12B
Gene Aliases
INSSA3
UniGene
Hs.437167
Gene Chromosome Location
Chr.2: 240676418-240685743 on Build GRCh38
Transcripts
NM_001102467.1 XM_011511671.2 XM_011511672.2 XM_011511673.2 XM_011511675.2 XM_011511676.2 XM_011511677.2 XM_011511678.2 XM_017004736.1 XM_017004737.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4662571 c.213G>A p.L71L Chr.2: 240682625-240682625 on Build GRCh38
318740 c.112G>T p.A38S Chr.2: 240682726-240682726 on Build GRCh38
4662573 c.116G>A p.R39Q Chr.2: 240682722-240682722 on Build GRCh38
4662575 c.15C>T p.N5N Chr.2: 240682823-240682823 on Build GRCh38
3579934 c.118G>A p.E40K Chr.2: 240682720-240682720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761181821 Chr.2: 240682566-240682566 on Build GRCh38
rs1047496389 Chr.2: 240682772-240682772 on Build GRCh38
rs769955023 Chr.2: 240682847-240682847 on Build GRCh38
rs745862809 Chr.2: 240682555-240682555 on Build GRCh38
rs767393771 Chr.2: 240682847-240682847 on Build GRCh38
rs368082796 Chr.2: 240682823-240682823 on Build GRCh38
rs757561287 Chr.2: 240682605-240682605 on Build GRCh38
rs201534317 Chr.2: 240682791-240682791 on Build GRCh38
rs745524394 Chr.2: 240682769-240682769 on Build GRCh38
rs778550757 Chr.2: 240682685-240682685 on Build GRCh38
rs547736661 Chr.2: 240682755-240682755 on Build GRCh38
rs763351629 Chr.2: 240682703-240682703 on Build GRCh38
rs773680074 Chr.2: 240682782-240682782 on Build GRCh38
rs770357699 Chr.2: 240682696-240682696 on Build GRCh38
rs772755374 Chr.2: 240682779-240682779 on Build GRCh38
rs368514296 Chr.2: 240682851-240682851 on Build GRCh38
rs780541240 Chr.2: 240682723-240682723 on Build GRCh38
rs543814740 Chr.2: 240682706-240682706 on Build GRCh38
rs763858804 Chr.2: 240682758-240682758 on Build GRCh38
rs747275019 Chr.2: 240682580-240682580 on Build GRCh38
rs531938199 Chr.2: 240682711-240682711 on Build GRCh38
rs752560158 Chr.2: 240682756-240682756 on Build GRCh38
rs566733461 Chr.2: 240682621-240682621 on Build GRCh38
rs369678598 Chr.2: 240682727-240682727 on Build GRCh38
rs750234915 Chr.2: 240682855-240682855 on Build GRCh38
rs776749172 Chr.2: 240682744-240682744 on Build GRCh38
rs759364765 Chr.2: 240682846-240682847 on Build GRCh38
rs775434578 Chr.2: 240682561-240682561 on Build GRCh38
rs73117922 Chr.2: 240682707-240682707 on Build GRCh38
rs373980987 Chr.2: 240682589-240682589 on Build GRCh38
rs759610365 Chr.2: 240682753-240682753 on Build GRCh38
rs376925022 Chr.2: 240682594-240682594 on Build GRCh38
rs4081909 Chr.2: 240682770-240682770 on Build GRCh38
rs372427935 Chr.2: 240682565-240682565 on Build GRCh38
rs544610485 Chr.2: 240682672-240682674 on Build GRCh38
rs750575054 Chr.2: 240682612-240682612 on Build GRCh38
rs747482212 Chr.2: 240682836-240682836 on Build GRCh38
rs764528780 Chr.2: 240682601-240682601 on Build GRCh38
rs772562887 Chr.2: 240682740-240682740 on Build GRCh38
rs781474336 Chr.2: 240682841-240682841 on Build GRCh38
rs769641721 Chr.2: 240682558-240682558 on Build GRCh38
rs763516852 Chr.2: 240682658-240682658 on Build GRCh38
rs375801155 Chr.2: 240682797-240682797 on Build GRCh38
rs774356588 Chr.2: 240682704-240682705 on Build GRCh38
rs560641543 Chr.2: 240682704-240682704 on Build GRCh38
rs374081283 Chr.2: 240682822-240682822 on Build GRCh38
rs959286311 Chr.2: 240682716-240682716 on Build GRCh38
rs192451353 Chr.2: 240682709-240682709 on Build GRCh38
rs768270919 Chr.2: 240682642-240682642 on Build GRCh38
rs747743140 Chr.2: 240682645-240682645 on Build GRCh38
rs199595129 Chr.2: 240682725-240682725 on Build GRCh38
rs374703341 Chr.2: 240682593-240682593 on Build GRCh38
rs369078305 Chr.2: 240682574-240682574 on Build GRCh38
rs184458523 Chr.2: 240682778-240682778 on Build GRCh38
rs187845451 Chr.2: 240682591-240682591 on Build GRCh38
rs370144346 Chr.2: 240682694-240682694 on Build GRCh38
rs189242283 Chr.2: 240682663-240682663 on Build GRCh38
rs373139392 Chr.2: 240682581-240682581 on Build GRCh38
rs201173715 Chr.2: 240682670-240682670 on Build GRCh38
rs778375961 Chr.2: 240682833-240682833 on Build GRCh38
rs777816086 Chr.2: 240682576-240682576 on Build GRCh38
rs369219301 Chr.2: 240682588-240682588 on Build GRCh38
rs372551464 Chr.2: 240682838-240682838 on Build GRCh38
rs201422028 Chr.2: 240682722-240682722 on Build GRCh38
rs896122282 Chr.2: 240682554-240682554 on Build GRCh38
rs376529999 Chr.2: 240682852-240682852 on Build GRCh38
rs773025520 Chr.2: 240682583-240682584 on Build GRCh38
rs767332173 Chr.2: 240682853-240682853 on Build GRCh38
rs771884214 Chr.2: 240682693-240682693 on Build GRCh38
rs192736899 Chr.2: 240682592-240682592 on Build GRCh38
rs4081908 Chr.2: 240682792-240682792 on Build GRCh38
rs753998513 Chr.2: 240682710-240682710 on Build GRCh38
rs780852278 Chr.2: 240682767-240682767 on Build GRCh38
rs756207344 Chr.2: 240682671-240682671 on Build GRCh38
rs184125501 Chr.2: 240682617-240682617 on Build GRCh38
rs754565566 Chr.2: 240682830-240682830 on Build GRCh38
rs755546146 Chr.2: 240682572-240682572 on Build GRCh38
rs767068318 Chr.2: 240682567-240682567 on Build GRCh38
rs182755222 Chr.2: 240682585-240682585 on Build GRCh38
rs758393875 Chr.2: 240682757-240682757 on Build GRCh38
rs765179725 Chr.2: 240682712-240682712 on Build GRCh38
rs757037681 Chr.2: 240682760-240682760 on Build GRCh38
rs373770837 Chr.2: 240682610-240682610 on Build GRCh38
rs550046466 Chr.2: 240682714-240682714 on Build GRCh38
rs369177771 Chr.2: 240682715-240682715 on Build GRCh38
rs776853080 Chr.2: 240682789-240682789 on Build GRCh38
rs754277716 Chr.2: 240682570-240682570 on Build GRCh38
rs755782261 Chr.2: 240682816-240682816 on Build GRCh38
rs766663764 Chr.2: 240682742-240682742 on Build GRCh38
rs762637257 Chr.2: 240682595-240682603 on Build GRCh38
rs746656089 Chr.2: 240682718-240682718 on Build GRCh38
rs759678385 Chr.2: 240682790-240682790 on Build GRCh38
rs928940963 Chr.2: 240682765-240682765 on Build GRCh38
rs577637446 Chr.2: 240682805-240682805 on Build GRCh38
rs4676387 Chr.2: 240682652-240682652 on Build GRCh38
rs749263916 Chr.2: 240682628-240682628 on Build GRCh38
rs373693752 Chr.2: 240682759-240682759 on Build GRCh38
rs370405826 Chr.2: 240682803-240682803 on Build GRCh38
rs573480304 Chr.2: 240682657-240682657 on Build GRCh38
rs753715441 Chr.2: 240682681-240682681 on Build GRCh38
rs755000716 Chr.2: 240682684-240682684 on Build GRCh38
rs533863219 Chr.2: 240682771-240682771 on Build GRCh38
rs375213961 Chr.2: 240682846-240682846 on Build GRCh38
rs370087461 Chr.2: 240682553-240682553 on Build GRCh38
rs531874196 Chr.2: 240682586-240682586 on Build GRCh38
rs769213334 Chr.2: 240682726-240682726 on Build GRCh38
rs202020511 Chr.2: 240682573-240682573 on Build GRCh38
rs555177924 Chr.2: 240682656-240682656 on Build GRCh38
rs780287254 Chr.2: 240682672-240682672 on Build GRCh38
rs555243128 Chr.2: 240682788-240682788 on Build GRCh38
rs543324817 Chr.2: 240682708-240682708 on Build GRCh38
rs200240739 Chr.2: 240682644-240682644 on Build GRCh38
rs577576670 Chr.2: 240682676-240682676 on Build GRCh38
rs773771969 Chr.2: 240682741-240682741 on Build GRCh38
rs747337536 Chr.2: 240682784-240682784 on Build GRCh38
rs200034703 Chr.2: 240682664-240682664 on Build GRCh38
rs751661293 Chr.2: 240682604-240682604 on Build GRCh38
rs370116541 Chr.2: 240682596-240682596 on Build GRCh38
rs746086433 Chr.2: 240682844-240682844 on Build GRCh38
rs555990436 Chr.2: 240682793-240682793 on Build GRCh38
rs116505219 Chr.2: 240682686-240682686 on Build GRCh38

View Full Product Details