Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 111783793-111784090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC1
Entrez Gene ID
64682
Gene Name
anaphase promoting complex subunit 1
Gene Aliases
APC1, MCPR, TSG24
UniGene
Hs.436527
Gene Chromosome Location
Chr.2: 111766147-111884196 on Build GRCh38
Transcripts
NM_022662.3 XM_011511633.2 XM_011511634.2 XM_017004710.1 XM_017004711.1 XM_017004712.1 XM_017004713.1 XM_017004714.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1005399 c.5034A>C p.I1678I Chr.2: 111783926-111783926 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777238148 Chr.2: 111783868-111783868 on Build GRCh38
rs747043648 Chr.2: 111783942-111783942 on Build GRCh38
rs577213586 Chr.2: 111784019-111784019 on Build GRCh38
rs745718417 Chr.2: 111783912-111783912 on Build GRCh38
rs756935487 Chr.2: 111783889-111783889 on Build GRCh38
rs755695831 Chr.2: 111783862-111783862 on Build GRCh38
rs753558032 Chr.2: 111783885-111783885 on Build GRCh38
rs574578224 Chr.2: 111783929-111783929 on Build GRCh38
rs2245224 Chr.2: 111784067-111784067 on Build GRCh38
rs776655137 Chr.2: 111783990-111783990 on Build GRCh38
rs545775241 Chr.2: 111784026-111784026 on Build GRCh38
rs762112964 Chr.2: 111784014-111784014 on Build GRCh38
rs1030227529 Chr.2: 111783853-111783853 on Build GRCh38
rs543691623 Chr.2: 111783932-111783932 on Build GRCh38
rs557338993 Chr.2: 111783978-111783978 on Build GRCh38

View Full Product Details