Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 102853691-102853820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIVM-ERCC5
Entrez Gene ID
100533467
Gene Name
BIVM-ERCC5 readthrough
Gene Aliases
ERCC5-202
UniGene
Hs.258429
Gene Chromosome Location
Chr.13: 102807146-102876001 on Build GRCh38
Transcripts
NM_001204425.1 NM_000123.3

Gene Symbol
ERCC5
Entrez Gene ID
2073
Gene Name
ERCC excision repair 5, endonuclease
Gene Aliases
COFS3, ERCC5-201, ERCM2, UVDR, XPG, XPGC
UniGene
Hs.258429
Gene Chromosome Location
Chr.13: 102845841-102876001 on Build GRCh38
Transcripts
NM_001204425.1 NM_000123.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6971195 c.293C>T p.A98V Chr.13: 102853785-102853785 on Build GRCh38
4605096 c.315G>A p.T105T Chr.13: 102853807-102853807 on Build GRCh38
6463055 c.317C>T p.T106I Chr.13: 102853809-102853809 on Build GRCh38
1628981 c.265-1G>T p.? Chr.13: 102853756-102853756 on Build GRCh38
6463054 c.317C>T p.T106I Chr.13: 102853809-102853809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757483997 Chr.13: 102853783-102853783 on Build GRCh38
rs375404851 Chr.13: 102853785-102853785 on Build GRCh38
rs150741329 Chr.13: 102853795-102853795 on Build GRCh38
rs764029659 Chr.13: 102853766-102853766 on Build GRCh38
rs372298533 Chr.13: 102853777-102853777 on Build GRCh38
rs199515913 Chr.13: 102853789-102853789 on Build GRCh38
rs759317041 Chr.13: 102853733-102853733 on Build GRCh38
rs767397228 Chr.13: 102853741-102853741 on Build GRCh38
rs759706438 Chr.13: 102853743-102853746 on Build GRCh38
rs777394192 Chr.13: 102853763-102853763 on Build GRCh38
rs752548779 Chr.13: 102853752-102853752 on Build GRCh38
rs969279305 Chr.13: 102853747-102853747 on Build GRCh38
rs780506840 Chr.13: 102853786-102853786 on Build GRCh38
rs1017806595 Chr.13: 102853734-102853734 on Build GRCh38
rs368957207 Chr.13: 102853722-102853722 on Build GRCh38
rs760695391 Chr.13: 102853762-102853762 on Build GRCh38

View Full Product Details