Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87046864-87047008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFF1
Entrez Gene ID
4299
Gene Name
AF4/FMR2 family member 1
Gene Aliases
AF4, MLLT2, PBM1
UniGene
Hs.480190
Gene Chromosome Location
Chr.4: 86935002-87141054 on Build GRCh38
Transcripts
NM_001166693.2 NM_001313959.1 NM_001313960.1 NM_005935.3 XM_005263007.3 XM_005263009.3 XM_005263011.3 XM_005263013.3 XM_011531973.2 XM_017008215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6595792 c.442G>A p.E148K Chr.4: 87046977-87046977 on Build GRCh38
4992194 c.365C>G p.S122C Chr.4: 87046900-87046900 on Build GRCh38
1058566 c.429G>A p.A143A Chr.4: 87046964-87046964 on Build GRCh38
3674238 c.433C>A p.P145T Chr.4: 87046968-87046968 on Build GRCh38
6476971 c.445C>T p.P149S Chr.4: 87046980-87046980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868847726 Chr.4: 87046967-87046967 on Build GRCh38
rs144461444 Chr.4: 87046964-87046964 on Build GRCh38
rs748778328 Chr.4: 87046916-87046916 on Build GRCh38
rs367720775 Chr.4: 87046959-87046959 on Build GRCh38
rs750394594 Chr.4: 87046911-87046911 on Build GRCh38
rs976133352 Chr.4: 87046937-87046937 on Build GRCh38
rs770309691 Chr.4: 87046980-87046980 on Build GRCh38
rs972805304 Chr.4: 87046984-87046984 on Build GRCh38
rs150065985 Chr.4: 87046915-87046915 on Build GRCh38
rs145392385 Chr.4: 87046962-87046962 on Build GRCh38
rs139425059 Chr.4: 87046889-87046889 on Build GRCh38
rs751739957 Chr.4: 87046891-87046891 on Build GRCh38
rs780981287 Chr.4: 87046901-87046901 on Build GRCh38
rs909775787 Chr.4: 87046973-87046973 on Build GRCh38
rs754500412 Chr.4: 87046951-87046951 on Build GRCh38
rs374201893 Chr.4: 87046892-87046892 on Build GRCh38
rs763051091 Chr.4: 87046985-87046985 on Build GRCh38
rs564970030 Chr.4: 87046884-87046884 on Build GRCh38
rs909804909 Chr.4: 87046886-87046886 on Build GRCh38
rs372635952 Chr.4: 87046983-87046983 on Build GRCh38
rs771430275 Chr.4: 87046963-87046963 on Build GRCh38
rs746415345 Chr.4: 87046972-87046972 on Build GRCh38

View Full Product Details