Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 91992154-91992308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP9
Entrez Gene ID
10142
Gene Name
A-kinase anchoring protein 9
Gene Aliases
AKAP-9, AKAP350, AKAP450, CG-NAP, HYPERION, LQT11, MU-RMS-40.16A, PPP1R45, PRKA9, YOTIAO
UniGene
Hs.651221
Gene Chromosome Location
Chr.7: 91940867-92110673 on Build GRCh38
Transcripts
NM_005751.4 NM_147185.2 XM_017011642.1 XM_017011643.1 XM_017011644.1 XM_017011645.1 XM_017011646.1 XM_017011647.1 XM_017011648.1 XM_017011649.1 XM_017011650.1 XM_017011651.1 XM_017011652.1 XM_017011653.1 XM_017011654.1 XM_017011655.1 XM_017011656.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2863564 c.397T>G p.L133V Chr.7: 91992203-91992203 on Build GRCh38
1550358 c.405G>T p.R135S Chr.7: 91992211-91992211 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs188318223 Chr.7: 91992179-91992179 on Build GRCh38
rs376736925 Chr.7: 91992256-91992256 on Build GRCh38
rs775043801 Chr.7: 91992244-91992244 on Build GRCh38
rs778747883 Chr.7: 91992216-91992216 on Build GRCh38
rs754335465 Chr.7: 91992178-91992178 on Build GRCh38
rs746539511 Chr.7: 91992249-91992249 on Build GRCh38
rs757027446 Chr.7: 91992212-91992212 on Build GRCh38
rs886471976 Chr.7: 91992229-91992229 on Build GRCh38
rs1008881176 Chr.7: 91992275-91992275 on Build GRCh38
rs755049262 Chr.7: 91992200-91992200 on Build GRCh38
rs200518712 Chr.7: 91992195-91992195 on Build GRCh38
rs771710726 Chr.7: 91992243-91992243 on Build GRCh38
rs749209057 Chr.7: 91992211-91992211 on Build GRCh38
rs765586191 Chr.7: 91992184-91992184 on Build GRCh38
rs750656682 Chr.7: 91992186-91992186 on Build GRCh38
rs758661269 Chr.7: 91992194-91992194 on Build GRCh38
rs751675949 Chr.7: 91992198-91992198 on Build GRCh38

View Full Product Details