Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47216330-47216504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDB2
Entrez Gene ID
1643
Gene Name
damage specific DNA binding protein 2
Gene Aliases
DDBB, UV-DDB2, XPE
UniGene
Hs.700338
Gene Chromosome Location
Chr.11: 47214942-47239218 on Build GRCh38
Transcripts
NM_000107.2 NM_001300734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1249843 c.210C>T p.V70V Chr.11: 47216418-47216418 on Build GRCh38
1298086 c.158T>C p.L53P Chr.11: 47216366-47216366 on Build GRCh38
6385218 c.172G>A p.A58T Chr.11: 47216380-47216380 on Build GRCh38
3809303 c.189G>A p.L63L Chr.11: 47216397-47216397 on Build GRCh38
5903778 c.227A>C p.H76P Chr.11: 47216435-47216435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758168272 Chr.11: 47216415-47216415 on Build GRCh38
rs753467757 Chr.11: 47216400-47216400 on Build GRCh38
rs1043766285 Chr.11: 47216457-47216457 on Build GRCh38
rs749426163 Chr.11: 47216445-47216445 on Build GRCh38
rs760456614 Chr.11: 47216391-47216391 on Build GRCh38
rs756368265 Chr.11: 47216431-47216431 on Build GRCh38
rs535591210 Chr.11: 47216350-47216350 on Build GRCh38
rs779731799 Chr.11: 47216416-47216416 on Build GRCh38
rs374094218 Chr.11: 47216480-47216480 on Build GRCh38
rs199630230 Chr.11: 47216426-47216426 on Build GRCh38
rs904410426 Chr.11: 47216466-47216466 on Build GRCh38
rs770922074 Chr.11: 47216462-47216462 on Build GRCh38
rs139674102 Chr.11: 47216408-47216408 on Build GRCh38
rs772355051 Chr.11: 47216366-47216366 on Build GRCh38
rs777824005 Chr.11: 47216432-47216432 on Build GRCh38
rs759879517 Chr.11: 47216355-47216355 on Build GRCh38
rs191773047 Chr.11: 47216397-47216397 on Build GRCh38
rs1015435321 Chr.11: 47216413-47216413 on Build GRCh38
rs568187738 Chr.11: 47216405-47216405 on Build GRCh38
rs765161300 Chr.11: 47216407-47216407 on Build GRCh38
rs150620642 Chr.11: 47216374-47216374 on Build GRCh38

View Full Product Details