Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 51054919-51055044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAD4
Entrez Gene ID
4089
Gene Name
SMAD family member 4
Gene Aliases
DPC4, JIP, MADH4, MYHRS
UniGene
Hs.75862
Gene Chromosome Location
Chr.18: 51030213-51085042 on Build GRCh38
Transcripts
NM_005359.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
14104 c.1_*del p.0 Chr.18: 51047047-51078467 on Build GRCh38
5369896 c.664_665insC p.S223fs*12 Chr.18: 51054990-51054991 on Build GRCh38
6476005 c.661T>C p.S221P Chr.18: 51054987-51054987 on Build GRCh38
6951875 c.664A>T p.T222S Chr.18: 51054990-51054990 on Build GRCh38
14157 c.607_608delCC p.P203fs*2 Chr.18: 51054933-51054934 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776523203 Chr.18: 51055002-51055002 on Build GRCh38
rs760785405 Chr.18: 51055004-51055004 on Build GRCh38
rs748615724 Chr.18: 51054940-51054940 on Build GRCh38
rs757977781 Chr.18: 51054964-51054964 on Build GRCh38
rs760777245 Chr.18: 51054995-51054996 on Build GRCh38
rs764527544 Chr.18: 51054998-51054999 on Build GRCh38
rs1046411210 Chr.18: 51054948-51054948 on Build GRCh38
rs878854767 Chr.18: 51054965-51054965 on Build GRCh38
rs770461626 Chr.18: 51054990-51054990 on Build GRCh38
rs138386557 Chr.18: 51054973-51054973 on Build GRCh38
rs370726274 Chr.18: 51055010-51055010 on Build GRCh38
rs779564453 Chr.18: 51055000-51055001 on Build GRCh38
rs757971589 Chr.18: 51054996-51054996 on Build GRCh38
rs200331016 Chr.18: 51055003-51055003 on Build GRCh38
rs905151346 Chr.18: 51054953-51054953 on Build GRCh38
rs777495692 Chr.18: 51054970-51054970 on Build GRCh38
rs763433258 Chr.18: 51054997-51054998 on Build GRCh38
rs773336735 Chr.18: 51054994-51054995 on Build GRCh38
rs766541692 Chr.18: 51055008-51055008 on Build GRCh38
rs775133117 Chr.18: 51055000-51055000 on Build GRCh38
rs769835478 Chr.18: 51054998-51054998 on Build GRCh38
rs547532436 Chr.18: 51055013-51055013 on Build GRCh38
rs997151197 Chr.18: 51054977-51054977 on Build GRCh38
rs753440205 Chr.18: 51055013-51055015 on Build GRCh38
rs112891188 Chr.18: 51054986-51054986 on Build GRCh38
rs765834260 Chr.18: 51055012-51055012 on Build GRCh38
rs863224732 Chr.18: 51054960-51054960 on Build GRCh38
rs771895353 Chr.18: 51054992-51054993 on Build GRCh38

View Full Product Details