Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 66443283-66443440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP2K1
Entrez Gene ID
5604
Gene Name
mitogen-activated protein kinase kinase 1
Gene Aliases
CFC3, MAPKK1, MEK1, MKK1, PRKMK1
UniGene
Hs.145442
Gene Chromosome Location
Chr.15: 66386873-66491544 on Build GRCh38
Transcripts
NM_002755.3 XM_011521783.2 XM_017022411.1 XM_017022412.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3690651 c.449C>T p.S150F Chr.15: 66443290-66443290 on Build GRCh38
1724124 c.484C>T p.P162S Chr.15: 66443325-66443325 on Build GRCh38
6968948 c.473C>T p.A158V Chr.15: 66443314-66443314 on Build GRCh38
5849601 c.460G>A p.V154I Chr.15: 66443301-66443301 on Build GRCh38
6906560 c.479G>C p.R160T Chr.15: 66443320-66443320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033086799 Chr.15: 66443384-66443384 on Build GRCh38
rs1001637882 Chr.15: 66443373-66443373 on Build GRCh38
rs761845350 Chr.15: 66443369-66443369 on Build GRCh38
rs754563110 Chr.15: 66443404-66443404 on Build GRCh38
rs768080091 Chr.15: 66443367-66443367 on Build GRCh38
rs776863823 Chr.15: 66443366-66443366 on Build GRCh38
rs184208600 Chr.15: 66443385-66443385 on Build GRCh38
rs955368719 Chr.15: 66443398-66443398 on Build GRCh38
rs568143190 Chr.15: 66443413-66443413 on Build GRCh38
rs768918234 Chr.15: 66443357-66443357 on Build GRCh38
rs772312180 Chr.15: 66443318-66443318 on Build GRCh38
rs987807830 Chr.15: 66443412-66443412 on Build GRCh38
rs369343022 Chr.15: 66443402-66443402 on Build GRCh38
rs906058031 Chr.15: 66443370-66443370 on Build GRCh38
rs201208360 Chr.15: 66443310-66443310 on Build GRCh38
rs376919469 Chr.15: 66443395-66443395 on Build GRCh38
rs200487483 Chr.15: 66443312-66443312 on Build GRCh38
rs199504372 Chr.15: 66443307-66443307 on Build GRCh38
rs372680156 Chr.15: 66443387-66443387 on Build GRCh38
rs747171317 Chr.15: 66443336-66443336 on Build GRCh38
rs140822439 Chr.15: 66443333-66443333 on Build GRCh38
rs1057518013 Chr.15: 66443359-66443359 on Build GRCh38
rs765301197 Chr.15: 66443383-66443383 on Build GRCh38
rs1022480087 Chr.15: 66443354-66443354 on Build GRCh38
rs886051366 Chr.15: 66443344-66443344 on Build GRCh38

View Full Product Details