Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26731191-26731361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID1A
Entrez Gene ID
8289
Gene Name
AT-rich interaction domain 1A
Gene Aliases
B120, BAF250, BAF250a, BM029, C1orf4, CSS2, ELD, MRD14, OSA1, P270, SMARCF1, hELD, hOSA1
UniGene
Hs.468972
Gene Chromosome Location
Chr.1: 26696031-26782110 on Build GRCh38
Transcripts
NM_006015.4 NM_139135.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
318771 c.1476G>T p.Q492H Chr.1: 26731277-26731277 on Build GRCh38
4848070 c.1528C>T p.Q510* Chr.1: 26731329-26731329 on Build GRCh38
7356381 c.1487C>G p.A496G Chr.1: 26731288-26731288 on Build GRCh38
307377 c.1508_1509insAAGGCCCCAGCGGGTATGGTCAACAGGGC p.P504fs*125 Chr.1: 26731309-26731310 on Build GRCh38
907685 c.1449_1450insCC p.Y485fs*135 Chr.1: 26731250-26731251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772741483 Chr.1: 26731291-26731291 on Build GRCh38
rs770551517 Chr.1: 26731323-26731323 on Build GRCh38
rs771412871 Chr.1: 26731271-26731271 on Build GRCh38
rs756017930 Chr.1: 26731235-26731236 on Build GRCh38
rs757458581 Chr.1: 26731231-26731231 on Build GRCh38
rs754027692 Chr.1: 26731228-26731228 on Build GRCh38
rs746775479 Chr.1: 26731241-26731241 on Build GRCh38
rs901983604 Chr.1: 26731298-26731298 on Build GRCh38
rs61749357 Chr.1: 26731313-26731313 on Build GRCh38
rs748935625 Chr.1: 26731297-26731297 on Build GRCh38
rs780033196 Chr.1: 26731234-26731234 on Build GRCh38
rs748065625 Chr.1: 26731274-26731274 on Build GRCh38
rs781266953 Chr.1: 26731270-26731270 on Build GRCh38
rs1049214262 Chr.1: 26731253-26731253 on Build GRCh38
rs200920292 Chr.1: 26731330-26731330 on Build GRCh38
rs370698486 Chr.1: 26731322-26731323 on Build GRCh38
rs866935743 Chr.1: 26731264-26731264 on Build GRCh38
rs755020333 Chr.1: 26731250-26731250 on Build GRCh38
rs774757600 Chr.1: 26731324-26731324 on Build GRCh38
rs769213510 Chr.1: 26731287-26731287 on Build GRCh38
rs368949162 Chr.1: 26731312-26731312 on Build GRCh38

View Full Product Details