Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 48434901-48435075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP22
Entrez Gene ID
58504
Gene Name
Rho GTPase activating protein 22
Gene Aliases
RhoGAP2, RhoGap22
UniGene
Hs.655672
Gene Chromosome Location
Chr.10: 48429831-48656265 on Build GRCh38
Transcripts
XM_011540013.2 XM_017016473.1 XM_017016476.1 NM_001278547.1 NM_001278548.1 NM_001323302.1 NM_001323320.1 NM_001323321.1 NM_001323322.1 NM_001323323.1 NM_001323324.1 NM_001323325.1 NM_001323326.1 NM_001323327.1 NM_001323328.1 NM_001323329.1 NM_001323330.1 NM_001323331.1 NM_139046.3 NM_139049.3 NR_136583.1 NR_136584.1 NR_136585.1

Gene Symbol
MAPK8
Entrez Gene ID
5599
Gene Name
mitogen-activated protein kinase 8
Gene Aliases
JNK, JNK-46, JNK1, JNK1A2, JNK21B1/2, PRKM8, SAPK1, SAPK1c
UniGene
Hs.138211
Gene Chromosome Location
Chr.10: 48306639-48439360 on Build GRCh38
Transcripts
XM_011540013.2 XM_017016473.1 XM_017016476.1 NM_001278547.1 NM_001278548.1 NM_001323302.1 NM_001323320.1 NM_001323321.1 NM_001323322.1 NM_001323323.1 NM_001323324.1 NM_001323325.1 NM_001323326.1 NM_001323327.1 NM_001323328.1 NM_001323329.1 NM_001323330.1 NM_001323331.1 NM_139046.3 NM_139049.3 NR_136583.1 NR_136584.1 NR_136585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1745809 c.1162C>A p.Q388K Chr.10: 48434907-48434907 on Build GRCh38
7328193 c.1177T>C p.S393P Chr.10: 48434922-48434922 on Build GRCh38
6707878 c.1176A>G p.S392S Chr.10: 48434921-48434921 on Build GRCh38
918481 c.1179G>A p.S393S Chr.10: 48434924-48434924 on Build GRCh38
538903 c.1160C>G p.S387C Chr.10: 48434905-48434905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762432631 Chr.10: 48434934-48434936 on Build GRCh38
rs759451457 Chr.10: 48434934-48434934 on Build GRCh38
rs992324943 Chr.10: 48434940-48434940 on Build GRCh38
rs763758430 Chr.10: 48434950-48434950 on Build GRCh38
rs972332361 Chr.10: 48435000-48435000 on Build GRCh38
rs750925995 Chr.10: 48434963-48434963 on Build GRCh38
rs142606131 Chr.10: 48435004-48435004 on Build GRCh38
rs781538114 Chr.10: 48435016-48435016 on Build GRCh38
rs770839347 Chr.10: 48435036-48435036 on Build GRCh38
rs200811159 Chr.10: 48435043-48435043 on Build GRCh38
rs149971269 Chr.10: 48434927-48434927 on Build GRCh38
rs1035626313 Chr.10: 48435049-48435049 on Build GRCh38
rs377754613 Chr.10: 48435046-48435046 on Build GRCh38
rs147673874 Chr.10: 48434998-48434998 on Build GRCh38
rs765524640 Chr.10: 48435048-48435048 on Build GRCh38
rs367599489 Chr.10: 48434935-48434935 on Build GRCh38
rs778866733 Chr.10: 48435039-48435039 on Build GRCh38
rs959786981 Chr.10: 48434939-48434939 on Build GRCh38
rs765655352 Chr.10: 48434962-48434962 on Build GRCh38
rs774869055 Chr.10: 48434949-48434949 on Build GRCh38
rs748451475 Chr.10: 48435024-48435024 on Build GRCh38
rs755161674 Chr.10: 48435008-48435008 on Build GRCh38
rs1011736095 Chr.10: 48435003-48435003 on Build GRCh38
rs753537417 Chr.10: 48434957-48434957 on Build GRCh38
rs142330800 Chr.10: 48434924-48434924 on Build GRCh38
rs549658964 Chr.10: 48435045-48435045 on Build GRCh38
rs933003988 Chr.10: 48434929-48434929 on Build GRCh38
rs531537127 Chr.10: 48435044-48435044 on Build GRCh38

View Full Product Details