Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 225510-225655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDHA
Entrez Gene ID
6389
Gene Name
succinate dehydrogenase complex flavoprotein subunit A
Gene Aliases
CMD1GG, FP, PGL5, SDH1, SDH2, SDHF
UniGene
Hs.440475
Gene Chromosome Location
Chr.5: 218223-264816 on Build GRCh38
Transcripts
NM_001294332.1 NM_004168.3 XM_005248331.3 XM_011514072.1 XM_011514073.1 XM_017009685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1265106 c.428C>T p.T143M Chr.5: 225534-225534 on Build GRCh38
3013716 c.441C>T p.P147P Chr.5: 225547-225547 on Build GRCh38
6374247 c.407A>T p.Q136L Chr.5: 225513-225513 on Build GRCh38
3013717 c.444C>T p.A148A Chr.5: 225550-225550 on Build GRCh38
7423663 c.424A>T p.M142L Chr.5: 225530-225530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763191656 Chr.5: 225546-225546 on Build GRCh38
rs748175384 Chr.5: 225578-225578 on Build GRCh38
rs193283468 Chr.5: 225582-225582 on Build GRCh38
rs778737664 Chr.5: 225560-225560 on Build GRCh38
rs752187837 Chr.5: 225553-225553 on Build GRCh38
rs367618662 Chr.5: 225550-225550 on Build GRCh38
rs200565489 Chr.5: 225571-225571 on Build GRCh38
rs749893972 Chr.5: 225610-225610 on Build GRCh38
rs767097738 Chr.5: 225603-225603 on Build GRCh38
rs201453889 Chr.5: 225547-225547 on Build GRCh38
rs575617625 Chr.5: 225551-225551 on Build GRCh38
rs375576259 Chr.5: 225548-225548 on Build GRCh38
rs771236880 Chr.5: 225592-225592 on Build GRCh38
rs776848209 Chr.5: 225530-225530 on Build GRCh38
rs781436294 Chr.5: 225572-225572 on Build GRCh38
rs149101259 Chr.5: 225624-225624 on Build GRCh38
rs770457461 Chr.5: 225580-225580 on Build GRCh38
rs542980860 Chr.5: 225554-225554 on Build GRCh38
rs906281170 Chr.5: 225535-225535 on Build GRCh38
rs371735891 Chr.5: 225568-225568 on Build GRCh38
rs138917116 Chr.5: 225559-225559 on Build GRCh38
rs35464369 Chr.5: 225594-225594 on Build GRCh38
rs200675907 Chr.5: 225534-225534 on Build GRCh38
rs763490338 Chr.5: 225583-225583 on Build GRCh38
rs765784494 Chr.5: 225539-225539 on Build GRCh38
rs752206435 Chr.5: 225608-225608 on Build GRCh38
rs143148642 Chr.5: 225557-225557 on Build GRCh38
rs773634017 Chr.5: 225542-225542 on Build GRCh38

View Full Product Details